Protein enrichment encompasses numerous techniques to isolate cellular proteins based on their sequence, antibody epitope, post-translational modification (PTM) or active binding site. Protein enrichment is essential for isolating low-abundant proteins and/or reducing sample complexity for proteomic analysis. Enrichment of specific proteins or protein complexes can most easily be accomplished using immunoaffinity techniques such as immunoprecipitation, or through PTM specific affinity binding, such as phosphoprotein enrichment. The novel Thermo Scientific ActivX™ probes and enrichment kits enable the specific targeting of several enzyme classes, including kinases, GTPases, and serine hydrolases.
  • Comprehensive—reagents and kits for multiple protein enrichment strategies
  • Optimized—all reagents and kits are designed to maximize protein yield and reduce nonspecific binding
  • Convenient—easy-to-use spin or magnetic formats enable more rapid enrichment of proteins
  • Flexible—kits can be used with either cultured mammalian cells or tissues
  • Validated—all products have been fully tested and processed samples have been analyzed using Thermo Scientific Mass Spectrometers.
Choose the right protein enrichment products for your application
 Phosphoprotein
Enrichment Kit
Streptavidin IP-MS KitProtein A/G IP-MS KitKinase Enrichment Kit with ATP ProbeActivX TAMRA-FP Serine Hydrolase Probe
Typical research goalsDiscovery proteomicsTargeted proteomicsTargeted proteomicsDiscovery proteomicsDiscovery proteomics
TargetPhosphorylated proteinsSingle or homologous proteinsSingle or homologous proteinsATP binding sitesActive serine hydrolase enzymes
Binding/labeling mechanismMetal chelate affinity to phosphate groups
Biotinylated antibody affinity to target
Antibody affinity to targetBiotinylated ATP analog to active site lysineTAMRA-labeled fluorophosphonate binds to active site serine
Enrichment of targetPre-digestion (protein)Pre-digestion (protein)Pre-digestion (protein)Post digestion (peptide)Post digestion (peptide)
Enrichment formatIMACImmobilized StreptavidinImmobilized Protein A/GImmobilized StreptavidinImmobilized anti-TAMRA mAb
Base beadAgaroseMagneticMagneticAgaroseAgarose
 Order nowOrder nowOrder nowOrder nowOrder now
Automated Magnetic Separations for Proteomics
Magnetic beads and KingFisher Purification Systems enable the automation of immunoprecipitation (IP) and other affinity purification protocols for proteomics analysis by mass spectrometry.
Learn more
Featured data
 TargetA431HEK293
Anti-target AbNegative controlAnti-target AbNegative control
% Sequence
coverage
EGFR65%0%16%0%
AKT136%2%68%6%
AKT250%0%82%0%
AKT38%0%62%0%
PTEN16%0%36%0%
Enrichment of medium- to low-abundant EGFR-AKT pathway targets using Thermo Scientific Pierce Streptavidin Magnetic Beads for MS analysis. EGFR-AKT pathway targets were immunoprecipitated from two cell lines (A431 and HEK293) with biotinylated antibodies and captured with Pierce Streptavidin Magnetic Beads. IP eluates were digested in-solution, and analyzed by LC-MS/MS to assess sequence coverage and identify isoform-specific peptides.
Western blot and MS workflows for targeted capture and analysis of enzymes.
Western blot and MS workflows for targeted capture and analysis of enzymes. The schematic depicts two parallel workflows for the profiling, capture and detection of kinases, GTPases or serine hydrolases with ActivX Probes. Preincubation of enzymes with inhibitors allows for the determination of inhibitor specificity, binding affinity and potency by Western blot (or fluorescent SDS-PAGE for serine hydrolases only) of probe-labeled proteins or MS of probe-labeled peptides.
 TargetA431HEK293
Anti-target AbNegative controlAnti-target AbNegative control
% Sequence
coverage
EGFR65%0%16%0%
AKT136%2%68%6%
AKT250%0%82%0%
AKT38%0%62%0%
PTEN16%0%36%0%
Enrichment of medium- to low-abundant EGFR-AKT pathway targets using Thermo Scientific Pierce Streptavidin Magnetic Beads for MS analysis. EGFR-AKT pathway targets were immunoprecipitated from two cell lines (A431 and HEK293) with biotinylated antibodies and captured with Pierce Streptavidin Magnetic Beads. IP eluates were digested in-solution, and analyzed by LC-MS/MS to assess sequence coverage and identify isoform-specific peptides.
Western blot and MS workflows for targeted capture and analysis of enzymes.
Western blot and MS workflows for targeted capture and analysis of enzymes. The schematic depicts two parallel workflows for the profiling, capture and detection of kinases, GTPases or serine hydrolases with ActivX Probes. Preincubation of enzymes with inhibitors allows for the determination of inhibitor specificity, binding affinity and potency by Western blot (or fluorescent SDS-PAGE for serine hydrolases only) of probe-labeled proteins or MS of probe-labeled peptides.
Additional protein enrichment mass spec products
Catalog # Name Size Price (USD) Qty
88312 Pierce™ Kinase Enrichment Kit with ADP Probe Each
964.00
90407 Pierce™ Antibody Biotinylation Kit for IP Each
370.65
Online Exclusive
422.00
Save 51.35 (12%)
90410 Low Protein Binding Collection Tubes (1.5 mL), 250 tubes Each
33.65
Online Exclusive
38.51
Save 4.86 (13%)
90411 Low Protein Binding Collection Tubes (1.5 mL) Each
214.65
Online Exclusive
252.00
Save 37.35 (15%)
88311 ActivX™ Desthiobiotin-ATP Probe Each
467.00
88313 ActivX™ Desthiobiotin-ADP Probe Each
471.00
88315 ActivX™ Desthiobiotin-GTP Probe Each
404.00
88316 ActivX™ Azido-FP Serine Hydrolase Probe Each
604.00
88318 ActivX™ TAMRA-FP Serine Hydrolase Probe Each
529.65
Online Exclusive
594.00
Save 64.35 (11%)
MA1041 TAMRA Monoclonal Antibody (5G5) 100 µg
546.00
For Research Use Only. Not for use in diagnostic procedures.