Whole transcriptome analysis is of growing importance in understanding how altered expression of genetic variants contributes to complex diseases such as cancer, diabetes, and heart disease. Analysis of genome-wide differential RNA expression provides researchers with greater insights into biological pathways and molecular mechanisms that regulate cell fate, development, and disease progression.  With a dynamic range to detect subtle changes in expression level in a hypothesis neutral environment, next generation sequencing enables the understanding of biological response to stimuli or environmental changes.
Step-by-step guide to whole transcriptome analysis sample prep products
PureLink RNA Mini Kit
Recommended RNA purification kit for next generation sequencing applications.  Achieve high quality RNA from a wide arrange of cell and tissue types.
Learn more ›
Ordering Information
Catalog # Name Size Price (USD) Qty
12280050 PureLink™ Viral RNA/DNA Mini Kit Each
341.65
Online Exclusive
365.00
Save 23.35 (6%)
12280096A PureLink™ Pro 96 Viral RNA/DNA Purification Kit Each
2,055.00
RiboMinus RNA-Seq kits
Enrichment of the true whole transcriptome through selective depletion of 99.9% of ribosomal RNA.
Learn more ›
Ion Total RNA-Seq Kit v2
The Ion Total RNA-Seq Kit v2 provides a fast, simple workflow with as little as 100 ng of total RNA input, while maintaining strand orientation and minimizing bias and error.
Learn more ›
Ordering Information
Catalog # Name Size Price (USD) Qty
4479789 Ion Total RNA-Seq Kit v2, 48 reactions Each
7,640.00
4475936 Ion Total RNA-Seq Kit v2, 12 reactions Each
2,325.00
Ion Xpress RNA-Seq Barcode 1-16 Kit
Ion Xpress RNA-Seq Barcode 1-16 Kit provides a set of 16 unique barcode primers for sequencing RNA on the Ion Torrent PGM semiconductor sequencer. When used in combination with the Ion Total RNA-Seq Kit v2, the Ion Xpress RNA-Seq Barcode 1-16 Kit enables users to pool up to 16 fragment libraries prior to template preparation and then conduct multiplexed sequencing analysis, simplifying the Ion semiconductor sequencing workflow for small and whole transcriptome RNA.
Ordering Information
Catalog # Name Size Price (USD) Qty
4475485 Ion Xpress™ RNA-Seq Barcode 1-16 Kit Each
3,140.00
PureLink RNA Mini Kit
Recommended RNA purification kit for next generation sequencing applications.  Achieve high quality RNA from a wide arrange of cell and tissue types.
Learn more ›
Ordering Information
Catalog # Name Size Price (USD) Qty
12280050 PureLink™ Viral RNA/DNA Mini Kit Each
341.65
Online Exclusive
365.00
Save 23.35 (6%)
12280096A PureLink™ Pro 96 Viral RNA/DNA Purification Kit Each
2,055.00
RiboMinus RNA-Seq kits
Enrichment of the true whole transcriptome through selective depletion of 99.9% of ribosomal RNA.
Learn more ›
Ordering Information
Catalog # Name Size Price (USD) Qty
A1083708 RiboMinus™ Eukaryote Kit for RNA-Seq Each
946.00
A1083808 RiboMinus™ Plant Kit for RNA-Seq Each
842.00
Ion Total RNA-Seq Kit v2
The Ion Total RNA-Seq Kit v2 provides a fast, simple workflow with as little as 100 ng of total RNA input, while maintaining strand orientation and minimizing bias and error.
Learn more ›
Ordering Information
Catalog # Name Size Price (USD) Qty
4479789 Ion Total RNA-Seq Kit v2, 48 reactions Each
7,640.00
4475936 Ion Total RNA-Seq Kit v2, 12 reactions Each
2,325.00
Ion Xpress RNA-Seq Barcode 1-16 Kit
Ion Xpress RNA-Seq Barcode 1-16 Kit provides a set of 16 unique barcode primers for sequencing RNA on the Ion Torrent PGM semiconductor sequencer. When used in combination with the Ion Total RNA-Seq Kit v2, the Ion Xpress RNA-Seq Barcode 1-16 Kit enables users to pool up to 16 fragment libraries prior to template preparation and then conduct multiplexed sequencing analysis, simplifying the Ion semiconductor sequencing workflow for small and whole transcriptome RNA.
Ordering Information
Catalog # Name Size Price (USD) Qty
4475485 Ion Xpress™ RNA-Seq Barcode 1-16 Kit Each
3,140.00