HEK 293 Cells Using Lipofectamine® LTX Reagent
Introduction
Invitrogen™ Lipofectamine™ LTX Reagent is a proprietary, animal-origin free formulation for the transfection of DNA into eukaryotic cells with low cytotoxicity. This reference provides a recommended procedure to transfect plasmid DNA into HEK 293, human embryonic kidney cells (ATCC No. CRL-1573) using Lipofectamine LTX Reagent.
Important guidelines for transfection
Follow these important guidelines when transfecting HEK 293 cells using Lipofectamine LTX Reagent:
  • Maintain the same seeding conditions between experiments. Use low-passage cells; make sure cells are healthy and greater than 90% viable before transfection.
  • Transfection can be performed both in the presence or absence of serum. Test serum-free media for compatibility with Lipofectamine LTX Reagent.
  • We recommend Gibco™ Opti-MEM™ I Reduced Serum Medium (Cat. No. 31985-070) to dilute the DNA Lipofectamine LTX Reagent before complexing.
  • Visit www.lifetechnologies.com/genedelivery or contact Technical Services for other specialized transfection protocols.
  • Lipofectamine LTX Reagent performs well with vector-based RNAi experiments. For Invitrogen™ siRNA and Stealth RNAi™ transfections, we recommend Invitrogen™ Lipofectamine™ RNAiMAX Transfection Reagent. Go to www.lifetechnologies.com/RNAi or contact Technical Service for more information.
Materials needed
Have the following reagents on hand before beginning:
  • HEK 293 cells maintained in Gibco™ Dulbecco's Modified Eagle Medium (DMEM) (Cat. No. 11960-044) supplemented with 4 mM Gibco™ L-Glutamine (Cat. No. 25030-081), 10% Gibco™ Fetal Bovine Serum (Cat No. 16000-044). Grow cells at 37o C with 5% CO2.
  • Plasmid DNA of interest.
  • Lipofectamine LTX Reagent (store at +4oC until ready to use)
  • Opti-MEM I Reduced Serum Media
  • Appropriate tissue culture plates and supplies
Ordering Information
Catalog # Name Size Price (USD) Qty
31985070 Opti-MEM™ I Reduced Serum Medium, 500 mL Each
72.65
Online Exclusive
78.75
Save 6.10 (8%)
11960044 DMEM, high glucose, no glutamine, 500 mL Each
34.65
Online Exclusive
37.64
Save 2.99 (8%)
25030081 L-Glutamine (200 mM), 100 mL Each
37.65
Online Exclusive
40.88
Save 3.23 (8%)
16000044 Fetal Bovine Serum, certified, United States, 500 mL Each -
Transfecting HEK 293 cells
Use this procedure to transfect plasmid DNA into HEK 293 cells in a 24-well format (for other formats, see Scaling up or down transfections, below). All amounts and volumes are given on a per well basis.
  1. The day before transfection, trypsinize and count the cells. Plate 0.5 -1.25x105 cells per well in 0.5 mL of complete growth medium. Cell density should be 50–80% confluent on the day of transfection.
  2. (Optional) The day of transfection, remove growth medium from cells and replace with 0.5 mL of complete growth medium.
  3. For each well of cells to be transfected, dilute 0.5 μg of DNA in 100 μL of Opti-MEM I Reduced Serum Media without serum.
  4. For each well of cells, add 0.75-1.75 μL of Lipofectamine LTX Reagent into the above diluted Opti-MEM:DNA solution, mix gently and incubate 30 minutes at room temperature to form DNA- Lipofectamine LTX Reagent complexes.
  5. After 30 minute incubation, add 100 μl of the DNA- Lipofectamine LTX Reagent complexes directly to each well containing cells and mix gently by rocking the plate back and forth.
  6. Complexes do not have to be removed following transfection. Incubate the cells at 37oC in a CO2 incubator for 18–24 hours post-transfection before assaying for transgene expression.
TOP
Scaling Up or Down Transfections
Culture vesselSurface
area per
well
Volume plating mediumCells per wellVolume
dilution
medium
DNALipofectamine
LTX Reagent
96-well0.3 cm2100 μl2.5 x 10420 μl100 ng0.15 - 0.35 μl
48-well1 cm2200 μl5 x 10440 μl200 ng0.30 - 0.7 μl
24-well2 cm2500 μl1.25 x 105100 μl500 ng0.75 - 1.75  μl
12-well4 cm21 ml2.5 x 105200 μl1 μg1.5 - 3.5 μl
6-well10 cm2 2 ml6.25 x 105500 μl2.5 μg3.75 - 8.75 μl
25-0989W   17  Nov  2006