What is allergy cross-reactivity? 
Allergy cross-reactivity occurs when the body's immune system identifies the proteins in one substance (e.g., pollen) and the proteins in another (e.g., a fruit or vegetable) as being similar.1  When an individual comes into contact with either, whether or not it’s a protein in something that he or she is truly allergic to, the immune system can react in the same way, which in some cases can cause allergic symptoms.  
All that matters to the immune system is that the proteins are structurally similar or biologically related. If they are, cross-reactivity can occur. The risk of allergy cross-reactivity needs to be assessed by a healthcare professional, and important foods should never be removed from a diet without a confirmed diagnosis. The potential for cross-reactivity can make diagnosing specific allergies somewhat complicated.2
Symptoms that have manifested as a result of cross-reacting antibodies (i.e., proteins produced by the immune system to fight disease) cannot always be distinguished from those caused by genuine sensitization. However, by determining the primary sensitization, treatment or avoidance of symptoms can be effective. 
What is Pollen Food Allergy Syndrome (PFAS)? 
Pollen Food Allergy Syndrome (PFAS), also known as Oral Allergy Syndrome (OAS), is an allergic reaction that typically occurs after a patient who is sensitive to pollen eats certain foods, e.g., fruits, vegetables, nuts, and grains.PFAS is possibly the most common food allergy in adults, with up to 60 percent of patients allergic to pollen being affected.3  Additionally, up to 25 percent of children with allergic rhinitis (i.e., hay fever) also suffer from OAS. 4
If you're sensitized to pollen, you may experience PFAS when certain foods touch your oral mucosa, lips, tongue, or throat. The symptoms of OAS appear within minutes of eating a raw form of the food but can last for hours. In the majority of cases, the responsible allergens are rapidly denatured by cooking and digestion, though certain patients can still present with systemic reactions.3  
Common PFAS or OAS symptoms include: 
  • Scratchy throat 
  • Itchy mouth or lips  
  • Swollen lips, tongue, or throat  
These symptoms could become worse during times of the year when pollen counts are high (usually in spring or fall).  
Common Allergy Cross-Reactivity Between Pollen and Foods
People with Oral Allergy Syndrome (OAS) typically have an allergy to birch trees, ragweed, or grass pollen.5 Foods that may cause OAS include tree nutssoypeanutfruits, and vegetables.5
Pollen
Fruits
Vegetables
Nuts
Grains
Birch 
Kiwi, apple, pear, plum, peach, nectarine, apricot, cherry, banana, fig, avocado, strawberry, dried plum, mango 
Celery, carrot, parsnip, parsley, dill, cumin, cilantro, fennel, potato, tomato, pepper (green), chicory
Hazelnut, almond, walnut, peanut  
Soy, wheat, lentils, peas, beans 
Alder 
Apple, cherry, peach, pear, strawberry, raspberry
Celery, parsley
Hazelnut, almond, walnut 
Grass
Fig, melon, orange, kiwi, watermelon 
Tomato, potato 
Peanut 
Mugwort 
Mango 
Celery, carrot 
Ragweed
Banana,  watermelon, melon,  honeydew, cantaloupe 
Squash, pepper, cucumber, artichoke, hibiscus, zucchini,  chamomile  tea 
Sunflower seeds  
Other Cross-Reactivity Considerations
Cross-reactivity between shellfish:1 There is a high degree of cross-reactivity among crustacean shellfish (e.g., shrimp, lobster, crab, crawfish). The risk may be lower for cross-reactivity between crustacean shellfish and noncrustacean shellfish (mollusks) such as clams, oysters, scallops, and mussels. Of note, there is cross-reactivity between chitins (a component of the exoskeleton of shellfish and insects) in dust mites and crustacean shellfish in the form of a cross-reactive risk protein called tropomyosin. 
Cross-reactivity between latex and food:1 Approximately 30 to 70 percent of people with IgE-mediated allergies to latex can experience symptoms with any of several fruits cross-reactive to latex, including most commonly banana, avocado, kiwi, and chestnut. Several types of proteins have been identified to be involved in Latex-Fruit Syndrome.
Cross-reactivity between peanut and tree nuts or seeds: Peanut and tree nut allergies are often characterized by life-threatening anaphylactic reactions and lifelong persistence. While peanut allergy is more commonly seen in patients, many subjects develop a hypersensitivity to both peanuts and tree nuts.6 Although peanuts are actually legumes, approximately 20 to 30 percent of those with a peanut allergy are also allergic to one or more types of tree nuts.7  Additionally, walnut, pecan, and hazelnut comprise a group of strongly cross-reactive tree nuts. Hazelnut, cashew, Brazil nut, pistachio, and almond form another group of moderately cross-reactive tree nuts.8 
What is cross-contact? 
Cross-contact happens when a small amount of allergen gets into a food that it shouldn’t be in. Some food labels include “may contain” or “processed in the same facility as”  identifiers due to cross-contact. When it comes to food allergies, it’s important to understand the difference between cross-contact and cross-reactivity to avoid confusion and/or an unnecessary reaction. 
Find Answers 
Knowing if you’re allergic and what you’re allergic to can help you avoid or minimize symptoms when it comes to cross-reactivity and Pollen Food Allergy Syndrome. A blood test—together with an allergy-focused medical history—may help identify underlying allergen triggers. 
  1. Matricardi PM, Kleine-Tebbe J, Hoffmann HJ, Valenta R, Hilger C, Hofmaier S, Aalberse RC, Agache I, Asero R, Ballmer-Weber B, Barber D, Beyer K, Biedermann T, Bilò MB, Blank S, Bohle B, Bosshard PP, Breiteneder H, Brough HA, Caraballo L, Caubet JC, Crameri R, Davies JM, Douladiris N, Ebisawa M, EIgenmann PA, Fernandez-Rivas M, Ferreira F, Gadermaier G, Glatz M, Hamilton RG, Hawranek T, Hellings P, Hoffmann-Sommergruber K, Jakob T, Jappe U, Jutel M, Kamath SD, Knol EF, Korosec P, Kuehn A, Lack G, Lopata AL, Mäkelä M, Morisset M, Niederberger V, Nowak-Węgrzyn AH, Papadopoulos NG, Pastorello EA, Pauli G, Platts-Mills T, Posa D, Poulsen LK, Raulf M, Sastre J, Scala E, Schmid JM, Schmid-Grendelmeier P, van Hage M, van Ree R, Vieths S, Weber R, Wickman M, Muraro A, Ollert M. EAACI Molecular Allergology User's Guide. Pediatr Allergy Immunol. 2016 May;27 Suppl 23:1-250. doi: 10.1111/pai.12563. PMID: 27288833. 
  2. American Academy of Allergy, Asthma & Immunology [Internet]. Milwaukee, WI: American Academy of Allergy, Asthma & Immunology; 2021. Available from: https://www.aaaai.org/tools-for-the-public/allergy,-asthma-immunology-glossary/cross-reactivity-defined.  
  3. Kashyap RR, Kashyap RS. Oral Allergy Syndrome: An Update for Stomatologists. J Allergy. 2015;2015:543928. 
  4. Roberts, Graham & Xatzipsalti, M & Borrego, Luis & Custovic, Adnan & Halken, Susanne & Hellings, Peter & Papadopoulos, Nikolaos & Rotiroti, G & Scadding, Glenis & Timmermans, Frans & Valovirta, Erkka. (2013). Pediatric rhinitis: Position paper of the European Academy of Allergy and Clinical Immunology. Allergy. 68. 10.1111/all.12235. 
  5. American College of Allergy, Asthma & Immunology [Internet]. Arlington Heights, IL: American College of Allergy, Asthma & Immunology; 2019 Mar 21. 
  6.  De Leon MP, Glaspole IN, Drew AC, Rolland JM, O'Hehir RE, Suphioglu C. Immunological analysis of allergenic cross-reactivity between peanut and tree nuts. Clin Exp Allergy. 2003 Sep;33(9):1273-80. doi: 10.1046/j.1365-2222.2003.01761.x. PMID: 12956750. 
  7.  McWilliam V, Koplin J, Lodge C, Tang M, Dharmage S, Allen K. The Prevalence of Tree Nut Allergy: A Systematic Review. Curr Allergy Asthma Rep. 2015 Sep;15(9):54. doi: 10.1007/s11882-015-0555-8. PMID: 26233427. 
  8. Goetz DW, Whisman BA, Goetz AD. Cross-reactivity among edible nuts: double immunodiffusion, crossed immunoelectrophoresis, and human specific igE serologic surveys. Ann Allergy Asthma Immunol. 2005 Jul;95(1):45-52. doi: 10.1016/S1081-1206(10)61187-8. PMID: 16095141.