showcase.par.86277.image.0.0.1
Yeast culture for maintaining and propagating various strains, such as Saccharomyces cerevisiae (S. cerevisiae) and Pichia pastoris (P. pastoris), requires specific formulations of complex media for use in cloning and protein expression. We offer a selection of yeast growth media for your specific application in both powder and ready-to-use liquid formats for convenience and ease of use.
Choose your type of yeast growth media:
PichiaPink Media
Pre-packaged media pouches for use with PichiaPink Yeast Expression System, containing media to grow P. pastoris yeast strains for transformation and selection
CSM Media
Specialty media for making agar plates to grow S. cerevisiae (MaV203) competent cells
YPD BrothCommonly used yeast media for maintenance and propagation of P. pastoris and S. cerevisiae yeast strains.
Yeast Nitrogen Base (YNB)Base medium for preparation of minimal and synthetic defined yeast media
Ordering Information
Catalog # Name Size Price (USD) Qty
A1374501 YPD Broth Each
57.65
Online Exclusive
60.00
Save 2.35 (4%)
Q30007 Yeast Nitrogen Base Each
268.00
A11156 PichiaPink™ Media Kit Each
1,196.00
Q30009 Yeast Nitrogen Base Each
956.00
A13292 CSM Media for Mav203 Yeast Cells Each
433.00