Real-time PCR master mixes for gene expression analysis
Applied Biosystems qPCR and RT-qPCR master mixes are engineered to deliver exceptional specificity and reliability for your gene expression analysis. Broadly compatible and supported by trusted expertise, our master mixes are ideal for applications in infectious disease research, cancer research, biomarker discovery, and pharmacogenomics. Whether you need single-target SYBR green dye-based detection or advanced TaqMan probe-based multiplexing, our reagents enable fast, accurate, and reproducible results across a wide range of target concentrations. Accelerate your path to discovery with the accuracy and consistency you can trust.
Compare SYBR Green vs TaqMan chemistries for gene expression analysis
Graphic of female scientist over blue background
PCR and RT-qPCR master mixes for gene expression
For optimal performance, we recommended using one of the following qPCR master mixes for gene expression analysis.
TaqMan Fast Advanced Master Mix
TaqMan Fast Virus 1-Step master mixes
PowerTrack SYBR Green Master Mix*
Overview Bench-top stable reagent with robust performance in singleplex or duplex reactions Highly sensitive, inhibitor-tolerant reagents capable of 4-target multiplex Universal 2X reagent master mix  with built-in visual indicators for accurate reaction setup
Chemistry
Probes, 2-step qPCR
Probes, 1-step RT-qPCR
SYBR Green dye, 2-step qPCR
Instrument compatibility Any qPCR instrument, from any manufacturer
Target
cDNA
RNA
cDNA
Concentration
2X
4X
2X
Multiplexing and passive reference dye
With ROX:  up to 2 targets
Available in 2 versions:
With ROX: up to 3 targets
No ROX: 3 or more targets 
With ROX: 1 target
Cycling mode: run time
Fast:  ~40 min
Standard: ~50 min
Fast: ~60 min
Standard: ~90 min
Fast: ~30 min
Standard: ~60 min
Carryover contamination control
2 versions:
With UNG
No UNG
No UNG
With UNG
Sample tracking dye No No Yes
Reverse transcriptase
SuperScript IV VILO Master Mix
Thermostable MMLV RT included in master mix
SuperScript IV VILO Master Mix
Assay recommendations
TaqMan Gene Expression assays
TaqMan Gene Expression assays
qPCR primers
Product page
Check price or order now
Check price or order now
Check price or order now
* For SYBR Green detection: when you need up to 72 hours of benchtop stability, choose PowerUp SYBR Green Master Mix in combination with High-Capacity RNA-to-cDNA Kit Reverse Transcriptase.
Reagents for diagnostic testing, bioanalysis, and clinical trial assay development
For biomarkers or drug targets moving beyond research and development, we offer master mixes that scale with growing study size and complexity. TaqPath general purpose reagents are made in ISO 13485-certified and FDA-registered facilities, following cGMP principles to enable lot-to-lot reproducibility and consistent Ct values. Maintain confidence in the precision of your gene expression data, while driving efficiency with master mixes that are optimized for high-throughput workflows and multiplex analysis
TaqPath qPCR general purpose reagents
Other real-time PCR master mixes
Our master mixes have long supported researchers with their qPCR gene expression analysis. However, for optimal results we recommend upgrading to one of our newer master mixes listed in the table above, which offer superior performance.
Legacy TaqMan Master Mixes include:
Legacy SYBR Green master mixes include:
Complementary products for gene expression analysis
Elevate qPCR performance and eliminate uncertainty with our high-performance products for every step of your workflow. Designed to work seamlessly with our master mixes, our sample preparation kits, reverse transcription reagents, and TaqMan Assays ensure high-quality results every time. Achieve reliable qPCR results with our comprehensive solutions
TaqMan Predesigned Gene Expression Assays
Choose from over 2.8 million predesigned assays covering more than 30 species to detect virtually any gene transcript with reliable precision.  
SuperScript VILO Master Mix for two-step RT-qPCR
SuperScript VILO Master Mix is a first-strand cDNA synthesis mix featuring Superscript IV Reverse Transcriptase for reliable synthesis and high cDNA yields.
Specificity in gene expression analysis
Target specificity is a critical and sometimes challenging aspect of gene expression analysis. Without it, your data can be compromised, leading to inaccurate results and potentially flawed conclusions. Before running your next experiment, read our white paper and infographic to learn why specificity can be challenging in gene expression analysis. Review the role of qPCR detection chemistry in assay specificity and discover key insights to help you achieve reliably accurate results.
Educational Resources for Gene Expression Analysis
For Research Use Only. Not for use in diagnostic procedures.
*For Laboratory Use.