cb12 - Apps/Samples/Products/Resources/Documents/Contact Us
As the demand for high-performance materials increases, so does the importance of surface engineering. The material’s surface is its point of interaction with the external environment and other materials, and will influence factors such as corrosion rates, catalytic activity, adhesive properties, wettability, contact potential, and failure mechanisms.
Surface modification can be used to alter or improve these characteristics; surface analysis is used to understand surface chemistry and investigate the efficacy of surface engineering. From non-stick cookware coatings to thin-film electronics and bio-active surfaces, X-ray photoelectron spectroscopy is one of the standard tools for surface characterization.
X-ray photoelectron spectroscopy (XPS), also known as electron spectroscopy for chemical analysis (ESCA), is a technique for analyzing a material’s surface chemistry. XPS can measure elemental composition as well as the chemical and electronic state of the atoms within a material.
Animation of XPS surface analysis.
XPS spectra are obtained by irradiating a solid surface with a beam of X-rays and measuring the kinetic energy of electrons that are emitted from the top 1-10 nm of the material. A photoelectron spectrum is recorded by counting ejected electrons over a range of kinetic energies. The energies and intensities of the photoelectron peaks enable identification and quantification of all surface elements (except hydrogen).
Surface characterization
The surface represents a discontinuity between one phase and another; the physical and chemical properties of the surface are, therefore, different from those of the bulk material. These differences affect the topmost atomic layer of the material to a large extent because a surface atom is not surrounded by atoms on all sides. This results in the surface atom having a bonding potential, which makes it more reactive than atoms in the bulk.
Diagram of surface layers that can be analyzed with XPS surface analysis.
A surface layer is defined as being up to three atomic layers thick (~1 nm), depending upon the material. Layers up to approximately 10 nm are considered ultra-thin films, and layers up to approximately 1 μm are thin films. The remainder of the solid is referred to as bulk material. This terminology is not definitive however, and the distinction between the layer types can vary depending upon the material and its application.
Single-instrument, multi-technique XPS workflow
When you look at your sample, do you wonder which analysis technique will get you all the pertinent information you need? Is the answer usually that no one single technique will give you everything you need? To fully understand materials, you need to be able to analyze them using multiple techniques. When you use a single-instrument, multi-technique workflow, you can better expose your sample’s properties by exploring it with a combination of many techniques, including XPS, ISS, REELS, UPS, and Raman. Learn how you can get on the fast track to comprehensive surface analysis.
Learn more
Surface Properties
Surface analysis contributes to the understanding of each of these areas:
  • Corrosion
  • Welding
  • Fatigue
  • Grain boundary segregation
  • Glass
  • Coatings
  • Motor/Avionics
  • Lubrication
  • Corrosion
  • Oxidation
  • Fatigue/Failure
  • Fiber Composites
  • Adhesives
  • Semiconductor/ Microelectronics
  • Microcircuits
  • Ultra-thin Films
  • Soldering
  • Cleaning
  • Thin-film Stability
  • Barrier Layers
  • Lubrication
  • Chemical Industry
  • Plastics/Coatings
  • Catalysis
  • Fibers
  • Metal/Steel Industry
Diagram of information found in atomic layers with XPS surface analysis.
Common properties and processes as a function of material depth or thickness.
Periodic Table
Explore our information-packed knowledge base of elemental properties and XPS analysis.
Analysis Features
Explore the many applications of XPS analysis.
Testimonials – Solving materials problems with XPS analysis
50 years of XPS
Discover how XPS has impacted research over the last 50 years.
Video Player is loading.
Current Time 0:00
Duration 7:16
Loaded: 2.27%
Stream Type LIVE
Remaining Time 7:16
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
      1. Now Playing
        Up Next50 years of XPS
        Discover how XPS has impacted research over the last 50 years.
      2. Now Playing
        Up NextXPS - John Watts, Professor of Materials Science, University of Surrey
        Professor John Watts goes over how to solve materials problems with XPS.
      3. Now Playing
        Up NextXPS Overview with XPS Facility Manager Robert Palgrave
        Mr. Palgrave of University College London goes over analyzing solid-state and thin film materials using XPS.
    Resources
    The Surface Analysis Toolbox Combining XPS with ISS, REELS, AES, UPS, and Raman in a workflow
    This webinar will introduce these additional techniques and show how they can be used with XPS to build comprehensive understanding of the surfaces of materials.
    Video Player is loading.
    Current Time 0:00
    Duration 59:43
    Loaded: 0.00%
    Stream Type LIVE
    Remaining Time 59:43
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
        1. Now Playing
          Up NextThe Surface Analysis Toolbox Combining XPS with ISS, REELS, AES, UPS, and Raman in a workflow
          This webinar will introduce these additional techniques and show how they can be used with XPS to build comprehensive understanding of the surfaces of materials.
        2. Now Playing
          Up NextSurface Analysis The Basics & Beyond session (part 1) of the Ignite Surface Analysis Virtual Scientific Conference
          The speakers Tim Nunney (Thermo Fisher Scientific), Raheleh Azmi (KIT) and Dr Vinod C. Prabhakaran (NCL) give presentations on the basics of XPS.
        3. Now Playing
          Up NextMulti-technique and Complementary Analysis session (part 2) of the Ignite Surface Analysis Virtual Scientific Conference
          The speakers Roland Barbosa (Covalent Metrology), Paul Mack and Herman Lemmens (Thermo Fisher Scientific) give presentations on multi-technique and complementary analysis.
        4. Now Playing
          Up NextAnalysis of Materials for Advanced Applications (part 3) of the Ignite Surface Analysis Virtual Scientific Conference
          The speakers Damien Aureau (UVSQ), Robert Palgrave (UCL) and Albert Ge (Thermo Fisher Scientific) give presentations on analysis of materials for advanced applications.
        5. Now Playing
          Up NextApplications Q&APanel discussion (part 4) of the Ignite Surface Analysis Virtual Scientific Conference
          The speakers Tim Nunney, Paul Mack, Robin Simpson, Hsiang-Han Tseng, Stuart Blackburn (Thermo Fisher Scientific) answer applications questions.
        6. Now Playing
          Up NextThermo Fisher Scientific Avantage Datasystem
          The XPS knowledge view is a powerful reference tool included within Avantage Datasystem
        7. Now Playing
          Up NextXPS Depth profiling
          Depth profiling is a common experiment using XPS surface analysis instrumentation. This presentation explains how a depth profile is set up, the types of equipment available to work with different types of sample, and is illustrated with example anal
        8. Now Playing
          Up NextThe Basics of XPS Surface Analysis, Part 1
          This presentation gives an introduction to X-ray photoelectron spectroscopy, a key technique for understanding materials. The first part of this two part series describes the fundamentals of the technique, the instrumentation, and is illustrated with
        9. Now Playing
          Up NextThe Basics of XPS Surface Analysis, Part 2
          The second part of this two part presentation shows how X-ray photoelectron spectroscopy can solve a problem by walking through a complete analysis of a sample, with tips on how to successfully collect and process XPS data.
        10. Now Playing
          Up NextThermo Scientific Magcis Dual Beam Ion Source
          The Thermo Scientific MAGCIS dual mode ion source enables depth profiling analysis and surface cleaning of both soft and hard materials on the same XPS instrument.
      Webinars
      Understanding XPS images and depth profiles with Avantage Software - Part 2
      By watching the webinar, you will learn how to use the right tools to understand multi-level data sets, such as depth profiles and images.
      Understanding Surface Chemistry with Avantage Software
      This webinar is designed to offer training to current users of Avantage software and act as an introduction to those unfamiliar with it.
      Surface Analysis of Thin Films
      Explore a number of applications for thin film coating analysis including forensic studies, graphene, multi-layered glass, coated fabrics and photovoltaics.
      Understanding Metal Surfaces and Oxides
      XPS delivers chemical state information from the topmost nanometers of a sample surface, enabling you to measure passivation coatings, understand catalyst chemistries, and develop bio-compatibility coatings.
      Multi-technique Surface Analysis and Cluster Ion Sample Cleaning
      We present how a dual mode ion source can be used for multi-technique sample analysis, showing the importance of sample cleaning and how depth profiles can be performed.
      Characterizing Polymers with XPS
      This webinar covers the basics of X-ray photoelectron spectroscopy, with a special emphasis on how it can be used in the field of polymer surface analysis. X-ray photoelectron spectroscopy is a powerful technique for the chemical analysis of the surface of materials.
      The Surface Analysis Toolbox Combining XPS with ISS, REELS, AES, UPS, and Raman in a workflow
      This webinar will introduce these additional techniques and show how they can be used with XPS to build comprehensive understanding of the surfaces of materials.
      Video Player is loading.
      Current Time 0:00
      Duration 59:43
      Loaded: 0.28%
      Stream Type LIVE
      Remaining Time 59:43
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
          1. Now Playing
            Up NextThe Surface Analysis Toolbox Combining XPS with ISS, REELS, AES, UPS, and Raman in a workflow
            This webinar will introduce these additional techniques and show how they can be used with XPS to build comprehensive understanding of the surfaces of materials.
          2. Now Playing
            Up NextSurface Analysis The Basics & Beyond session (part 1) of the Ignite Surface Analysis Virtual Scientific Conference
            The speakers Tim Nunney (Thermo Fisher Scientific), Raheleh Azmi (KIT) and Dr Vinod C. Prabhakaran (NCL) give presentations on the basics of XPS.
          3. Now Playing
            Up NextMulti-technique and Complementary Analysis session (part 2) of the Ignite Surface Analysis Virtual Scientific Conference
            The speakers Roland Barbosa (Covalent Metrology), Paul Mack and Herman Lemmens (Thermo Fisher Scientific) give presentations on multi-technique and complementary analysis.
          4. Now Playing
            Up NextAnalysis of Materials for Advanced Applications (part 3) of the Ignite Surface Analysis Virtual Scientific Conference
            The speakers Damien Aureau (UVSQ), Robert Palgrave (UCL) and Albert Ge (Thermo Fisher Scientific) give presentations on analysis of materials for advanced applications.
          5. Now Playing
            Up NextApplications Q&APanel discussion (part 4) of the Ignite Surface Analysis Virtual Scientific Conference
            The speakers Tim Nunney, Paul Mack, Robin Simpson, Hsiang-Han Tseng, Stuart Blackburn (Thermo Fisher Scientific) answer applications questions.
          6. Now Playing
            Up NextThermo Fisher Scientific Avantage Datasystem
            The XPS knowledge view is a powerful reference tool included within Avantage Datasystem
          7. Now Playing
            Up NextXPS Depth profiling
            Depth profiling is a common experiment using XPS surface analysis instrumentation. This presentation explains how a depth profile is set up, the types of equipment available to work with different types of sample, and is illustrated with example anal
          8. Now Playing
            Up NextThe Basics of XPS Surface Analysis, Part 1
            This presentation gives an introduction to X-ray photoelectron spectroscopy, a key technique for understanding materials. The first part of this two part series describes the fundamentals of the technique, the instrumentation, and is illustrated with
          9. Now Playing
            Up NextThe Basics of XPS Surface Analysis, Part 2
            The second part of this two part presentation shows how X-ray photoelectron spectroscopy can solve a problem by walking through a complete analysis of a sample, with tips on how to successfully collect and process XPS data.
          10. Now Playing
            Up NextThermo Scientific Magcis Dual Beam Ion Source
            The Thermo Scientific MAGCIS dual mode ion source enables depth profiling analysis and surface cleaning of both soft and hard materials on the same XPS instrument.
        Webinars
        Understanding XPS images and depth profiles with Avantage Software - Part 2
        By watching the webinar, you will learn how to use the right tools to understand multi-level data sets, such as depth profiles and images.
        Understanding Surface Chemistry with Avantage Software
        This webinar is designed to offer training to current users of Avantage software and act as an introduction to those unfamiliar with it.
        Surface Analysis of Thin Films
        Explore a number of applications for thin film coating analysis including forensic studies, graphene, multi-layered glass, coated fabrics and photovoltaics.
        Understanding Metal Surfaces and Oxides
        XPS delivers chemical state information from the topmost nanometers of a sample surface, enabling you to measure passivation coatings, understand catalyst chemistries, and develop bio-compatibility coatings.
        Multi-technique Surface Analysis and Cluster Ion Sample Cleaning
        We present how a dual mode ion source can be used for multi-technique sample analysis, showing the importance of sample cleaning and how depth profiles can be performed.
        Characterizing Polymers with XPS
        This webinar covers the basics of X-ray photoelectron spectroscopy, with a special emphasis on how it can be used in the field of polymer surface analysis. X-ray photoelectron spectroscopy is a powerful technique for the chemical analysis of the surface of materials.
        Applications
        Process control using electron microscopy
        Process control using electron microscopy
        Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
        Quality control and failure analysis using electron microscopy
        Quality control and failure analysis
        Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
        Fundamental Materials Research_R&D_Thumb_274x180_144DPI
        Fundamental Materials Research
        Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
        Aluminum mineral grain found with SEM during parts cleanliness testing
        Technical Cleanliness
        More than ever, modern manufacturing necessitates reliable, quality components. With scanning electron microscopy, parts cleanliness analysis can be brought inhouse, providing you with a broad range of analytical data and shortening your production cycle.
        Samples
        Battery Research
        Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
        Learn more ›
        Metals Research and Characterization
        Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
        Learn more ›
        Geological Research
        Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
        Learn more ›
        Oil and Gas
        As the demand for oil and gas continues, there is an ongoing need for efficient and effective extraction of hydrocarbons. Thermo Fisher Scientific offers a range of microscopy and spectroscopy solutions for a variety of petroleum science applications.
        Learn more ›
        Nanoparticles
        Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
        Learn more ›
        Forensics
        Micro-traces of crime scene evidence can be analyzed and compared using electron microscopy as part of a forensic investigation. Compatible samples include glass and paint fragments, tool marks, drugs, explosives, and GSR (gunshot residue).
        Learn more ›
        Catalysis Research
        Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
        Learn more ›
        Fibers and Filters
        The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
        Learn more ›
        2D Materials
        Novel materials research is increasingly interested in the structure of low-dimensional materials. Scanning transmission electron microscopy with probe correction and monochromation allows for high-resolution two-dimensional materials imaging.
        Learn more ›
        Automotive Materials Testing
        Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
        Learn more ›
        Products
        Style Sheet for Instrument Cards Original
        K-Alpha XPS
        • High resolution XPS
        • Fast, efficient, automated workflow
        • Ion source for depth profiling
        Nexsa G2 XPS
        • Micro-focus X-ray sources
        • Unique multi-technique options
        • Dual-mode ion source for monoatomic & cluster ion depth profiling
        ESCALAB QXi XPS
        • High spectral resolution
        • Multi-technique surface analysis
        • Extensive sample preparation and expansion options
        Documents
        Style Sheet for Komodo Tabs
        Style Sheet to change H2 style to p with em-h2-header class
        Contact us
        Style Sheet for Support and Service footer
        Style Sheet for Fonts
        Style Sheet for Cards
        Electron microscopy services for
        the materials science
        To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.