Organism TypeHumanA small part of the thoracic aorta was dissected from human donor and recipient hearts during heart transplantation. The adventitial layer was dissected from the aorta and cells were isolated with the use of collagenase. The cells that attached were cultured in DMEM, supplemented with L-glutamine, 100 U/mL penicillin, 100 ug/mL streptomycin, and 10% fetal bovine serum.
TissuesAorta
PhenotypeAdherent
Primaryyes
ApplicationCell Culture / Growth Conditions
Products
Catalog # Name Size Price (USD) Qty
11965092 DMEM, high glucose, 500 mL Each
34.65
Online Exclusive
37.14
Save 2.49 (7%)
15070063 Penicillin-Streptomycin (5,000 U/mL) Each
26.65
Online Exclusive
28.76
Save 2.11 (7%)
25030081 L-Glutamine (200 mM), 100 mL Each
37.65
Online Exclusive
40.88
Save 3.23 (8%)
25030149 L-Glutamine (200 mM), 20 mL Each
13.65
Online Exclusive
14.67
Save 1.02 (7%)
Every step of the way, a wide range of cell health products
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.