Food Profiling and Authenticity
Testing food and feed authenticity for consumer and public health protection
In today's global economy, a food product or its ingredients may be sourced from a single farm or from producers and manufacturers worldwide. Truth-in-labeling regulations are enforced to verify the authenticity and origins of these products as they are exported around the globe. Recurring food contamination scandals have shown that constant monitoring is required to prevent counterfeiting and intentional or unintentional adulteration.
Next generation food screening and identification requires an expanded toolset to address the increase in food fraud cases and ensure food authenticity.
Contact us  Food analysis brochure  
On this page
Download our three-part eBook series
This three-part application eBook series illustrates how our chromatography and mass spectrometry solutions can be used to improve analytical testing in three important application areas: food authenticity, veterinary drug residues and QA/QC product testing.
Download eBooks ›
Routine food testing and forensic investigations benefit from a variety of food profiling techniques. Genomic approaches utilize nuclear or mitochondrial DNA to generate genetic profiles. Proteomics characterizes food and food products through the analysis of proteins and peptides. Metabolomics provides deep insights into the overall composition of food products, while isotopolomics can provide the analysis of isotopic ratios and rare earth elements.
Food profiling techniques
By analyzing food products with these various food profiling techniques, databases can be generated to easily identify quality parameters like geographic origin, production type, variety and many more.
Fingerprinting and origin using isotopic ratio analysis
Because food fraud is a problem at every stage of the food and beverage supply chain and consumers are increasingly concerned about food safety, quality, and sourcing; product labels should be accurate. To protect the reputations of your company and clients and encourage customer loyalty, food fingerprinting and isotope analysis can be used to determine the authenticity of ingredients.
Isotope fingerprint in food and beverage products is region or process specific
Isotope fingerprintWhat is the biogeochemical interpretation?What is an example of food fraud interpretation?What products can be affected?
CarbonBotanical origin
(C3, C4 and CAM photosynthesis)
Adulteration
(e.g. sweetening with cheap sugar)
Honey, liquor, wine, olive oil, butter, flavors
NitrogenSoil processes, plant fertilizer processesMislabeling
(Differentiate organic and non-organic)
Fruits and vegetables, animal meat
SulfurLocal soil conditions, proximity to shorelineOrigin of productFruits and vegetables, animal meat, honey
OxygenPrincipally related to local-regional rainfall and hence geographical areaWatering of beverages, place of origin of productCoffee, wine, liquor, water, sugar, animal meat, flavors
HydrogenRelated to local-regional rainfall and hence geographical areaWatering of beverages, origin of productCoffee, wine, liquor, water, sugar, animal meat, flavors
Configuration/Use case
These processes can be traced using carbon, nitrogen, sulfur, oxygen and hydrogen isotopes, indicating the origin and history of food and beverage products.
Instruments and SoftwareVarious configurations available
Gas Isotope Ratio Mass Spectrometry (IRMS)
Qtegra ISDS Software for gas IRMS
H, C, N, O, S
AccessoriesTypical use case
Thermo Scientific EA IsoLink IRMS SystemAnalyze bulk samples
Thermo Scientific GC IsoLink II IRMS SystemAnalyze volatile compounds
Thermo Scientific LC IsoLink II IRMS SystemAnalyze polar compounds
Thermo Scientific GasBench Plus IRMS SystemAnalyze beverages using headspace gas equilibration
Isotope fingerprint in food and beverage products is region or process specific
Isotope fingerprintWhat is the biogeochemical interpretation?What is an example of food fraud interpretation?What products can be affected?
CarbonBotanical origin
(C3, C4 and CAM photosynthesis)
Adulteration
(e.g. sweetening with cheap sugar)
Honey, liquor, wine, olive oil, butter, flavors
NitrogenSoil processes, plant fertilizer processesMislabeling
(Differentiate organic and non-organic)
Fruits and vegetables, animal meat
SulfurLocal soil conditions, proximity to shorelineOrigin of productFruits and vegetables, animal meat, honey
OxygenPrincipally related to local-regional rainfall and hence geographical areaWatering of beverages, place of origin of productCoffee, wine, liquor, water, sugar, animal meat, flavors
HydrogenRelated to local-regional rainfall and hence geographical areaWatering of beverages, origin of productCoffee, wine, liquor, water, sugar, animal meat, flavors
Configuration/Use case
These processes can be traced using carbon, nitrogen, sulfur, oxygen and hydrogen isotopes, indicating the origin and history of food and beverage products.
Instruments and SoftwareVarious configurations available
Gas Isotope Ratio Mass Spectrometry (IRMS)
Qtegra ISDS Software for gas IRMS
H, C, N, O, S
AccessoriesTypical use case
Thermo Scientific EA IsoLink IRMS SystemAnalyze bulk samples
Thermo Scientific GC IsoLink II IRMS SystemAnalyze volatile compounds
Thermo Scientific LC IsoLink II IRMS SystemAnalyze polar compounds
Thermo Scientific GasBench Plus IRMS SystemAnalyze beverages using headspace gas equilibration
Become an Isotope Hunter - Isotope Fingerprints Compendium
Are you performing origin analysis? Don't miss this introduction to Isotope Fingerprints and how they are used for origin and authenticity for food integrity applications.
Download now
Detecting Clues, tracking Origin, unraveling History with isotope fingerprints
Isotope fingerprints: Finding clues, tracking origins and unraveling history
History can't hide from the isotope hunter. Geography, geology, and growth conditions of foods, fibers, liquids, and stone are embedded in their unique isotope fingerprints. Trace your sample history with our complete portfolio of IRMS systems.
Investigate more
Foodomics using the analysis of the molecular composition of foodstuffs
Rapidly obtain valuable information about the biochemical composition of food through mass spectrometry-based proteomics and metabolomics approaches. Total confirmational confidence in authenticity and adulteration testing with specificity and sensitivity is enabled through Orbitrap high resolution accurate mass (HRAM) LCMS and GCMS workflows.
Sensitive and accurate meat speciation investigation with HRAM LCMS
Food testing laboratories traditionally perform speciation testing using polymerase chain reaction (PCR) and enzyme-linked immunosorbent assay (ELISA). For public health and food safety authenticity investigations, these methods require intense customization to achieve the required sensitivity and accuracy and the molecular information is incomplete, making data mining post-analysis impossible.
Using a bottom up LCMS proteomic strategy, meat speciation testing can achieve the sensitivity and selectivity required to detect meat adulteration with accuracy.
Illustration of the analytical strategy used based on a proteogenomic approach to develop a targeted mass spectrometry-based proteomic method for meat authenticity and adulteration
Illustration of the analytical strategy used based on a proteogenomic approach to develop a targeted mass spectrometry-based proteomic method for meat authenticity and adulteration.
Characterization and authentication of food samples using integrated metabolomics
Flavor profiling and authentication can be a challenging analysis as the sample matrices encountered are chemically complex, the compounds are present over a wide dynamic range, and profiling requires sensitive and stable systems. An integrated metabolomics approach can accelerate characterization and identification of volatile and non-volatile food samples for profiling and food investigations.
V-plot scatterplot
V-plot scatterplot showing the statistical significance (P value) versus magnitude of change (fold change) when comparing the oregano adulterated samples (with thyme) versus the native oregano. The main chemical components that are responsible for sample diversity between two sample groups are selected (light blue dots).
Related products
Transform your food discovery characterization and identification studies
Discovery-based small molecule research has re-emerged as a key sector due to the creation and expansion of many routine applications. Targeted screening and quantitative workflows may only profile a fraction of the sample-specific compounds, potentially missing critical factors, whether this means limited coverage of metabolites and degradants, outstanding unknown or unexpected pesticides, or the misidentification or characterization of natural products that could potentially provide health benefits.
Download ebook
Next generation sequencing technologies, including high-throughput digital PCR, are enabling rapid identification of pathogenic bacteria and fungi to ensure food sources are safe before they reach shelves and tables.
GMO-specific DNA detection and quantification
Find GMO real-time PCR testing solutions for your complete workflow with Applied Biosystems TaqMan GMO Detection and Quantification Kits, supplied in partnership with Imegen™.
Reliable detection and quantification of species DNA in meat and feed samples. Identification of specific meat or fish species present in food samples is an essential step for verification of origin and traceability of raw materials, as well as for quality control of handling and cleaning processes in production lines. We offer a broad portfolio of high sensitivity, real-time PCR based meat species and fish detection and quantification testing solutions.
  • Rapid: Same day results
  • Full workflow solution: DNA extraction options for low and high sample input
  • High sensitivity: Detection of mitochondrial DNA
  • Quantitative: Quantification based on DNA standards, species DNA/total animal DNA result (%)
Learn more ›
Meat species detection, screening, and quantification
When verifying the origin and traceability of raw materials and for quality control of handling and cleaning processes in production lines, it is essential to accurately identify meat and fish species in food and ingredient samples. Learn more about the real-time PCR-based species detection and quantification solutions we have developed in collaboration with Imegen*.
Meat species detection, screening and quantification
Next generation sequencing for species screening and identification
Combat food fraud with a complete next generation sequencing (NGS) workflow for meat, fish and plant species screening and identification (ID). Initiate in-house multi-species ID testing of even the most complex food samples for accurate detection and differentiation of expected or unexpected species.
Next Generation Sequencing for species screening and identification
  • Simple: ready-to-use kits include all necessary reagents for DNA labeling and amplification, and provide results with any sample type, including complex and processed samples
  • Rapid: allows preparation of NGS library from extracted DNA within 4 hours
  • Flexible: utilizes a unique labelling system in which the DNA of each food sample is individually identified with an exclusive barcoded DNA sequence, enabling analysis of multiple samples together for meat, fish and plant species
  • Supported: our experienced technical team is ready to assist you
Learn more ›
Keep informed about topics related to safety, quality and authenticity of foods and ingredients
Read the latest news, articles and updates in the Food Safety and Authenticity Bulletin.
Learn more
Learn more about the latest food microbiology testing issues, trends, and solutions
Explore our Food Safety and Authenticity Webinar Hub to learn about the latest issues and trends within food microbiology testing, from experts working in the fields of food pathogen detection, authenticity, and food safety testing.
View webinar hub
How Isotopes Can Help with Food Authenticity Control
How isotopes can help with food authenticity control
Isotopic analyses of foods have become a widespread tool to evaluate the quality, authenticity and origin of food products. See how IRMS technology is improving food quality.