Vials of bovine serum albumin product
What is Bovine Serum Albumin?
Bovine serum albumin (BSA) is a soluble monomeric protein (66.5 kDa) synthesized in the liver. BSA stabilizes extracellular fluid volume and is a carrier for a variety of small molecules, including steroids, fatty acids, and thyroid hormones in serum. In addition, BSA protein can serve as an antioxidant, trapping harmful free radicals.
Bovine serum albumin is used in many laboratory procedures:
  • Used as a standard for protein quantification
  • Used as a blocking reagent for immunoassays
  • Added as a stabilizing component to restriction enzyme digest reactions
  • Added to antibody solutions for increased antibody stability and longevity
  • Supplemented into cell culture media for enhanced nutrition
BSA selection guide
What is bovine serum albumin used for in cell culture?
Supplementation of cell culture media with BSA protein can benefit cell growth and survival, especially when using serum-free media. Bovine serum albumin binds essential molecules present in the media (e.g., fatty acids, hormones, peptides), allowing for efficient delivery of these molecules into cultured cells. The antioxidative properties of BSA protein can also reduce cellular stress and damage.
Thermo Fisher Scientific offers a variety of Gibco BSA products optimized for supplementing cell culture media, including AlbuMAX Lipid-Rich BSA, which can reduce or eliminate the need for supplementation of cell culture media with serum.
Why is bovine serum albumin used for blocking?
BSA protein is also commonly used for blocking during immunoassays, including ELISA and western blotting. As a single protein, BSA is less likely to exhibit cross-reactivity with antibodies than milk, which is a complex protein mixture. In addition, BSA protein, which is generally unphosphorylated, is often recommended for assays involving anti-phosphoprotein antibodies. Anti-phosphoprotein antibodies may nonspecifically bind phosphorylated proteins found in milk blockers (i.e., casein). However, anti-phosphoprotein antibodies are unlikely to bind BSA, reducing background.
Blocker BSA in PBS and Blocker BSA in TBS are both available from Thermo Fisher Scientific as valuable additions to your immunoassay protocols.
Fluorescent BSA proteins for tracer studies
Thermo Fisher Scientific offers bovine serum albumin conjugated to fluorescent tags, which can be used for tracer studies in combination with anti-BSA antibodies. In addition, BSA fluorescent conjugates can be used in quantitative studies of electroporation, and also to measure plasma in rats.
Catalog # Name Size Price (KRW) Qty
A23016 Albumin from Bovine Serum (BSA), Tetramethylrhodamine conjugate Each
251,000
A34785 Albumin from Bovine Serum (BSA), Alexa Fluor™ 647 conjugate Each
358,000
Online offer
Ends: 30-Jun-2025
447,000
Save 89,000 (20%)
A23015 Albumin from Bovine Serum (BSA), FITC conjugate Each
197,000
Online offer
Ends: 30-Jun-2025
246,000
Save 49,000 (20%)
A34786 Albumin from Bovine Serum (BSA), Alexa Fluor™ 555 conjugate Each
406,000
Online offer
Ends: 30-Jun-2025
507,000
Save 101,000 (20%)
A11133 Bovine Serum Albumin Polyclonal Antibody 500 µL
356,000
Online offer
Ends: 30-Jun-2025
418,000
Save 62,000 (15%)
A23018 DNP-BSA (Albumin from Bovine Serum (BSA), 2,4-Dinitrophenylated) Each
195,000
Online offer
Ends: 30-Jun-2025
243,000
Save 48,000 (20%)
A34787 Albumin from Bovine Serum (BSA), Alexa Fluor™ 680 conjugate Each
394,000
Online offer
Ends: 30-Jun-2025
492,000
Save 98,000 (20%)
A23017 Albumin from Bovine Serum (BSA), Texas Red™ conjugate Each
200,000
Online offer
Ends: 30-Jun-2025
249,000
Save 49,000 (20%)
A13101 Albumin from Bovine Serum (BSA), Alexa Fluor™ 594 conjugate Each
357,000
Online offer
Ends: 30-Jun-2025
446,000
Save 89,000 (20%)
A13100 Albumin from Bovine Serum (BSA), Alexa Fluor™ 488 conjugate Each
359,000
Online offer
Ends: 30-Jun-2025
448,000
Save 89,000 (20%)
BSA selection guide
ApplicationNameSizeFormPurificationCat. No.
Media bottle icon
Cell culture
Bovine Albumin Fraction V (7.5% solution)100 mLLiquid 15260037
Media bottle icon
Cell culture
AlbuMAX I Lipid-Rich BSA25 gPowderChromatographical (IgG content ≤0.1%)11020021
Media bottle icon
Cell culture
AlbuMAX I Lipid-Rich BSA1 kgPowderChromatographical (IgG content ≤0.1%)11020039
Media bottle icon
Cell culture
AlbuMAX II Lipid-Rich BSA25 gPowderChromatographical (IgG content ≤1.0%)11021029
Media bottle icon
Cell culture
AlbuMAX II Lipid-Rich BSA100 gPowderChromatographical (IgG content ≤1.0%)11021037
Media bottle icon
Cell culture
AlbuMAX II Lipid-Rich BSA1 kgPowderChromatographical (IgG content ≤1.0%)11021045
Antibody icon
Tracer studies
and immunoassays
Bovine Serum Albumin Polyclonal Antibody500 µLLiquidIgG fractionA11133
Labeled protein icon
Tracer studies
DNP-BSA (Albumin from Bovine Serum (BSA), 2,4-Dinitrophenylated)5 mgLyophilized powder A23018
Labeled protein icon
Tracer studies
Albumin from Bovine Serum (BSA), FITC conjugate5 mgLyophilized powder A23015
Labeled protein icon
Tracer studies
Albumin from Bovine Serum (BSA), Alexa Fluor 488 conjugate5 mgLyophilized powder A13100
Labeled protein icon
Tracer studies
Albumin from Bovine Serum (BSA), Tetramethylrhodamine conjugate5 mgLyophilized powder A23016
Labeled protein icon
Tracer studies
Albumin from Bovine Serum (BSA), Alexa Fluor 555 conjugate5 mgLyophilized powder A34786
Labeled protein icon
Tracer studies
Albumin from Bovine Serum (BSA), Texas Red conjugate5 mgLyophilized powder A23017
Labeled protein icon
Tracer studies
Albumin from Bovine Serum (BSA), Alexa Fluor 594 conjugate5 mgLyophilized powder A13101
Labeled protein icon
Tracer studies
Albumin from Bovine Serum (BSA), Alexa Fluor 647 conjugate5 mgLyophilized powder A34785
Labeled protein icon
Tracer studies
Albumin from Bovine Serum (BSA), Alexa Fluor 680 conjugate5 mgLyophilized powder A34787
DNA icon
Restriction enzyme
cloning, chromatin biology
Bovine Serum Albumin150 mgLiquid 15561020
cell culture media icon GMP manufacturing
Gibco cell culture products are manufactured in facilities compliant with current good manufacturing practices (GMP) and adhere to a robust quality management system, ensuring the consistency, reliability, and high quality you can rely on.
Learn more about GMP manufacturing
leaf icon Sustainable solutions
We are committed to delivering products that serve the research needs of our customers, while striving to develop them in a way that minimizes our use of natural resources and our impact on the environment.
Learn more about sustainable solutions
Resources
Cell Culture & Transfection Learning Center
Access cell culture and transfection educational resources for better experiment planning and execution.
Gibco Cell Culture Basics
Learn the fundamentals of cell culture for achieving consistent results, including laboratory setup, safety, and aseptic techniques.
Media Formulation Tool
Find the right Gibco media formulation for DMEM, DMEM/F-12, MEM, and RPMI-1640 media.
Related products
Support
Cell Culture & Transfection Support Center
Find technical support recommendations for your cell culture and transfection workflows, including tips for experimental setup and in-depth troubleshooting help.
Need technical support? Contact our expert team for technical and application support of Laboratory Products.
For Research Use Only. Not for use in diagnostic procedures.