AlbuMAX™ I Lipid-Rich BSA
Product Image
Gibco™

AlbuMAX™ I Lipid-Rich BSA

Lipid-rich bovine serum albumin for cell culture (1). It is chromatographically purified and has an IgG content ≤ 0.1%.Origin: NewRead more
Have Questions?
Catalog NumberQuantity
1102002125 g
Catalog number 11020021
Price (USD)
142.00
Each
Add to cart
Quantity:
25 g
Recurring order eligible. Learn more »
Price (USD)
142.00
Each
Add to cart
Ask our AI about this Product

Lipid-rich bovine serum albumin for cell culture (1). It is chromatographically purified and has an IgG content ≤ 0.1%.

Origin: New Zealand

For Research Use Only. Not for use in diagnostic procedures.
Specifications
Product LineAlbuMAX™, Gibco™
Quantity25 g
Shipping ConditionRoom Temperature
Product TypeBSA
Unit SizeEach
Contents & Storage
Store in refrigerator (2–8°C).
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Customers who viewed this item also viewed



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3083143Certificate of AnalysisMay 10, 202511020039, 11020088, 11020021, 11020013
3083142Certificate of AnalysisMay 02, 202511020039, 11020088, 11020021, 11020013
3024056Certificate of AnalysisApr 25, 202511020039, 11020088, 11020021, 11020013
2833722Certificate of AnalysisJan 26, 202511020039, 11020088, 11020021, 11020013
2494919BCertificate of AnalysisJan 26, 202511020039, 11020088, 11020021, 11020013
5 results displayed, search above for a specific certificate

Safety Data Sheets

Scientific Resources

Product Information

Limited Use Label Licenses (LULL)

'

Notice to Purchaser: The purchase of this product conveys to the purchaser the limited, non- transferable right to use the purchased amount of the product to (a) perform internal research for the sole benefit of the purchaser; (b) manufacture protein (or other biological material) for resale; and (c) perform research or manufacturing services conducted by the purchaser on a fee for service or contract basis for or on behalf of third parties. However, the purchaser may transfer this product, its components, or materials made using this product to a third party (including contract research/manufacturing organizations), provided that each such third party agrees in writing to use such product, components, or materials solely on behalf of the purchaser, and such third party is restricted from further transferring any such product, components, or materials to any individual or entity other than the purchaser. No additional rights are granted. By purchasing this product, the purchaser agrees not to: (1) resell the product in any form; (2) use the product as a therapeutic agent or diagnostics test component; (3) reverse engineer the product or cause the product to be reverse engineered; or (4) use the product for purposes other than what is indicated in this Limited Use Label License. Life Technologies is not aware of Intellectual Property ("IP") that would be infringed by the manufacture or sale of its products. Customers are urged to perform an IP search and analysis specific to their manufacturing processes and target biologic products. Should that analysis require information about Life Technologies products, Life Technologies will provide to the customer's IP counsel the information necessary to analyze any relevant IP in a manner that protects Life Technologies proprietary information. The purchaser is responsible for obtaining all regulatory approvals necessary for any therapeutic or diagnostic use of the protein (or biological material) manufactured using this product. For information on obtaining additional rights, please contact outlicensing@thermofisher.com, Licensing and Commercial Supply, Thermo Fisher Scientific, 5823 Newton Drive, Carlsbad, CA, 92008, United States.

'

Frequently asked questions (FAQs)

Below are the concentrations of free fatty acids obtained from the HPLC lipid profiles of an older lot of AlbuMAX I:

Alpha-linolenic acid: 0.59 mg FFA/g protein
Linoleic acid: 0.65 mg FFA/g protein
Oleic acid: 2.12 mg FFA/g protein
Stearic acid: 2.09 mg FFA/g protein
Palmitic acid: 1.90 mg FFA/g protein
Note: The individual lipid concentrations may vary between lots, but the values above can be used as a good approximation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Share catalog number, name or link

1x1 image pixel for data collection