KingFisher Apex System
The Thermo Scientific KingFisher Apex represents the pinnacle of automated purification for DNA, RNA, proteins, and cells. Building on decades of KingFisher product value and excellence, we’ve combined exceptional instrument capabilities with complete touchscreen-based control to deliver excellent flexibility and performance. The KingFisher Apex makes sample prep easy.
Video Player is loading.
Current Time 0:00
Duration 2:01
Loaded: 0%
Stream Type LIVE
Remaining Time 2:01
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Explore KingFisher Apex with a self-guided tour
    Virtually navigate the KingFisher Apex instrument and get a closer look at the features, supporting kits and reagents, and more!
    Note: This tool is best viewed in Chrome, Firefox, or Safari browser. Learn more
    Features
    In addition to the common features of all KingFisher Purification Systems, the Apex offers:
    High-throughput purification
    1–96 samples per run
    Choose from two plate formats
    Wide 20 μL–5 mL volume range
    Download protocols from our library
    Or easily customize protocols using Thermo Scientific BindIx Software
    Easy to install and run with simple workflow
    Follow on-screen instructions (including color guided plate loading) and start first run in <10 minutes
    Security & maintenance features
    User login and access levels for added security. Pop-up reminders for yearly maintenance.
    Ultraviolet lamp
    Decontamination is standard
    Elute directly to PCR plates
    Store your samples for later use with PCR compatible storage tubes
    kingfisher-apex-brochure
    Touchscreen control
    A modern touchscreen display and an intuitive interface puts you in control of instrument operation. Follow the guided visuals for easy plate loading, protocol changes, and instrument setup.
    It’s easy to edit or write protocols directly on the instrument, and you can customize protocols at every step—bind, wash, elute. You also have the option to manage protocols on your PC with BindIx software. Instrument-defined limits help reduce potential errors in any protocol modifications.
    KingFisher Apex touchscreen interactive demo
    Click buttons to see preview
    Dual magnets, tip combs, and plates
    dualmagnets2
    Dual magnets, multiple formats
    When you install two magnets and heating blocks, KingFisher Apex intelligently selects the correct magnetic head based on your protocol. Work from large to small volume ranges by choosing from 24-well, 96-well, or PCR formats with corresponding magnetic heads and plates.
    Interested in diagnostic nucleic acid extraction?
    Demo videos
    Protocols and software
    Thermo Scientific BindIx software is included with KingFisher Apex instruments. The software makes it easy to create, download, run, modify, and store protocols directly on the instrument or on your PC.
    Protocols are available for nucleic acid and protein purification procedures with reagent kits. The software allows you to define and edit steps, parameters, plates, and reagents for every part of your protocol—bind, wash, and elute. You can even generate status reports including a run log, plate layout, and step parameters.
    For more information about BindIx, see the Features tab.
    Attention KingFisher Flex users: KingFisher Flex scripts and protocols can be readily opened and converted to KingFisher Apex scripts.
    To download BindIx software, choose one of the methods in the table. 
    Three ways to download BindIx software
    Method
    Instructions
    Web link
    Double-click the downloaded package to start the installation process
    Microsoft Store
    Click Get, then Install
    USB drive from manufacturer
    Double-click the .msix bundle file, then click Install
     System requirements: Windows 10 version 1809 (17763.0) or higher
    Applications
    The KingFisher Apex can gently and efficiently isolate and purify nucleic acids, proteins, and cells from a range of sample types for a wide variety of downstream applications. For a selection of application kits, reagents, and protocols that run on the KingFisher Apex, see the Kits, Reagents, and Protocols tab.
    Plates, plastics, and setup
    For information about plastics and plate setup products designed to work with the KingFisher Apex, see the Accessories tab.
    Specifications
    Attribute
    Description
    Instrument size
    Benchtop
    Throughput level
    Medium to high
    Processing volume by plate format
    96-well plate
    • Deep-well: 50–1,000 µL
    • KingFisher standard: 15–200 µL
    • PCR: 10–80 µL
    24-well plate
    • Deep-well: 30–5,000 µL
    Storage tubes
    • 96 tubes: 30–200 µL
    • 24 tubes: 200–1,000 µl
    Samples per run
    24 or 96
    Software included
    BindIx
    Customizable protocols
    Yes, created with instrument user interface or BindIx PC software*
    Heating/cooling ranges
    Heating up to 100ºC
    Cooling down to 4ºC
    Ultraviolet lamps
    2 UV lamps, max run time 23 h 59 min
    Dimensions (H x W x D)
    45 x 78 x 59 cm (18 x 31 x 23 in)
    Weight
    56 kg (123 lb)
    Plate formats
    96 deep-well plate
    96-well KingFisher standard plate
    96-well PCR plate (skirted and semiskirted)
    24 deep-well plate
    96 storage tubes
    24 storage tubes
    Instrument memory
    ~30 GB
    Display interface
    Graphical user interface with touch screen
    Power
    100–240 VAC 50/60 Hz, 280 VA
    Barcode reader
    On instrument; 1D barcodes
    Magnetic head options
    4 options, customer interchangeable
    Data connectivity
    USB device for PC
    USB Wi-Fi or LAN adapter for network
    RS-232 for automation
    Cloud enabled
    * Requires Windows 10 version 1809 (17763.0) or higher.