Membrane protein extraction and isolation is critical to understanding the structural and functional role of proteins within biological systems. Membrane proteins play crucial roles in various biological processes, serving as key players in cell signaling, transport, and communication between cells and their environment. However, these proteins embedded with the cell membranes lipid bilayer, presents unique challenges in their separation from other cellular components due to their hydrophobic nature and localization. Membrane protein extraction and isolation methods utilize strategies to obtain pure and functional membrane proteins for further analysis and research applications.
Thermo Fisher Scientific offers specialized membrane protein extraction and isolation reagents, kits, polymers, and detergents that are optimized for high protein yield, preserving protein function, minimizing contamination, and helping ensure compatibility with downstream applications. Choose from our comprehensive range of membrane protein extraction and isolation products based on your specific isolation needs. 
Strategies for extraction and stabilization
Pre-formulated reagents and kits
Detergents
Polymers
✓  Traditional protein extraction method
  Optimized buffers and reagents
  No upfront screening
•   May need to buffer exchange downstream
Maintains protein-protein interactions, protein activity and function, and protein structure
•  Need to ensure compatibility with downstream application
•  Requires screening to balance efficiency and compatibility
  Detergent-free method for extracting proteins and surrounding lipids
•  Need to ensure native conformation
•  Requires screening/research to confirm fit for purpose
Choose the appropriate membrane protein extraction method
 GPCR Extraction and Stabilization ReagentMem-PER Plus Membrane Protein Extraction KitCell Surface Protein Biotinylation and Isolation Kit
 GPCR Extraction and Stabilization ReagentMem-PER Plus Membrane Protein Extraction KitCell Surface Protein Biotinylation and Isolation Kit
Membrane protein enrichmentG Protein-Coupled Receptors (GPCR)Plasma membrane proteinsIntegral and membrane associated proteins
Protein functionalityMaintains structural integrity as measured by immunoprecipitation and receptor ligand binding assaysEnrichment compatible with western blottingEnrichment compatible with western blotting and MS applications
Compatible sample typesTissues and cultured mammalian cellsTissues and cultured mammalian cellsCultured mammalian cells
Cytosolic contaminationWhole cell lysateLess than 5% contamination of cytosolic proteinsLess than 20% contamination of cytosolic proteins
Sample processing time1–2 hours1 hour2 hours
Mechanical disruption required?Yes, for tissueYes, for tissueNo
Amount of sample processed100 samples with 107 cells or 100 samples with 50–100 mg tissue50 samples with 5 million cells or 25 samples of 20–40 mg tissue per kit8 experiments, with two confluent 15 cm dishes or four T75 flasks
Compatible protein assaysBCA Protein AssaysBCA Protein Assays, Detergent-Compatible BradfordLysis buffer is compatible with BCA and Rapid Gold BCA; elution buffer is not compatible
Downstream compatibilityWestern blot, IP, protein purification, radio-ligand binding assaysIP, western blot, ELISA, amine reactive labelingWestern blot, ELISA, mass spectrometry analysis
Protease/phosphatase inhibitors recommended?Yes, bothYes, bothYes, both
Available size(s)100 mL300 mL8 samples
User guideUser Guide: GPCR Extraction and Stabilization ReagentUser Guide: Mem-PER Plus Membrane Protein Extraction KitUser Guide: Pierce Cell Surface Protein Biotinylation and Isolation Kit
Reagent-based membrane protein extraction and isolation
Reagent-based lysis has replaced traditional physical lysis as the method of choice for membrane disruption and extraction of membrane proteins. Reagent-based lysis methods do not require expensive, cumbersome equipment and protocols that are difficult to implement. The Thermo Scientific membrane protein extraction and membrane protein isolation reagents consist of optimized concentrations of detergents, buffers, salts, and reducing agents developed for particular species and types of cells. Reagents also have the added benefits of both lysing and solubilizing effects. Our membrane protein isolation and membrane protein extraction kits are optimized specifically for your needs to isolate high-quality protein samples essential to completing successful downstream applications.
Features:
  • Extraction and isolation—produces minimal cross-contamination (typically less than 10%) of cytosolic protein into the membrane protein fraction
  • Cells or tissues—effective for extraction from cultured mammalian cells and mammalian tissues
  • Downstream compatibility—analyze membrane protein extracts by SDS-PAGE, western blotting, immunoprecipitation, and protein assays
  • Fast and simple—isolation of membrane proteins in approximately one hour
  • No special equipment required—only a benchtop microcentrifuge, tubes, homogenizers, and pipettors are needed
Membrane protein isolation kits
Extraction and stabilization of GPCR’s
The GPCR Extraction and Stabilization Reagent is an easy-to-use solution that extracts and stabilizes G-couples protein receptors (GPCR’s) and other membrane-associated protein from cells and tissues in 1–2 hours (Figure 1). This all-in-one reagent stabilizes the receptor by encapsulating it in a detergent micelle that maintains the structural and functional integrity of the protein (Figure 2). This stabilization process also allows for more flexibility for receptor analysis by enabling extended storage of the extracted protein at 4°C for up to one week and at –20°C for up to one month. 
Application note:Extraction and Purification of Functional G Protein-Coupled Receptors from Expression System
 Poster:Effective solubilization and stabilization of functional G protein-coupled receptors
Figure 2. Preserve functionality of receptors in cells and tissues. Extracts from GeneBLAzer expression cell lines or mouse brain tissue were prepared using the GPCR Extraction and Stabilization Reagent to assess activity using radioligand binding assays. Extracts (400–600 µg) containing Muscarinic Acetylcholine Receptor 3 (M3) and serotonin receptor (5HT1A) (GeneBLAzer expression cell lines) or adenosine A2a receptor (ADORA2A) (mouse brain tissue) were then incubated with H3-4-DAMP, H3-propanolol, or H3-adenosine (PerkinElmer) alone or in a competition reaction of cold H3-ligand for 75 minutes at room temperature. Free radioligand was then removed and samples were then analyzed using a TRI-CARB 2000 TR scintillation counter.
Extraction and enrichment of integral membrane proteins
MEM-PER Plus Membrane Protein Extraction Kit allows for solubilization and enrichment of integral membrane proteins and membrane-associated proteins. This membrane protein isolation kit offers a user-friendly alternative to traditional protein isolation techniques, utilizing a straightforward benchtop microcentrifuge procedure (Figure 3). 
Application note:Efficient Mammalian Membrane Protein Extraction
Membrane protein extraction and enrichment can be obtained from adherent and suspension cells as well as hard and soft tissues (Figure 4). The extraction efficiencies and yields of integral membrane proteins will depend on the cell type and the number of transmembrane domains. Typically, proteins with one or two transmembrane domains have a high extraction efficiency (up to 90%), while cross-contamination of cytosolic proteins into the membrane fraction is typically minimal (less than 10%).
isolated membrane proteins from different tissues have minimal contamination from the cytosol
Figure 4. Isolation and enrichment of membrane proteins from different tissues. Membrane proteins were isolated from frozen mouse heart or brain (30 mg) following the Mem-PER Plus Membrane Protein Extraction Kit protocol. Membrane and cytosolic fractions (10 μg) were separated by SDS-PAGE and transferred to a nitrocellulose membrane. Western blots were done using the Thermo Scientific Fast Western Rabbit Dura Kit and primary antibodies diluted 1:1000. Images were generated using the Thermo Scientific myECL Imager.
Biotinylation and enrichment of plasma membrane proteins
The Cell Surface Biotinylation and Isolation Kit biotinylates, solubilizes, and enriches plasma membrane proteins. This membrane protein isolation kit offers a less harsh alternative to traditional protein isolation techniques that can result in denatured and contaminated plasma membrane proteins. The cell surface biotinylation and isolation kit is easy to use with a reduced number of steps which reduces processing time, and reagent formulations to improve enrichment and extraction while minimizing intracellular protein contamination. This procedure involves the biotinylation of proteins using a thiol-cleavable amine-reactive biotinylation reagent, EZ-Link Sulfo-NHS-SS-Biotin. Cells are then lysed, and labeled proteins are captured with NeutrAvidin Agarose and eluted with dithiothreitol (DTT) to reduce disulfide bonds in the biotin label (Figure 5).
  Poster: Proteomic analysis of cell surface proteins with improved specificity of enrichment
The cell surface biotinylation and isolation kit is compatible with a range of adherent and suspension cell lines. It also allows for the comparative protein expression analysis in treated versus non-treated cells as well as different cell lines. Selectivity and efficiency of isolated membrane protein can be confirmed by western blot (Figure 6) and LC-MS analysis (Figure 7).
Figure 6. Western blot analysis of enriched cell surface proteins. Samples were prepared in duplicate from two cell lines using the Cell Surface Protein Biotinylation and Isolation Kit. Flow-through (F), elution (E), and bead boil (B) were normalized by volume and analyzed by western blot for cell surface proteins (EGFR, CD55, cadherin, and integrin α5) or intracellular proteins (HSP90, actin, calnexin, and β-tubulin). The blots were imaged using the iBright FL Imaging System. The data demonstrates effective enrichment of cell surface proteins with minimal contamination of intracellular proteins.
Figure 7. Classification of cell surface protein type in the top 50 most abundant proteins. Cell surface proteins were enriched from three mammalian cell lines, in duplicate, and then analyzed by LC-MS. Raw data was searched using Proteome Discoverer Software 2.2. The top 50 most abundant proteins were identified using the UniProt Knowledgebase. Cell surface proteins were classified as single- or multi-spanning (6–17 spans) transmembrane (TM) proteins, glycosylphosphatidylinositol (GPI) anchored proteins, or extracellular matrix (ECM) proteins. A similar distribution was observed in the three cell lines.
Non-denaturing detergents are excellent for membrane protein isolation as they maintain protein-protein interactions, protein activity and function, and protein structure. These types of detergents include non-ionic and zwitterionic. Non-ionic detergents are milder and are recommended to be used when maintaining protein interactions and activity is critical. Zwitterionic detergents are stronger and usually result in better membrane solubilization. See the different characteristics of non-denaturing detergents in the table below. 
Characteristics of membrane protein isolation detergents
NonionicZwitterionic
Maltosides Glucosides 
  • Lipid-like nonionic detergent
  • Especially useful for isolating and stabilizing hydrophobic membrane proteins
  • Preserves activity of membrane protein better than most of the detergents
  • High-purity compounds with low UV absorptivity
  • Gentle on proteins, minimizing denaturation 
  • Reduce electrostatic interactions 
  • Low critical micelle concentration (CMC)
  • High-purity with low UV absorptivity
  • Gentle on proteins, minimizing denaturation and aggregation 
  • Effectively disrupt lipid-protein interactions 
  • Exhibit low critical micelle concentration (CMC)
  • High-purity compounds with low UV absorptivity
When choosing a detergent for membrane protein extraction, it is important to remember the detergent best for solubilization may not be the best for your application.
Learn more about non-ionic versus ionic, and non-denaturing versus denaturing detergents and see other detergents for protein solubilization
Matoside detergents
Maltosides are highly efficient at extracting and stabilizing hydrophobic membrane proteins. They assist in the purification and characterization of membrane proteins as such can be used in structural and functional analysis.
 n-Dodecyl-β-Maltoside Detergent (DDM)Lauryl Maltose Neopentyl Glycol (LMNG)n-Undecyl-β-D-maltopyranoside (UDM)n-Octyl-β-D-Maltopyranoside (OM)n-Nonyl-β-Maltoside (NM)
Description typeNon-ionic
Functional propertiesUsed most often for hydrophobic membrane proteins.Ideal for integral membrane proteins.Forms stronger interactions with hydrophobic regions of membranes proteins.Flexible for use in a wide range of experimental conditions, buffer systems, and pH rangers.Forms stable micelles at lower concentrations important for protein stability and integrity.
Dialyzable NoYesYesYesYes
Aggregation number*98 average (70–140 range)Unknown~71 in 100 mM NaCl, 20 mM HEPES pH 7.5~47 in 100 mM NaCl, 20 mM HEPES pH 7.5
~55 in 100 mM NaCl, 20 mM HEPES pH 7.5
MW510.61005.2496.6454.4468.5
CMC %w/v (mM)0.009% (0.17) in water0.001% (~0.01)0.03% (~0.59)0.89% (~19.5) in 100 mM NaCl, 20 mM HEPES pH 7.50.28% (~6)
Compatible with Detergent Removal ResinYesNoYesNoYes
Available size(s)1 g
5 g
1 g (solid)
10 x 1 mL (liquid)
1 g
5 g
1 g
5 g
1 g
5 g
See all maltoside detergents
Cyclohexyl maltoside detergents
Cyclohexyl maltosides are designed for the extraction of intact complexes. They can be used with protein purification, crystallization, spectroscopy, and structural studies.
 5-Cyclohexyl-1-Pentyl-β-D-Maltoside6-Cyclohexyl-1-Hexyl-β-D-Maltoside7-Cyclohexyl-1-Heptyl-β-D-Maltoside
Description typeNon-ionic
Functional propertiesIdeal for membrane protein complexes, solubilization efficiency depends on specific protein, structural features, and experimental conditions.Increased hydrophobicity may result in enhanced solubilization of higher hydrophobic regions or multiple transmembrane domains. Well suited for structural biology studies.Provides improved stability, maintaining structural integrity and functionality. Well suited for structural biology studies.
Dialyzable YesYesYes
Aggregation number*~47~91~150
MW494.5508.5522.5
CMC %w/v (mM)0.12% (~2.4–5.0)0.028% (~0.56)0.0099% (~0.19)
Compatible with Detergent Removal ResinYesYesYes
Available size(s)1 g
5 g
1 g
5 g
1 g
5 g
See all cyclohexyl maltoside detergents
*Aggregation number is the number of molecules per micelle
Glucoside and other non-ionic detergents 
Glucosides are gentler detergents that enhance solubilization of hydrophobic biomolecules. They can be used with protein purification, crystallization, and structural studies. Tetraethylene Glycol Monooctyl Ether (C8E4) is good for solubilization of sensitive biomolecules. It can be used in native mass spectrometry.
 n-Nonyl-β-D-glucoside (NG)Octyl-β-Glucoside Detergent (OG)Octylthioglucoside (OTG)Tetraethylene Glycol Monooctyl Ether (C8E4)
Description typeNon-ionic
Functional propertiesContains a longer alkyl chain resulting in higher hydrophobicity, enhancing solubilization and improving extraction of membrane proteins.
Widely used for membrane protein solubilization
Advantageous for application such as protein purification and structural studies since they provide better solubility and reduced downstream interference.
Effective for cell lysis and nondenaturing protein solubilization
Useful for applications where enzyme stability is desired since it can remain stable in the presence of beta-D-glucoside glucohydrolase enzymes.
Good for solubility in both aqueous and organic solvents
Ideal for sensitive biomolecules, and membrane proteins or protein complexes used for MS.
Dialyzable YesYesYesYes
Aggregation number*Unknown27Unknown~82
MW308.4292.444306.5
CMC %w/v (mM)0.20% (6.5)0.67–0.73% (23-25)0.28% (9)0.25% (~8)
Cloud point (°C)Unknown>100>100Unknown
Compatible with Detergent Removal ResinYesYesYesYes
Available size(s)1 g
5 g
5 g5 g10 mL
See all glucoside detergents
* Aggregation number is the number of molecules per micelle
Zwitterionic detergents 
Tetraethylene Glycol Monooctyl Ether (C8E4), Lauryldimethylamine-N-oxide (LDAO), and Fos-Choline detergents can all be used for protein purification and structural studies. LDAO are ideal for MS and crystallization studies; while fos-choline is also used for membrane protein research. 
 Lauryldimethylamine-N-oxide (LDAO)Fluorinated Fos-Choline-8Fos-choline-12Fos-choline-15
Description typeZwitterionic
Functional propertiesIdeal for the study of intact membrane protein structure and non-covalent interactions.Efficient at solubilization of challenging and delicate membrane proteins. 
Ideal for sensitive membrane proteins, minimizes potential cytotoxic effects such as denaturation and aggregation.Optimal for wide range of membrane protein types by maintaining native conformation and functionality of proteins.Increased hydrophobicity results in enhanced solubilization and stabilization of larger hydrophobic regions or transmembrane domains.
Dialyzable YesYesYesYes
Aggregation number*~76Unknown~54~131
MW229.4529.2351.5393.5
CMC % w/v (mM)0.023% (~1-2)-- (~2.9)0.047% (1.5)0.0027% (0.07)
Compatible with Detergent Removal ResinYesYesYesNo
Available size(s)1 g
5 g
1 g
5 g
1 g
5 g
1 g
5 g
See all fos-choline detergents
* Aggregation number is the number of molecules per micelle
Not worried about maintaining protein function? See our detergents for protein solubilization
Polymers for detergent-free membrane protein isolation
Polymers represents a cutting-edge approach to membrane protein extraction, offering a detergent-free method for isolating membrane proteins along with their surrounding lipid bilayer. This innovative technology provides researchers with a powerful tool for studying membrane proteins in their native lipid environment, potentially leading to more accurate structural and functional analyses. 
Like detergents, it is important to understand polymer characteristics in order to determine the best polymer for membrane protein target and intended experimental goal. A recent publication highlights the importance of selecting a polymer that effectively interacts with the specific lipid environment to maintain protein stability and functionality (1).
Characteristics of membrane protein extraction polymers
Attribute​Unit​SMALP® 200​SMALP® 300​
Polymer styrene-to-maleic anhydride ratio​n:m​2:1​3:1​
Molecular weight​Da​6,500​10,000​
Dry solids content​% w/w​20​20​
Solvent​Water​Water ​
pH​7.5​8.0​
Absolute viscosity​mPaꞏs​≤ 5,000​≤ 5,000​
 See all SMALP 200 polymersSee all SMALP 300 polymers
 GPCR Extraction and Stabilization ReagentMem-PER Plus Membrane Protein Extraction KitCell Surface Protein Biotinylation and Isolation Kit
 GPCR Extraction and Stabilization ReagentMem-PER Plus Membrane Protein Extraction KitCell Surface Protein Biotinylation and Isolation Kit
Membrane protein enrichmentG Protein-Coupled Receptors (GPCR)Plasma membrane proteinsIntegral and membrane associated proteins
Protein functionalityMaintains structural integrity as measured by immunoprecipitation and receptor ligand binding assaysEnrichment compatible with western blottingEnrichment compatible with western blotting and MS applications
Compatible sample typesTissues and cultured mammalian cellsTissues and cultured mammalian cellsCultured mammalian cells
Cytosolic contaminationWhole cell lysateLess than 5% contamination of cytosolic proteinsLess than 20% contamination of cytosolic proteins
Sample processing time1–2 hours1 hour2 hours
Mechanical disruption required?Yes, for tissueYes, for tissueNo
Amount of sample processed100 samples with 107 cells or 100 samples with 50–100 mg tissue50 samples with 5 million cells or 25 samples of 20–40 mg tissue per kit8 experiments, with two confluent 15 cm dishes or four T75 flasks
Compatible protein assaysBCA Protein AssaysBCA Protein Assays, Detergent-Compatible BradfordLysis buffer is compatible with BCA and Rapid Gold BCA; elution buffer is not compatible
Downstream compatibilityWestern blot, IP, protein purification, radio-ligand binding assaysIP, western blot, ELISA, amine reactive labelingWestern blot, ELISA, mass spectrometry analysis
Protease/phosphatase inhibitors recommended?Yes, bothYes, bothYes, both
Available size(s)100 mL300 mL8 samples
User guideUser Guide: GPCR Extraction and Stabilization ReagentUser Guide: Mem-PER Plus Membrane Protein Extraction KitUser Guide: Pierce Cell Surface Protein Biotinylation and Isolation Kit
Reagent-based membrane protein extraction and isolation
Reagent-based lysis has replaced traditional physical lysis as the method of choice for membrane disruption and extraction of membrane proteins. Reagent-based lysis methods do not require expensive, cumbersome equipment and protocols that are difficult to implement. The Thermo Scientific membrane protein extraction and membrane protein isolation reagents consist of optimized concentrations of detergents, buffers, salts, and reducing agents developed for particular species and types of cells. Reagents also have the added benefits of both lysing and solubilizing effects. Our membrane protein isolation and membrane protein extraction kits are optimized specifically for your needs to isolate high-quality protein samples essential to completing successful downstream applications.
Features:
  • Extraction and isolation—produces minimal cross-contamination (typically less than 10%) of cytosolic protein into the membrane protein fraction
  • Cells or tissues—effective for extraction from cultured mammalian cells and mammalian tissues
  • Downstream compatibility—analyze membrane protein extracts by SDS-PAGE, western blotting, immunoprecipitation, and protein assays
  • Fast and simple—isolation of membrane proteins in approximately one hour
  • No special equipment required—only a benchtop microcentrifuge, tubes, homogenizers, and pipettors are needed
Membrane protein isolation kits
Extraction and stabilization of GPCR’s
The GPCR Extraction and Stabilization Reagent is an easy-to-use solution that extracts and stabilizes G-couples protein receptors (GPCR’s) and other membrane-associated protein from cells and tissues in 1–2 hours (Figure 1). This all-in-one reagent stabilizes the receptor by encapsulating it in a detergent micelle that maintains the structural and functional integrity of the protein (Figure 2). This stabilization process also allows for more flexibility for receptor analysis by enabling extended storage of the extracted protein at 4°C for up to one week and at –20°C for up to one month. 
Application note:Extraction and Purification of Functional G Protein-Coupled Receptors from Expression System
 Poster:Effective solubilization and stabilization of functional G protein-coupled receptors
Figure 2. Preserve functionality of receptors in cells and tissues. Extracts from GeneBLAzer expression cell lines or mouse brain tissue were prepared using the GPCR Extraction and Stabilization Reagent to assess activity using radioligand binding assays. Extracts (400–600 µg) containing Muscarinic Acetylcholine Receptor 3 (M3) and serotonin receptor (5HT1A) (GeneBLAzer expression cell lines) or adenosine A2a receptor (ADORA2A) (mouse brain tissue) were then incubated with H3-4-DAMP, H3-propanolol, or H3-adenosine (PerkinElmer) alone or in a competition reaction of cold H3-ligand for 75 minutes at room temperature. Free radioligand was then removed and samples were then analyzed using a TRI-CARB 2000 TR scintillation counter.
Extraction and enrichment of integral membrane proteins
MEM-PER Plus Membrane Protein Extraction Kit allows for solubilization and enrichment of integral membrane proteins and membrane-associated proteins. This membrane protein isolation kit offers a user-friendly alternative to traditional protein isolation techniques, utilizing a straightforward benchtop microcentrifuge procedure (Figure 3). 
Application note:Efficient Mammalian Membrane Protein Extraction
Membrane protein extraction and enrichment can be obtained from adherent and suspension cells as well as hard and soft tissues (Figure 4). The extraction efficiencies and yields of integral membrane proteins will depend on the cell type and the number of transmembrane domains. Typically, proteins with one or two transmembrane domains have a high extraction efficiency (up to 90%), while cross-contamination of cytosolic proteins into the membrane fraction is typically minimal (less than 10%).
isolated membrane proteins from different tissues have minimal contamination from the cytosol
Figure 4. Isolation and enrichment of membrane proteins from different tissues. Membrane proteins were isolated from frozen mouse heart or brain (30 mg) following the Mem-PER Plus Membrane Protein Extraction Kit protocol. Membrane and cytosolic fractions (10 μg) were separated by SDS-PAGE and transferred to a nitrocellulose membrane. Western blots were done using the Thermo Scientific Fast Western Rabbit Dura Kit and primary antibodies diluted 1:1000. Images were generated using the Thermo Scientific myECL Imager.
Biotinylation and enrichment of plasma membrane proteins
The Cell Surface Biotinylation and Isolation Kit biotinylates, solubilizes, and enriches plasma membrane proteins. This membrane protein isolation kit offers a less harsh alternative to traditional protein isolation techniques that can result in denatured and contaminated plasma membrane proteins. The cell surface biotinylation and isolation kit is easy to use with a reduced number of steps which reduces processing time, and reagent formulations to improve enrichment and extraction while minimizing intracellular protein contamination. This procedure involves the biotinylation of proteins using a thiol-cleavable amine-reactive biotinylation reagent, EZ-Link Sulfo-NHS-SS-Biotin. Cells are then lysed, and labeled proteins are captured with NeutrAvidin Agarose and eluted with dithiothreitol (DTT) to reduce disulfide bonds in the biotin label (Figure 5).
  Poster: Proteomic analysis of cell surface proteins with improved specificity of enrichment
The cell surface biotinylation and isolation kit is compatible with a range of adherent and suspension cell lines. It also allows for the comparative protein expression analysis in treated versus non-treated cells as well as different cell lines. Selectivity and efficiency of isolated membrane protein can be confirmed by western blot (Figure 6) and LC-MS analysis (Figure 7).
Figure 6. Western blot analysis of enriched cell surface proteins. Samples were prepared in duplicate from two cell lines using the Cell Surface Protein Biotinylation and Isolation Kit. Flow-through (F), elution (E), and bead boil (B) were normalized by volume and analyzed by western blot for cell surface proteins (EGFR, CD55, cadherin, and integrin α5) or intracellular proteins (HSP90, actin, calnexin, and β-tubulin). The blots were imaged using the iBright FL Imaging System. The data demonstrates effective enrichment of cell surface proteins with minimal contamination of intracellular proteins.
Figure 7. Classification of cell surface protein type in the top 50 most abundant proteins. Cell surface proteins were enriched from three mammalian cell lines, in duplicate, and then analyzed by LC-MS. Raw data was searched using Proteome Discoverer Software 2.2. The top 50 most abundant proteins were identified using the UniProt Knowledgebase. Cell surface proteins were classified as single- or multi-spanning (6–17 spans) transmembrane (TM) proteins, glycosylphosphatidylinositol (GPI) anchored proteins, or extracellular matrix (ECM) proteins. A similar distribution was observed in the three cell lines.
Non-denaturing detergents are excellent for membrane protein isolation as they maintain protein-protein interactions, protein activity and function, and protein structure. These types of detergents include non-ionic and zwitterionic. Non-ionic detergents are milder and are recommended to be used when maintaining protein interactions and activity is critical. Zwitterionic detergents are stronger and usually result in better membrane solubilization. See the different characteristics of non-denaturing detergents in the table below. 
Characteristics of membrane protein isolation detergents
NonionicZwitterionic
Maltosides Glucosides 
  • Lipid-like nonionic detergent
  • Especially useful for isolating and stabilizing hydrophobic membrane proteins
  • Preserves activity of membrane protein better than most of the detergents
  • High-purity compounds with low UV absorptivity
  • Gentle on proteins, minimizing denaturation 
  • Reduce electrostatic interactions 
  • Low critical micelle concentration (CMC)
  • High-purity with low UV absorptivity
  • Gentle on proteins, minimizing denaturation and aggregation 
  • Effectively disrupt lipid-protein interactions 
  • Exhibit low critical micelle concentration (CMC)
  • High-purity compounds with low UV absorptivity
When choosing a detergent for membrane protein extraction, it is important to remember the detergent best for solubilization may not be the best for your application.
Learn more about non-ionic versus ionic, and non-denaturing versus denaturing detergents and see other detergents for protein solubilization
Matoside detergents
Maltosides are highly efficient at extracting and stabilizing hydrophobic membrane proteins. They assist in the purification and characterization of membrane proteins as such can be used in structural and functional analysis.
 n-Dodecyl-β-Maltoside Detergent (DDM)Lauryl Maltose Neopentyl Glycol (LMNG)n-Undecyl-β-D-maltopyranoside (UDM)n-Octyl-β-D-Maltopyranoside (OM)n-Nonyl-β-Maltoside (NM)
Description typeNon-ionic
Functional propertiesUsed most often for hydrophobic membrane proteins.Ideal for integral membrane proteins.Forms stronger interactions with hydrophobic regions of membranes proteins.Flexible for use in a wide range of experimental conditions, buffer systems, and pH rangers.Forms stable micelles at lower concentrations important for protein stability and integrity.
Dialyzable NoYesYesYesYes
Aggregation number*98 average (70–140 range)Unknown~71 in 100 mM NaCl, 20 mM HEPES pH 7.5~47 in 100 mM NaCl, 20 mM HEPES pH 7.5
~55 in 100 mM NaCl, 20 mM HEPES pH 7.5
MW510.61005.2496.6454.4468.5
CMC %w/v (mM)0.009% (0.17) in water0.001% (~0.01)0.03% (~0.59)0.89% (~19.5) in 100 mM NaCl, 20 mM HEPES pH 7.50.28% (~6)
Compatible with Detergent Removal ResinYesNoYesNoYes
Available size(s)1 g
5 g
1 g (solid)
10 x 1 mL (liquid)
1 g
5 g
1 g
5 g
1 g
5 g
See all maltoside detergents
Cyclohexyl maltoside detergents
Cyclohexyl maltosides are designed for the extraction of intact complexes. They can be used with protein purification, crystallization, spectroscopy, and structural studies.
 5-Cyclohexyl-1-Pentyl-β-D-Maltoside6-Cyclohexyl-1-Hexyl-β-D-Maltoside7-Cyclohexyl-1-Heptyl-β-D-Maltoside
Description typeNon-ionic
Functional propertiesIdeal for membrane protein complexes, solubilization efficiency depends on specific protein, structural features, and experimental conditions.Increased hydrophobicity may result in enhanced solubilization of higher hydrophobic regions or multiple transmembrane domains. Well suited for structural biology studies.Provides improved stability, maintaining structural integrity and functionality. Well suited for structural biology studies.
Dialyzable YesYesYes
Aggregation number*~47~91~150
MW494.5508.5522.5
CMC %w/v (mM)0.12% (~2.4–5.0)0.028% (~0.56)0.0099% (~0.19)
Compatible with Detergent Removal ResinYesYesYes
Available size(s)1 g
5 g
1 g
5 g
1 g
5 g
See all cyclohexyl maltoside detergents
*Aggregation number is the number of molecules per micelle
Glucoside and other non-ionic detergents 
Glucosides are gentler detergents that enhance solubilization of hydrophobic biomolecules. They can be used with protein purification, crystallization, and structural studies. Tetraethylene Glycol Monooctyl Ether (C8E4) is good for solubilization of sensitive biomolecules. It can be used in native mass spectrometry.
 n-Nonyl-β-D-glucoside (NG)Octyl-β-Glucoside Detergent (OG)Octylthioglucoside (OTG)Tetraethylene Glycol Monooctyl Ether (C8E4)
Description typeNon-ionic
Functional propertiesContains a longer alkyl chain resulting in higher hydrophobicity, enhancing solubilization and improving extraction of membrane proteins.
Widely used for membrane protein solubilization
Advantageous for application such as protein purification and structural studies since they provide better solubility and reduced downstream interference.
Effective for cell lysis and nondenaturing protein solubilization
Useful for applications where enzyme stability is desired since it can remain stable in the presence of beta-D-glucoside glucohydrolase enzymes.
Good for solubility in both aqueous and organic solvents
Ideal for sensitive biomolecules, and membrane proteins or protein complexes used for MS.
Dialyzable YesYesYesYes
Aggregation number*Unknown27Unknown~82
MW308.4292.444306.5
CMC %w/v (mM)0.20% (6.5)0.67–0.73% (23-25)0.28% (9)0.25% (~8)
Cloud point (°C)Unknown>100>100Unknown
Compatible with Detergent Removal ResinYesYesYesYes
Available size(s)1 g
5 g
5 g5 g10 mL
See all glucoside detergents
* Aggregation number is the number of molecules per micelle
Zwitterionic detergents 
Tetraethylene Glycol Monooctyl Ether (C8E4), Lauryldimethylamine-N-oxide (LDAO), and Fos-Choline detergents can all be used for protein purification and structural studies. LDAO are ideal for MS and crystallization studies; while fos-choline is also used for membrane protein research. 
 Lauryldimethylamine-N-oxide (LDAO)Fluorinated Fos-Choline-8Fos-choline-12Fos-choline-15
Description typeZwitterionic
Functional propertiesIdeal for the study of intact membrane protein structure and non-covalent interactions.Efficient at solubilization of challenging and delicate membrane proteins. 
Ideal for sensitive membrane proteins, minimizes potential cytotoxic effects such as denaturation and aggregation.Optimal for wide range of membrane protein types by maintaining native conformation and functionality of proteins.Increased hydrophobicity results in enhanced solubilization and stabilization of larger hydrophobic regions or transmembrane domains.
Dialyzable YesYesYesYes
Aggregation number*~76Unknown~54~131
MW229.4529.2351.5393.5
CMC % w/v (mM)0.023% (~1-2)-- (~2.9)0.047% (1.5)0.0027% (0.07)
Compatible with Detergent Removal ResinYesYesYesNo
Available size(s)1 g
5 g
1 g
5 g
1 g
5 g
1 g
5 g
See all fos-choline detergents
* Aggregation number is the number of molecules per micelle
Not worried about maintaining protein function? See our detergents for protein solubilization
Polymers for detergent-free membrane protein isolation
Polymers represents a cutting-edge approach to membrane protein extraction, offering a detergent-free method for isolating membrane proteins along with their surrounding lipid bilayer. This innovative technology provides researchers with a powerful tool for studying membrane proteins in their native lipid environment, potentially leading to more accurate structural and functional analyses. 
Like detergents, it is important to understand polymer characteristics in order to determine the best polymer for membrane protein target and intended experimental goal. A recent publication highlights the importance of selecting a polymer that effectively interacts with the specific lipid environment to maintain protein stability and functionality (1).
Characteristics of membrane protein extraction polymers
Attribute​Unit​SMALP® 200​SMALP® 300​
Polymer styrene-to-maleic anhydride ratio​n:m​2:1​3:1​
Molecular weight​Da​6,500​10,000​
Dry solids content​% w/w​20​20​
Solvent​Water​Water ​
pH​7.5​8.0​
Absolute viscosity​mPaꞏs​≤ 5,000​≤ 5,000​
 See all SMALP 200 polymersSee all SMALP 300 polymers
Ordering information
Catalog # Name Size Price (USD) Qty
A65509 n-Undecyl-β-D-maltopyranoside (UDM), 1 g, 1 g Each
206.65
Online Exclusive
219.00
Save 12.35 (6%)
89903 n-Dodecyl-β-D-Maltoside Detergent, 5 g, 5 g Each
545.65
Online Exclusive
582.00
Save 36.35 (6%)
A65514 n-Nonyl-Beta-Maltoside (NM), 5 g, 5 g Each
540.65
Online Exclusive
574.00
Save 33.35 (6%)
89902 n-Dodecyl-β-D-Maltoside Detergent, 1 g, 1 g Each
173.65
Online Exclusive
184.00
Save 10.35 (6%)
A50940 Lauryl Maltose Neopentyl Glycol (LMNG), 1 g Each
148.65
Online Exclusive
160.00
Save 11.35 (7%)
A50941 LMNG/CHS Solution (10:1), 10 x 1 mL Each
231.65
Online Exclusive
252.00
Save 20.35 (8%)
A65510 n-Undecyl-β-D-maltopyranoside (UDM), 5 g, 5 g Each
773.65
Online Exclusive
822.00
Save 48.35 (6%)
A65511 n-Octyl-β-D-Maltopyranoside (OM), 1 g, 1 g Each
170.65
Online Exclusive
181.00
Save 10.35 (6%)
A65512 n-Octyl-β-D-Maltopyranoside (OM), 5 g, 1 g Each
540.65
Online Exclusive
574.00
Save 33.35 (6%)
A65513 n-Nonyl-Beta-Maltoside (NM), 1 g, 1 g Each
170.65
Online Exclusive
181.00
Save 10.35 (6%)
Catalog # Name Size Price (USD) Qty
28351 Octylthioglucoside (OTG) Detergent, 5 g, 5 g Each
326.65
Online Exclusive
353.00
Save 26.35 (7%)
A65515 n-Nonyl-β-D-glucoside (NG), 1 g, 1 g Each
152.65
Online Exclusive
164.00
Save 11.35 (7%)
A65516 n-Nonyl-β-D-glucoside (NG), 5 g, 5 g Each
507.65
Online Exclusive
548.00
Save 40.35 (7%)
28310 Octyl-beta-Glucoside Detergent, 5 g, 5 g Each
513.65
Online Exclusive
554.00
Save 40.35 (7%)
A65524 Tetraethylene Glycol Monooctyl Ether (C8E4), LC/MS grade Each
548.00
Catalog # Name Size Price (USD) Qty
A40002806 SMALP 200, 5 x 100 mg 5 x 100 mg
343.00
A40002805 SMALP 200, 1 g 1 g
634.00
A40002802 SMALP 300, 5 x 100 mg 5 x 100 mg
343.00
A40002801 SMALP 300, 5 x 1 g 5 x 1 g
2,850.00
A40002804 SMALP 300, 1 g 1 g
634.00
A40002803 SMALP 200, 5 x 1 g 5 x 1 g
2,850.00
Citations
Discover protein preparation tools
Protein sample preparation technical handbook
The Protein Preparation Handbook provides useful information on our broad selection of reagents and tools for protein extraction, clean-up, immunoprecipitation, and purification. Practical information, selection guides, and relevant data are included to help you improve your protein yield and downstream analysis.
protein preparation handbook
Protein sample preparation eLearning course
This modular, animated, and narrated eLearning course was developed to provide a succinct, contextual summary of common laboratory methods and techniques required to achieve optimal results in protein assays and experiments. There are knowledge checks throughout the course to test what you have learned.
eLearning courses
Hero banner dark gradient overlay
Stylesheet for Classic Wide Template adjustments
Hero banner dark gradient overlay
For Research Use Only. Not for use in diagnostic procedures.