48ab - Appl/Samples/Products/Resources/Contact
The ability to perform compositional analysis, and visualize the resulting chemical maps in 3D, is essential to obtain the true elemental distribution or composition of a material, ultimately delivering new insights into the structure-function relationship of the sample.
Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. Full 3D characterization includes chemical as well as imaging data, making 3D energy dispersive X-ray spectroscopy (EDS, also abbreviated EDX or XEDS) an indispensable technique. For the highest quality results, instrumentation with dynamic high-resolution imaging capabilities, as well as fast and quantitative data acquisition, is therefore required. The combination of flexibility in acquisition schemes (TEM, STEM, and EDS), the ability to easily and reproducibly optimize the experiment, and the fast and highly sensitive collection of the elemental distribution data are prerequisites for capturing the real 3D structure and composition of nanomaterials.
Electron tomography produces 3D reconstructions of materials by incrementally adjusting the angle at which the sample is observed. This produces a tilt series of images that can be digitally back projected to render the original sample volume. EDS spectra can be obtained alongside the the electron microscopy (EM) images, providing detailed elemental context. Below are just a few examples of EDS tomography use cases, covering a wide variety of scales, resolutions, and applications.
Thermo Fisher Scientific instruments offer a range of automation capabilities, allowing you to predetermine mapping conditions, drift compensation, and detector parameters, as well as autofocus and auto tilt conditions. This level of automation enables you to set up the EDS tomography experiment and then leave the system unattended for the complete data acquisition process. Visualization and reconstruction were performed with Thermo Scientific Inspect 3D and Avizo Software.
Resources
Video Player is loading.
Current Time 0:00
Duration 0:15
Loaded: 62.66%
Stream Type LIVE
Remaining Time 0:15
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    EDS tomography of P-Zn-In nanotubes, which are used as an electrode material for Na-ion and Li-ion batteries. The segregation of zinc has not been well known during synthesis; however, the elemental data clearly reveals the distribution of zinc relative to the other elements. It becomes evident that there is almost no concentration of zinc in the straight nanotubes. Sample courtesy of Dr. Reza Shahbazian Yassar, Michigan Tech University.
    3D EDS TEM tomography of precipitates in an AlMgSi alloy. Sample courtesy of Thomas Kremmer and Stefan Pogatscher, University of Leoben, Austria.
    Video Player is loading.
    Current Time 0:00
    Duration 0:08
    Loaded: 100.00%
    Stream Type LIVE
    Remaining Time 0:08
    1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
      • en (Main), selected
      EDS tomography data of organic nanoparticles in a polymer sheet.
      Video Player is loading.
      Current Time 0:00
      Duration 0:37
      Loaded: 25.94%
      Stream Type LIVE
      Remaining Time 0:37
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        EDS tomography data of a catalyst powder. Sample courtesy of Tampere University of Technology, Finland.
        Video Player is loading.
        Current Time 0:00
        Duration 0:28
        Loaded: 35.45%
        Stream Type LIVE
        Remaining Time 0:28
        1x
          • Chapters
          • descriptions off, selected
          • captions off, selected
          • default, selected
          EDS tomography data of copper indium sulfide (CuInS2) nanostructures. Sample courtesy of Prof. Neerish Revaprasadu, University of Zululand, South Africa.
          Video Player is loading.
          Current Time 0:00
          Duration 1:35
          Loaded: 10.33%
          Stream Type LIVE
          Remaining Time 1:35
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Example walkthrough of the EDS tomography workflow on the Talos S/TEM.
            Video Player is loading.
            Current Time 0:00
            Duration 0:48
            Loaded: 20.83%
            Stream Type LIVE
            Remaining Time 0:48
            1x
              • Chapters
              • descriptions off, selected
              • captions off, selected
              • default, selected
              STEM and EDS tomography showing the distribution of the Palladium particles (red) relative to other elements of the vehicle aged catalyst material.
              Video Player is loading.
              Current Time 0:00
              Duration 0:19
              Loaded: 49.62%
              Stream Type LIVE
              Remaining Time 0:19
              1x
                • Chapters
                • descriptions off, selected
                • captions off, selected
                • en (Main), selected
                Digital slice of the reconstructed Ag-Pt core shell nanoparticle volume. Sample courtesy Prof. Yi Ding and Prof. Jun Luo, Center for Electron Microscopy, Tianjin University of Technology.
                Video Player is loading.
                Current Time 0:00
                Duration 0:59
                Loaded: 16.68%
                Stream Type LIVE
                Remaining Time 0:59
                1x
                  • Chapters
                  • descriptions off, selected
                  • captions off, selected
                  • en (Main), selected
                  Segmented surface rendering of nanoparticles with elements present: Ag core, Platinum shell (to increase visibility, the Platinum shells have been colored semitransparent).
                  Video Player is loading.
                  Current Time 0:00
                  Duration 0:15
                  Loaded: 62.66%
                  Stream Type LIVE
                  Remaining Time 0:15
                  1x
                    • Chapters
                    • descriptions off, selected
                    • captions off, selected
                    • en (Main), selected
                    EDS tomography of P-Zn-In nanotubes, which are used as an electrode material for Na-ion and Li-ion batteries. The segregation of zinc has not been well known during synthesis; however, the elemental data clearly reveals the distribution of zinc relative to the other elements. It becomes evident that there is almost no concentration of zinc in the straight nanotubes. Sample courtesy of Dr. Reza Shahbazian Yassar, Michigan Tech University.
                    3D EDS TEM tomography of precipitates in an AlMgSi alloy. Sample courtesy of Thomas Kremmer and Stefan Pogatscher, University of Leoben, Austria.
                    Video Player is loading.
                    Current Time 0:00
                    Duration 0:08
                    Loaded: 100.00%
                    Stream Type LIVE
                    Remaining Time 0:08
                    1x
                      • Chapters
                      • descriptions off, selected
                      • captions off, selected
                      • en (Main), selected
                      EDS tomography data of organic nanoparticles in a polymer sheet.
                      Video Player is loading.
                      Current Time 0:00
                      Duration 0:37
                      Loaded: 25.94%
                      Stream Type LIVE
                      Remaining Time 0:37
                      1x
                        • Chapters
                        • descriptions off, selected
                        • captions off, selected
                        • en (Main), selected
                        EDS tomography data of a catalyst powder. Sample courtesy of Tampere University of Technology, Finland.
                        Video Player is loading.
                        Current Time 0:00
                        Duration 0:28
                        Loaded: 35.45%
                        Stream Type LIVE
                        Remaining Time 0:28
                        1x
                          • Chapters
                          • descriptions off, selected
                          • captions off, selected
                          • default, selected
                          EDS tomography data of copper indium sulfide (CuInS2) nanostructures. Sample courtesy of Prof. Neerish Revaprasadu, University of Zululand, South Africa.
                          Video Player is loading.
                          Current Time 0:00
                          Duration 1:35
                          Loaded: 10.50%
                          Stream Type LIVE
                          Remaining Time 1:35
                          1x
                            • Chapters
                            • descriptions off, selected
                            • captions off, selected
                            • en (Main), selected
                            Example walkthrough of the EDS tomography workflow on the Talos S/TEM.
                            Video Player is loading.
                            Current Time 0:00
                            Duration 0:48
                            Loaded: 20.83%
                            Stream Type LIVE
                            Remaining Time 0:48
                            1x
                              • Chapters
                              • descriptions off, selected
                              • captions off, selected
                              • default, selected
                              STEM and EDS tomography showing the distribution of the Palladium particles (red) relative to other elements of the vehicle aged catalyst material.
                              Video Player is loading.
                              Current Time 0:00
                              Duration 0:19
                              Loaded: 49.62%
                              Stream Type LIVE
                              Remaining Time 0:19
                              1x
                                • Chapters
                                • descriptions off, selected
                                • captions off, selected
                                • en (Main), selected
                                Digital slice of the reconstructed Ag-Pt core shell nanoparticle volume. Sample courtesy Prof. Yi Ding and Prof. Jun Luo, Center for Electron Microscopy, Tianjin University of Technology.
                                Video Player is loading.
                                Current Time 0:00
                                Duration 0:59
                                Loaded: 16.52%
                                Stream Type LIVE
                                Remaining Time 0:59
                                1x
                                  • Chapters
                                  • descriptions off, selected
                                  • captions off, selected
                                  • en (Main), selected
                                  Segmented surface rendering of nanoparticles with elements present: Ag core, Platinum shell (to increase visibility, the Platinum shells have been colored semitransparent).
                                  Applications
                                  Process control using electron microscopy
                                  Process control using electron microscopy
                                  Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
                                  Quality control and failure analysis using electron microscopy
                                  Quality control and failure analysis
                                  Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
                                  Aluminum mineral grain found with SEM during parts cleanliness testing
                                  Technical Cleanliness
                                  More than ever, modern manufacturing necessitates reliable, quality components. With scanning electron microscopy, parts cleanliness analysis can be brought inhouse, providing you with a broad range of analytical data and shortening your production cycle.
                                  Fundamental Materials Research_R&D_Thumb_274x180_144DPI
                                  Fundamental Materials Research
                                  Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
                                  Samples
                                  Battery Research
                                  Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
                                  Learn more ›
                                  Polymers Research
                                  Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
                                  Learn more ›
                                  Metals Research and Characterization
                                  Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
                                  Learn more ›
                                  Nanoparticles
                                  Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
                                  Learn more ›
                                  Oil and Gas
                                  As the demand for oil and gas continues, there is an ongoing need for efficient and effective extraction of hydrocarbons. Thermo Fisher Scientific offers a range of microscopy and spectroscopy solutions for a variety of petroleum science applications.
                                  Learn more ›
                                  Geological Research
                                  Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
                                  Learn more ›
                                  Catalysis Research
                                  Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
                                  Learn more ›
                                  Automotive Materials Testing
                                  Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
                                  Learn more ›
                                  Fibers and Filters
                                  The diameter, morphology and density of synthetic fibers are key parameters that determine the lifetime and functionality of a filter. Scanning electron microscopy (SEM) is the ideal technique for quickly and easily investigating these features.
                                  Learn more ›
                                  Products
                                  Style Sheet for Instrument Cards Original
                                  Iliad (S)TEM
                                  • Advanced integration of EELS and (S)TEM optics
                                  • Electrostatic beam blanker
                                  • High energy resolution electron source
                                  Helios 5 Laser PFIB System
                                  • Fast, millimeter-scale cross sections
                                  • Statistically relevant deep subsurface and 3D data analysis
                                  • Shares all capabilities of the Helios 5 PFIB platform
                                  Spectra Ultra
                                  • New imaging and spectroscopy capabilities on the most beam sensitive materials
                                  • A leap forward in EDX detection with Ultra-X
                                  • Column designed to maintain sample integrity.
                                  Spectra 300
                                  • Highest-resolution structural and chemical information at the atomic level
                                  • Flexible high-tension range from 30-300 kV
                                  • Three lens condenser system
                                  Spectra 200
                                  • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
                                  • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
                                  • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
                                  Thermo Scientific Talos F200C transmission electron microscope (TEM)
                                  Talos F200C TEM
                                  • High-contrast and high-quality TEM and STEM imaging
                                  • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
                                  • Large pole piece gap and multiple in situ options
                                  Thermo Scientific Talos L120C transmission electron microscope (TEM)
                                  Talos L120C TEM
                                  • High versatility and stability
                                  • 4k x 4k Ceta CMOS camera for speed and large FOV
                                  • TEM magnification range of 25X to 650kX
                                  • EDS and STEM options for compositional analyses
                                  Talos F200i TEM
                                  • Compact design with X-TWIN objective lens
                                  • Available with S-FEG, X-FEG, and X-CFEG
                                  • Flexible and fast EDS options for comprehensive elemental analysis
                                  Talos F200S TEM
                                  • Intuitive and easy-to-use automation software
                                  • Available with Super-X EDS for rapid quantitative chemical analysis
                                  • High-throughput with simultaneous multi-signal acquisition
                                  Talos F200X TEM
                                  • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
                                  • X-FEG and X-CFEG available for the highest brightness and energy resolution
                                  • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
                                  Phenom ParticleX Battery Desktop SEM
                                  • Versatile solution for high-quality, in-house analysis
                                  • Automated system and analysis of multiple samples
                                  • Testing 10x faster
                                  Velox
                                  • An experiments panel on the left side of the processing window.
                                  • Live quantitative mapping
                                  • Interactive detector layout interface for reproducible experiment control and setup
                                  Thermo Scientific Inspect 3D tomography software
                                  Inspect 3D Software
                                  • Image processing tools and filters for cross-correlation
                                  • Feature tracking for image alignment
                                  • Algebraic reconstruction technique for iterative projection comparison
                                  Avizo Software
                                  Materials Science
                                  • Support for multi-data/multi-view, multi-channel, time series, very large data
                                  • Advanced multi-mode 2D/3D automatic registration
                                  • Artifact reduction algorithms
                                  Style Sheet for Komodo Tabs
                                  Style Sheet to change H2 style to p with em-h2-header class
                                  Contact us
                                  Style Sheet for Support and Service footer
                                  Style Sheet for Fonts
                                  Style Sheet for Cards
                                  Electron microscopy services for
                                  the materials science
                                  To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.