Organism Type | Human | |
Tissues | Ovary |
Phenotype | Adherent |
Primary | no |
Application | Cell Culture / Growth Conditions,Protein Expression,Stable Cell Transfection,Transient Transfection |
Recommended Transfection Reagents
- Plasmid DNA transfection and co-transfection: Lipofectamine® 3000 (30–50% transfection efficiency and 3 fold protein expression improvement over Lipofectamine® 2000)
- RNAi transfection (siRNA/miRNA) and gene silencing: Lipofectamine® RNAiMAX
10131027,11668019,11668027,11668075,11811023,18324012,18324111,
Products
10131027,11668019,11668027,11811023,18324012,18324111