It starts with the sample container
Making the right selection of your centrifuge, rotor, buckets and adapters starts with the right sample container. Some rotors offer exceptional versatility with different adapters available to accommodate various formats, e.g. bottles, tubes, microplates. Some rotors might only be used for one sample container type, if there is only one application in the lab. With the forces a centrifuge runs your samples with, it is critical to select the adapter not only matching to the right volume, but also the shape of your sample tube/ bottle.
General accessories
Buckets, adapters and lids
Maximize throughput, convenience, and performance with the exact accessories for your rotor and application. Choose from a wide selection of adapter shapes and sizes for both swinging bucket and fixed angle rotors, and buckets for swinging bucket rotors. For biocontainment, ClickSeal lids are sized to fit many bucket shapes and fixed angle rotors.
centrifugeware
Consumables
You can find the matching container for your centrifuge separation application, from large-volume pelleting to DNA isolation, with Thermo Scientific Nalgene and Nunc centrifuge ware. We offer a broad selection of tubes and bottles in sizes from 10 mL to 2 L, in conical and round-bottom profiles, with a choice of secure closures.
Superspeed centrifuge consumables
Thermo Scientific Fiberlite Bottles are designed for the speeds and RCFs of our superspeed centrifuges and are excellent for bacterial and yeast isolations and viral harvesting. Choose from 250 mL, 500 mL, and 1,000 mL capacities in polypropylene copolymer and polycarbonate materials. Additionally, the 1L SteriBIOS bottles provide a sterilized, single use option; while the Sorvall HP bottles offer improved chemical resistance to phenol solutions.
Specifications
Fiberlite Bottle​
SteriBIOS Bottle​
Sorvall HP Bottle​
Maximum capacity/ bottle size​
1,000 mL
500 mL
250 mL
1,000 mL​
1,000 mL
Materials:
Bottles​
Cap​
Closure​
O-Rings
  • PC / PPCO​
  • Glass filled Nylon​
  • PP​
  • Silicone​
  • PPCO​
  • Glass filled Nylon​
  • PP​
  • Silicone​
  • PC / PPCO​
  • Aluminum​
  • PP​
  • Silicone​
Max speed / RCF
​(Compatible rotors)​
1,000 mL bottles​
10,500 / 20,584 (F10-4x1000 LEX)​
9,000 / 17,568 (F9-6x1000 LEX)​
6,250 / 12,000 (F6-10x1000 LEX)​
Bottle lifetime​
100 cycles​
Single use​
50 cycles ​
Autoclavable
Yes​ (PPCO)
N/A​
Yes​ (PPCO)
Certifications
  • USP Class VI per USP <88>
  • Non-cytotoxic per USP <87>
  • BSE / TSE transmission safe​
  • USP Class VI per USP <88>
  • Non-cytotoxic per USP <87>​
  • BSE / TSE transmission safe
  • Endotoxin per USP <85>​
  • Particle count per USP <788>​
  • Sterilized following ISO11137-1 and ISO1137-2​
  • USP Class VI per USP <88>
  • Non-cytotoxic per USP <87>​
  • BSE / TSE transmission safe​
Ultracentrifuge accessories
Since ultracentrifuges spin at high speeds to up to 100,000 rpm, consumables are specifically designed for stability. The following tubes are available in many different sizes to match your application needs.
Specifications
Thermo Scientific Ultracrimp and Clearcrimp Tubes
Open Top Thin Wall Style Tubes
Threaded Thick Wall Tubes​
Available size​s (note: maximum fill may be lower)
Fixed angle rotors - 6mL (5.7mL), 11.5mL (11.5mL), 35mL (35mL), 90mL (89mL)
Vertical Rotors – 2.0mL, 6.0mL, 13.5mL, 39.0mL
Fixed angle rotors – 2.0mL, 4.4mL, 5.0mL, 6.5mL, 12.5mL, 36.0mL
Swinging Bucket rotors – 4.4mL, 5.0mL, 12.0mL, 13.2mL 17.0mL, 36.0mL, 38.5mL
 13.5mL, 39.0mL, 100mL ​
Materials:
Tube/Bottle​
Plug​
Cap/Crown​
O-Ring​
  • Polyallomer (PA)/ Polyethylene terephthalate (PET)
  • PP​
  • Aluminum
  •   -
  • Polyallomer (PA)/ Polyethylene terephthalate (PET)
  •   -
  •   -
  •   -
  • PC​
  •   -
  • Aluminum​
  • Nitrile
Max speed / RCF
​Compatible rotors
Fixed angle rotors
90mL (PA) – 
47,500 / 250,259 (T-647.5)​
35mL (PA) – 
50,000 / 301,032 (T-1250)
60,000 / 340,596 (TV-860)
65,000 / 429,459 (T-865)
11.5mL (PA & PET) – 
90,000 / 692,149 (T-890)
70,000 / 448,811 (T-1270)
30,000 / 91,482 (T-865)
27,500 / 91,062 (T-1250)
6mL (PA & PET) – 
100,000 / 802,006 (T-8100)
45,000 / 238,859 (FT-45.6)
65,000 / 414,357 (TV-1665)
Vertical rotors 
39.0mL (PA & PET) – 
50,000 / 241,831 (StepSaver 50V39)
13.5mL (PA & PET) – 
50,000 / 228,706 (StepSaver 50V39)
65,000 / 400,617 (StepSaver 65V13)
6.0mL (PA) –  
65,000 / 393,120 (StepSaver 65V13)
70,000 / 467,420 (StepSaver 70V6)
2.0mL (PA & PET) – 
70,000 / 467,420 (StepSaver 70V6)
Fixed angle rotors
36.0mL (PA) – 
65,000 / 429,459 (T-865)
50,000 / 301,032 (T-1250)
12.5mL (PA) – 
75,000 / 480,659 (T-890)
70,000 / 448,811 (T-1270)
6.5mL (PA) –  
80,000 / 513,284 (T-8100)
5.0mL (PA) –  
65,000 / 414,357 (TV-1665)
4.4mL (PA) –  
80,000 / 468,246 (TFT80.4)
2.0mL (PA) – 
80,000 / 429,643 (TFT80.2)
Swing bucket rotors
38.5mL (PET) – 
32,500 / 187,295 (SureSpin 632)​
30,000 / 166,880 (SureSpin 630)​
29,000 / 151,243 (AH-629)
36mL (PA) – 
32,500 / 187,295 (SureSpin 632)​
30,000 / 166,880 (SureSpin 630)​
29,000 / 151,243 (AH-629)
17.0mL (PA & PET) – 
32,000 / 191,759 (SureSpin 632)​
30,000 / 166,880 (SureSpin 630)​
29,000 / 155,846 (AH-629)
13.2mL (PA) – 
41,000 / 287,660 (TH-641)
12.0mL (PET) – 
41,000 / 287,660 (TH-641)
5.0mL (PA) – 
50,000 / 296,005 (AH-650)
4.4mL (PA) – 
60,000 / 488,576 (TH-660)
100mL – 
37,000 / 182,460 (F37L-8x100)​
39mL – 
50,000 / 266,000 (F50L-8x39)
30,000 / 119,950 (F37L-8x100)
13.5mL
30,000 / 119,950 (F37L-8x100)
65,000 / 324,000 (F65L-6x13.5)
Tube lifetime​
Single use
Reusable (Only if not punctured or no significant deformation or stress cracking has occurred) 
Reusable 
Autoclavable
Yes​ (PA tubes)
Yes
Yes
Certifications      
  For 6mL, 11.5mL, 35mL & 90mL (PA tubes) 
  • USP Class VI per USP <88>
  • Non-cytotoxic per USP <87>​
  • BSE / TSE transmission safe​
Ultracrimp and clearcrimp tubes require a sealing tool.
Large capacity consumables
Thermo Scientific 2 liter bottles are designed for harvesting large volumes in our bioprocessing centrifuges. Harvest up to 16 liters in a single run in a swinging bucket rotor with these translucent polypropylene bottles.
CentriPAK BPC (Bioprocessing Container) provides a comprehensive, closed-system solution to safeguard samples throughout the bioprocessing workflow. The CentriPAK BPC is designed for single-use and undergoes gamma-irradiation sterilization, eliminating the need for post-use cleaning typically associated with reusable containers.
Specifications
2,000 mL Oval Bottle​
(Cat. No. 75003870)
2,000 mL BIOS Bottle​
(Cat. No. 75003872)
2,000 mL SteriBIOS Bottle
(Cat. No. 75003871)
CentriPAK BPC​
Maximum capacity/ bottle size​  
2,000 mL
2,000 mL
2,000 mL
10.7 L (1.7 L each)​
Materials:  
Bottles​
Closure​
Film​
  • PPCO​
  • PPCO​
  •   -
  • PP
  • PP
  •    -
  • PP
  • PP
  •   -
  •   -
  • CX3-9
  •     -
Max speed / RCF  
​(Compatible rotors)​
3,900 / 5,374 (8x2000 Large Capacity Swinging Bucket Rotor)​
4,700 / 7,187 (6x2000 Large Capacity Swinging Bucket Rotor)
3,900 / 5,374 (8x2000 Large Capacity Swinging Bucket Rotor)​
4,700 / 7,187 (6x2000 Large Capacity Swinging Bucket Rotor)
3,900 / 5,374 (8x2000 Large Capacity Swinging Bucket Rotor)​
4,700 / 7,187 (6x2000 Large Capacity Swinging Bucket Rotor)​
4,700 / 7,187 (6x2000 and 6x8000 Large Capacity Swinging Bucket Rotors)​
Bottle / BPC lifetime​  
500 cycles​
50 cycles​
Single use​
Single use​
Autoclavable  
Yes (Bottle and Closure)​ ​
Yes (Bottle and Closure)​ ​
N/A​
N/A​
Certifications  
  • Food grade material
  • USP Class VI per USP <88>
  • Non-cytotoxic per USP <87>​
  • BSE / TSE transmission safe​
  • USP Class VI per USP <88>
  • Non-cytotoxic per USP <87>​
  • BSE / TSE transmission safe​
  • Endotoxin per USP <85>​
  • Particle count per USP <788>​
  • Sterilized following ISO 11137-1 and ISO 1137-2
  • USP Class VI per USP <88>
  • Non-cytotoxic per USP <87>​
  • BSE / TSE transmission safe
  • Endotoxin per USP <85>​
  • Particle count per USP <788>​
  • Sterilized following ISO 11137-1:2006​
Intended use of the products mentioned on this page vary. For specific intended use statements please refer to the product label.