Desktop SEM for component cleanliness analysis
The Thermo Scientific Phenom ParticleX TC Desktop SEM is a multi-purpose desktop SEM enabling technical cleanliness at the microscale.
Materials characterization
A versatile solution for high-quality, in-house analysis, the Phenom ParticleX Desktop SEM gives you the ability to carry out speedy characterization, verification and classification of materials, supporting your production with fast, accurate and trusted data. The system is simple to operate and fast to learn, opening up particle and material analysis to a wider group of users.
Video Player is loading.
Current Time 0:00
Duration 2:59
Loaded: 5.58%
Stream Type LIVE
Remaining Time 2:59
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Thermo Scientific Phenom ParticleX TC demo
    Particle Analysis Learning Center
    Learn more
    Insights
    See how the Phenom ParticleX TC Desktop SEM performs in the real world with case studies, webinars, and more.
    Case study: The real reason why the automotive industry needs technical cleanliness analysis
    Read more
    Datasheet: Phenom Particle X TC
    Read more
    Brochure: EDS for Desktop SEMs
    Read more
    Blog article: Steel Surface Finish
    Read more
    Technical cleanliness webinar
    Webinar
    Technical cleanliness analysis with an SEM Why
    Watch webinar ›
    Key Features
    Technical cleanliness
    With the growing demand for analysis of smaller particles beyond the scope of light microscopy within (automotive) industries, the Phenom ParticleX TC (Technical Cleanliness) Desktop SEM enables automated scanning electron microscopy with energy-dispersive X-ray spectroscopy (EDS). This is a major advantage over light microscopy as it enables chemical classification of the particles, providing great insights in your production processes and/or environments. Standard reports compliant with VDA 19 / ISO 16232 or ISO 4406/4407 are available.
    Secondary electron detector
    A secondary electron detector (SED) is optionally available on the Phenom ParticleX TC (Technical Cleanliness) Desktop SEM. The SED collects low-energy electrons from the top surface layer of the sample. It is therefore the perfect choice to reveal detailed sample surface information. The SED can be of great use for applications where topography and morphology are important. This is often the case when studying microstructures, fibers or particles.
    General SEM usage
    The user interface is based on the proven ease-of-use technology applied in the successful Phenom desktop SEM products. The interface enables both existing and new users to quickly become familiar with the system with a minimum of training. 
    Elemental mapping and line scan
    For a user, it is simply click and go to work with the elemental mapping and line scan functionality of the Phenom ParticleX TC (Technical Cleanliness) Desktop SEM. The line scan functionality shows the quantified element distribution in a line plot. 
    ChemiSEM Technology
    Thermo Scientific ChemiSEM Technology revolutionizes and simplifies EDS analysis by fully integrating SEM and EDS functions into a single, cohesive user interface. Based on live quantification and building on decades of expertise in EDS analysis, the technology provides elemental information quickly and easily, guaranteeing reliable results in less time. ChemiSEM Technology now comes with a powerful new feature: ChemiPhase. ChemiPhase identifies unique phases with a big data approach, finding minor and trace elements while eliminating user bias and reducing possible mistakes.
    Learn more
    Specifications
    Style Sheet for Products Table Specifications
    Electron optical
    • Long lifetime thermionic source (CeB6 )
    • Multiple beam currents
    Electron optical magnification range
    • 160 - 200,000x
    Light optical magnification
    • 3–16x
    Resolution
    • <10 nm
    Image resolution options
    • 960 x 600, 1920 x 1200, 3840 x 2400 and 7680 x 4800 pixels
    Acceleration voltages
    • Default: 5 kV, 10 kV and 15 kV
    • Advanced mode: adjustable range between 4.8 kV and 20.5 kV imaging and analysis mode
    Vacuum levels
    • Low - medium - high
    Detector
    • Backscattered electron detector (standard)
    • Energy-dispersive X-ray spectroscopy (EDS) detector (standard)
    • Secondary electron detector (optional)
    Sample size
    • Max. 100 mm x 100 mm (up to 36 x 12 mm pin stubs)
    • Max. 40 mm (h)
    Sample loading time
    • Light optical <5 s 
    • Electron optical <60 s
    Industrial Manufacturing Product Selector
    The product selector is here to assist you in choosing the most suitable Scanning Electron Microscope (SEM) system and software for your research. Find out in a few minutes which SEM best suits your research application:
    Find your ideal Desktop SEM
    Resources
    Video Player is loading.
    Current Time 0:00
    Duration 1:13
    Loaded: 13.58%
    Stream Type LIVE
    Remaining Time 1:13
    1x
      • Chapters
      • descriptions off, selected
      • en (Main), selected
      Alexander Bouman on the new Phenom ParticleX
      Video Player is loading.
      Current Time 0:00
      Duration 2:54
      Loaded: 5.65%
      Stream Type LIVE
      Remaining Time 2:54
      1x
        • Chapters
        • descriptions off, selected
        • captions off, selected
        • en (Main), selected
        Watch Phenom ParticleX Desktop SEM: Quality Starts at the Microscale
        Watch this on-demand webinar to learn how Phenom ParticleX desktop SEMs are the ideal solution for:
        • ISO 16232 and VDA-19 Compliance for Technical Cleanliness.
        • ISO 4406 and ISO 4407 for contamination of hydraulic fluids.
        • Particle morphology for additive manufacturing feedstocks.
        • Workflow examples for particle classifications and reporting.
        Webinar: Scanning electron microscopy: selecting the right technology for your needs
        This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
        • How the needs for different microanalysis modalities are met (EDX, EBSD, WDS, CL, etc.).
        • How samples are characterized in their natural state without the need for sample preparation.
        • How new advanced automation allows researchers to save time and increase productivity.
        Watch on-demand: Particle Analysis Applications Using Desktop SEM Webinar Series
        In each on-demand webinar, our expert will focus on one particular analysis application and how the Phenom ParticleX Desktop SEM overcomes some of the most common challenges. See the abstracts for individual sessions below.
        • How to certify your NCM powder quality with SEM+EDS
        • Electron-microscope-grade cleanliness in electronics
        • Speed up your automated gunshot residue analysis
        • Technical cleanliness analysis with an SEM: Why?
        • Understand your steel with automated inclusion analysis
        • One tool for complete AM powder characterization
        Video Player is loading.
        Current Time 0:00
        Duration 1:13
        Loaded: 13.36%
        Stream Type LIVE
        Remaining Time 1:13
        1x
          • Chapters
          • descriptions off, selected
          • en (Main), selected
          Alexander Bouman on the new Phenom ParticleX
          Video Player is loading.
          Current Time 0:00
          Duration 2:54
          Loaded: 5.65%
          Stream Type LIVE
          Remaining Time 2:54
          1x
            • Chapters
            • descriptions off, selected
            • captions off, selected
            • en (Main), selected
            Watch Phenom ParticleX Desktop SEM: Quality Starts at the Microscale
            Watch this on-demand webinar to learn how Phenom ParticleX desktop SEMs are the ideal solution for:
            • ISO 16232 and VDA-19 Compliance for Technical Cleanliness.
            • ISO 4406 and ISO 4407 for contamination of hydraulic fluids.
            • Particle morphology for additive manufacturing feedstocks.
            • Workflow examples for particle classifications and reporting.
            Webinar: Scanning electron microscopy: selecting the right technology for your needs
            This on-demand webinar has been designed to help you decide which SEM best meets your unique needs. We present an overview of Thermo Fisher Scientific SEM technology for multi-user research labs and focus on how these wide-ranging solutions deliver performance, versatility, in situ dynamics and faster time to results. Watch this webinar if you are interested in:
            • How the needs for different microanalysis modalities are met (EDX, EBSD, WDS, CL, etc.).
            • How samples are characterized in their natural state without the need for sample preparation.
            • How new advanced automation allows researchers to save time and increase productivity.
            Watch on-demand: Particle Analysis Applications Using Desktop SEM Webinar Series
            In each on-demand webinar, our expert will focus on one particular analysis application and how the Phenom ParticleX Desktop SEM overcomes some of the most common challenges. See the abstracts for individual sessions below.
            • How to certify your NCM powder quality with SEM+EDS
            • Electron-microscope-grade cleanliness in electronics
            • Speed up your automated gunshot residue analysis
            • Technical cleanliness analysis with an SEM: Why?
            • Understand your steel with automated inclusion analysis
            • One tool for complete AM powder characterization
            Applications
            Process control using electron microscopy
            Process control using electron microscopy
            Modern industry demands high throughput with superior quality, a balance that is maintained through robust process control. SEM and TEM tools with dedicated automation software provide rapid, multi-scale information for process monitoring and improvement.
            Quality control and failure analysis using electron microscopy
            Quality control and failure analysis
            Quality control and assurance are essential in modern industry. We offer a range of EM and spectroscopy tools for multi-scale and multi-modal analysis of defects, allowing you to make reliable and informed decisions for process control and improvement.
            Fundamental Materials Research_R&amp;D_Thumb_274x180_144DPI
            Fundamental Materials Research
            Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
            Aluminum mineral grain found with SEM during parts cleanliness testing
            Technical Cleanliness
            More than ever, modern manufacturing necessitates reliable, quality components. With scanning electron microscopy, parts cleanliness analysis can be brought inhouse, providing you with a broad range of analytical data and shortening your production cycle.
            Techniques
            EDS Elemental Analysis
            Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
            Learn more ›
            3D EDS Tomography
            Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
            Learn more ›
            Atomic-Scale Elemental Mapping with EDS
            Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
            Learn more ›
            Particle analysis
            Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
            Learn more ›
            EDS Elemental Analysis
            Thermo Scientific Phenom Elemental Mapping Software provides fast and reliable information on the distribution of chemical elements within a sample.
            Learn more ›
            3D EDS Tomography
            Modern materials research is increasingly reliant on nanoscale analysis in three dimensions. 3D characterization, including compositional data for full chemical and structural context, is possible with 3D EM and energy dispersive X-ray spectroscopy.
            Learn more ›
            Atomic-Scale Elemental Mapping with EDS
            Atomic-resolution EDS provides unparalleled chemical context for materials analysis by differentiating the elemental identity of individual atoms. When combined with high-resolution TEM, it is possible to observe the precise organization of atoms in a sample.
            Learn more ›
            Particle analysis
            Particle analysis plays a vital role in nanomaterials research and quality control. The nanometer-scale resolution and superior imaging of electron microscopy can be combined with specialized software for rapid characterization of powders and particles.
            Learn more ›
            Style Sheet to change H2 style to p with em-h2-header class
            Style Sheet for Komodo Tabs
            Style Sheet for Global Design System
            Contact us
            Electron microscopy services for
            the materials science
            To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.
            Style Sheet for Support and Service footer
            Style Sheet for Fonts
            Style Sheet for Cards