Elemental mapping
Through the combination of atom-sized electron probes with high current and highly sensitive X-ray detectors, routine atomic-resolution spectroscopy is now possible with electron microscopes. Energy-dispersive X-ray spectroscopy (EDS, also abbreviated EDX or XEDS) can identify a wide range of elements due to their unique X-ray signals and is ideally suited for the differentiation of individual atoms. With this information, researchers can characterize and modify their materials at the atomic scale, providing unparalleled insight into the behavior of nanomaterials and particles.
As shown in the yttrium titanate examples on this page, individual atomic positions can be distinguished by their unambiguous chemical signal. The individual atomic columns are not only visible, but distinct from their neighbors due to their high contrast; even mixed and pure columns can be differentiated. Additionally, the EDS signal even makes it possible to detect light elements, which have been notoriously difficult to observe at these resolutions.
Such robust, high-quality results are only possible with Thermo Fisher Scientific’s unique ChemiSTEM Technology and the multi-SSD design of the Dual-X/Super-X Detection Systems. With both improved signal generation and detection, the low probe currents required for atomic-scale spatial resolution no longer pose a barrier to X-ray spectrum collection and analysis.
Y2Ti2O7 atomic elemental mapping with EDS analysis.
Atomic chemical mapping of Y2Ti2O7 in the [110] projection obtained with ChemiSTEM Technology and a probe Cs-corrector at 200 kV acceleration voltage. The maps have a size of 64 x 64 pixels with a mapping time of 306 s. A probe current of 76 pA was used. EDS maps of filtered data using 3-pixel averaging are shown. A model of the structure in [110] projection indicates that three different occupancies are present; pure yttrium, pure titanium, and 50:50 mixed Y/Ti columns.
Style Sheet to change H2 style to p with em-h2-header class
Applications
Fundamental Materials Research_R&D_Thumb_274x180_144DPI
Fundamental Materials Research
Novel materials are investigated at increasingly smaller scales for maximum control of their physical and chemical properties. Electron microscopy provides researchers with key insight into a wide variety of material characteristics at the micro- to nano-scale.
Samples
Battery Research
Battery development is enabled by multi-scale analysis with microCT, SEM and TEM, Raman spectroscopy, XPS, and digital 3D visualization and analysis. Learn how this approach provides the structural and chemical information needed to build better batteries.
Learn more ›
Oil and Gas
As the demand for oil and gas continues, there is an ongoing need for efficient and effective extraction of hydrocarbons. Thermo Fisher Scientific offers a range of microscopy and spectroscopy solutions for a variety of petroleum science applications.
Learn more ›
Metals Research and Characterization
Effective production of metals requires precise control of inclusions and precipitates. Our automated tools can perform a variety of tasks critical for metal analysis including; nanoparticle counting, EDS chemical analysis and TEM sample preparation.
Learn more ›
Nanoparticles
Materials have fundamentally different properties at the nanoscale than at the macroscale. To study them, S/TEM instrumentation can be combined with energy dispersive X-ray spectroscopy to obtain nanometer, or even sub-nanometer, resolution data.
Learn more ›
Polymers Research
Polymer microstructure dictates the material’s bulk characteristics and performance. Electron microscopy enables comprehensive microscale analysis of polymer morphology and composition for R&D and quality control applications.
Learn more ›
Geological Research
Geoscience relies on consistent and accurate multi-scale observation of features within rock samples. SEM-EDS, combined with automation software, enables direct, large-scale analysis of texture and mineral composition for petrology and mineralogy research.
Learn more ›
Automotive Materials Testing
Every component in a modern vehicle is designed for safety, efficiency, and performance. Detailed characterization of automotive materials with electron microscopy and spectroscopy informs critical process decisions, product improvements, and new materials.
Learn more ›
Catalysis Research
Catalysts are critical for a majority of modern industrial processes. Their efficiency depends on the microscopic composition and morphology of the catalytic particles; EM with EDS is ideally suited for studying these properties.
Learn more ›
Products
Style Sheet for Instrument Cards Original
Iliad (S)TEM
  • Advanced integration of EELS and (S)TEM optics
  • Electrostatic beam blanker
  • High energy resolution electron source
Spectra Ultra
  • New imaging and spectroscopy capabilities on the most beam sensitive materials
  • A leap forward in EDX detection with Ultra-X
  • Column designed to maintain sample integrity.
Spectra 200
  • High-resolution and contrast imaging for accelerating voltages from 30-200 kV
  • Symmetric S-TWIN/X-TWIN objective lens with wide-gap pole piece design of 5.4 mm
  • Sub-Angstrom STEM imaging resolution from 60 kV-200 kV
Spectra 300
  • Highest-resolution structural and chemical information at the atomic level
  • Flexible high-tension range from 30-300 kV
  • Three lens condenser system
Thermo Scientific Talos F200C transmission electron microscope (TEM)
Talos F200C TEM
  • High-contrast and high-quality TEM and STEM imaging
  • 4k x 4k Ceta CMOS camera options for large FOV and high read-out speeds
  • Large pole piece gap and multiple in situ options
Talos F200S TEM
  • Intuitive and easy-to-use automation software
  • Available with Super-X EDS for rapid quantitative chemical analysis
  • High-throughput with simultaneous multi-signal acquisition
Talos F200i TEM
  • Compact design with X-TWIN objective lens
  • Available with S-FEG, X-FEG, and X-CFEG
  • Flexible and fast EDS options for comprehensive elemental analysis
Thermo Scientific Talos L120C transmission electron microscope (TEM)
Talos L120C TEM
  • High versatility and stability
  • 4k x 4k Ceta CMOS camera for speed and large FOV
  • TEM magnification range of 25X to 650kX
  • EDS and STEM options for compositional analyses
Talos F200X TEM
  • High-resolution, EDS cleanliness, and quality in 2D as well as 3D
  • X-FEG and X-CFEG available for the highest brightness and energy resolution
  • High accuracy and repeatable results with integrated Thermo Scientific Velox Software
Phenom ParticleX Battery Desktop SEM
  • Versatile solution for high-quality, in-house analysis
  • Automated system and analysis of multiple samples
  • Testing 10x faster
Style Sheet for Media Gallery Tabs
Contact us
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.