lab-essentials-discoveries
Discoveries can happen any day.
So you need to be prepared every day.
Discover the right technique
For standard pipetting use Forward Technique:
Recommended for aqueous solutions, such as buffers, diluted acids, or alkalis, this technique is commonly used when pipetting and mixing a sample or reagent into another liquid.
For solutions with high viscosity or tendency to foam use Reverse Technique:
This technique is commonly used with air displacement pipettes, and is recommended for precisely pipetting small volumes. Reverse pipetting avoids the risk of sample splash, foaming, or bubble formation.
Isn’t all labware the same? No, and let us explain why
There are a lot of plastic labware choices, but the quality of your work often depends on using quality plastic labware. Even though they may look alike, not all labware products are created equal. Lower-cost alternatives come with a nice price, but remember that old saying, “You get what you pay for.”
Regulatory Standards:
High quality plastic labware must meet accepted ASTM and ISO accuracy standards and be made from laboratory-grade plastic materials that won’t leach impurities into your reagents and samples. Containers should provide guaranteed leak-proof performance to keep you safe from exposure and your work safely contained. 
5 Quick Tips | Select the right tip for your application
 Pre-wet the Pipette Tip
 Work at Temperature Equilibrium
 Examine the pipette tip for droplets
 Select forward or reverse pipetting based on the liquid
 Pause consistently
     Learn about quick tips 
Watch | Ergonomic advice for the bench
Did you know...
there are no toxic additives in our Nalgene plastics that can leach out into your lab solutions and ruin your experiments?
Top 10
We use laboratory-grade plastic resin materials to make our products. Customers trust Nalgene products to look and perform perfectly every time and rely on us to provide them with products that are good enough for their important lab work.
Here are a few Nalgene "must-haves" for your bench.
Request a printed poster for your lab
Receive a Nalgene Chemical Resistance Poster for your wall with everything you need to know at a glance.
 Improve Pipetting Accuracy with 10 Proven Steps
Aspirate and fully expel an amount of the liquid at least three times before aspirating for delivery. Failure to pre-wet the tip increases evaporation within the tip air space, which can cause significantly lower delivery volumes. Pre-wetting increases the humidity with the tip, thus reducing evaporation.
Allow liquids and equipment to equilibrate to ambient temperature prior to pipetting. The volume of liquid delivered by air displacement pipettes varies with the relative humidity and vapor pressure of the liquid – both of which are temperature-dependent. Working at a constant temperature minimizes variation of pipetted volume.
Temperature differences cause thermal expansion and shrinking in the air space. After temperature equilibrium, the influencing factor is liquid density. Cold liquid is more dense and hot liquid is less dense compared to room temperature liquids.
Before dispensing, carefully remove droplets from the outside of the tip by touching off the side of the reservoir, being sure to stay clear of the tip opening to avoid wicking liquid out of the tip. After dispensing, and before releasing the plunger, deliver any residual liquid remaining in the tip by touching the tip to the side of the container. Surface tension will help draw the remaining liquid out of the tip.
Depress the plunger to the first stop, immerse the tip into the liquid, and aspirate by releasing the plunger. Remove the pipette from the liquid and depress the plunger to the second stop to dispense the entire contents. Standard (or forward) mode pipetting yields better accuracy and precision than reverse mode for all but viscous or volatile liquids. Reverse mode often results in over-delivery. Hence, it’s recommended to evaluate the effect of possible over-delivery in the experiment and make adjustments if needed.
In this experiment 200µL of  viscous liquid (glycerol) was pipetted 10 times by using both forward and reverse pipetting techniques. The pipette used was adjusted for glycerol using forward pipetting. The chart describes the accuracy and precision obtained with both techniques.
Using the reverse method a smaller deviation between doses was observed and therefore reduced imprecision.
The reverse method gave bigger doses as the liquid column in the tip is taller and therefore the liquid amount above the dose presses a larger dose out.
After aspirating, and before removing the tip from the liquid, pause for one second. Make this pause as consistent as possible. Liquid continues to flow into the tip for a short time after the plunger stops. At the same time, evaporation within the tip is occurring. Pausing consistently balances these two effects and ensures correct aspiration.
When aspirating liquid, hold the pipette vertically and pull the pipette straight out from the center of the reservoir. This technique is especially important when pipetting small volumes (less than 50µL). Holding the pipette at an angle as it is removed from the liquid alters the aspirated volume.
Hold the pipette loosely and utilize the finger rest. Remember to return the  pipette to the pipette stand between deliveries. Avoid handling pipette tips or reservoirs with bare hands. Body heat transferred during handling disturbs temperature equilibrium, which leads to variations in delivered volume.
Before aspirating, immerse the tip adequately below the meniscus. Too little immersion, particularly with large volume pipettes, can lead to aspiration of air. Too much immersion can cause liquid to cling to the outside of the tip. Contacting the container bottom with the tip may restrict aspiration.
Step 1: The tip is immersed to the correct depth and correctly held vertically.
Step 2: Inaccuracy doubles when immersing the tip too deeply.
Step 3: Inaccuracy increases three to five times by immersing too deeply while holding the pipette at a 30-40° angle.
Use high-quality tips intended for use with the pipette. System tips are designed to work with their matching pipettes. Mismatched tips and pipettes can result in inaccuracy, imprecision, or both. Quality system tips provide an airtight seal, are made of superior materials, and are free of molding defects. They also ensure dependable liquid delivery.
Use the Correct Pipette Tip diagram
 Click image to enlarge
Depress the plunger smoothly until coming to rest with a light and consistent force at the first stop. Immerse the tip, and then release the plunger at a constant rate. Repeatable actions produce repeatable results.
Special Offers
Resources
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.