microplates-dishes-flasks-banner-470x200
Superior performance in virtually every application
Our quality control standards ensure you receive Thermo Scientific and Applied Biosystems plates, dishes, and flasks that are free of defects and exhibit very tight dimensional tolerances, to help enable superior performance.
Featured microplates, dishes, and flasks categories
A wide range of surfaces to suit a variety of needs. Dishes in more than 100 combinations of format and surface, with excellent optical quality for manual or automated imaging.
Prepare and store culture media and perform a broad range of culturing techniques with our flat-sided flasks and accessories, plastic spinner flasks, and plastic Erlenmeyer flasks.
Ideal for high-throughput screening. Work with small volumes and screen 4 times as many samples in one plate.
Accommodate sequencing and wash steps. Increased well volume allows for post-PCR applications to be carried out directly in the same plate.
Polypropylene's low binding capacity prevent proteins and DNA from binding, allowing for complete sample recovery. Plates withstand extreme temperatures and are available in a variety of colors.
Our range of accessories enables you to maximize your microplate performance utilizing superior sealing, tracking, and organization.
Polypropylene storage plates are an economical and dependable choice for low binding assays and storage of genomic samples and cells. Plate sealers secure samples with a reliable seal and prevent contamination or evaporation.
Plates, tubes, strips, sealing options, and storage plates for PCR and related applications.
Applied Biosystems MicroAmp plastics consumables offer excellent PCR or qPCR performance in formats developed to meet your experimental needs. 
Our cell culture inserts and carrier plates meet ANSI standards and are equipped with alphanumeric well identification. All items are USP Class VI, sterile, and include lids.
Tools and supplies for ELISA, including blank polystyrene microplates, pipetting reagent reservoirs, plate sealing tape, racked tube systems, and a unique vacuum-manifold apparatus for filter-based ELISA procedures.
Thermo Scientific Multi-well plates and Microplates
Select the ideal cell culture plate for your application
Simply select your criteria from the available options; the cell culture plate guide will return all plates that match your selections.
Explore cell culture plates
Support
For Research Use Only. Not for use in diagnostic procedures.
California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.