Search Thermo Fisher Scientific
Applied Biosystems TaqMan MicroRNA Assays quantitate miRNAs with the specificity and sensitivity of TaqMan Assay chemistry. A simple, two-step protocol requires only reverse transcription with a miRNA-specific primer, followed by real-time PCR with TaqMan probes. TaqMan MicroRNA Assays are:
Our TaqMan MicroRNA Assays catalog is kept up to date with the Sanger database to provide comprehensive coverage with the most current annotation.
What chromosome position are you interested in?
Genome Version: Build GRCh38 for
Availability & sizes: Made to Order assays are available in four different scales and usually ship within two weeks. Inventoried assays are prepackaged in Small scale and are ready to ship. Inventoried assays may also be purchased in larger sizes by ordering as MTO.
Format | Recommended TaqMan assay/array |
Individual qPCR assays | TaqMan miRNA Assays 2,000 pre-designed assays are inventoried and ship out in 1-2 days. |
384-well microfluidic TaqMan array card | TaqMan miRNA array cards Search for pre-designed 384-well arrays › |
OpenArray plate | Predesigned TaqMan miRNA OpenArray panels TaqMan Human miRNA OpenArray panel › |
Custom 96- and 384-well plates | Contact: specialty_plates@thermofisher.com |
Product Categories
Related Applications
Related Products
Species | TaqMan® MicroRNA Assays are available for a range of species including human, mouse, rat, Drosophila, C. elegans, and Arabidopsis. We will continue to increase the number of miRNA assays for these species in alignment with the Sanger miRBase Registry. |
Assay Components | TaqMan® MicroRNA Assays are delivered in two tubes: one containing the RT primer, the other containing the specific pre-formulated TaqMan® Assay (TaqMan® probe and PCR primer set). |
Available Assays | Download: Complete list of TaqMan® MicroRNA Assays |
Shipping and Storage | Shipped at ambient temperature. |
Catalog # | Availability | # of RT/PCR Reactions |
---|---|---|
4427975 | Inventoried | S: 50/150 |
4440885 | Made to Order | XS: 25/75 |
4440886 | Made to Order | S: 50/150 |
4440887 | Made to Order | M: 750/750 |
4440888 | Made to Order | L: 2900/2900 |
Application Note: Endogenous Controls for Real-Time Quantitation of miRNA Using TaqMan® MicroRNA Assays
Application Note: High Correlation of miRNA Quantitation Data from Matched FFPE and Snap-Frozen Tissues using TaqMan® MicroRNA Assays
Customer Focus Story: TaqMan® MicroRNA Assays help researchers validate miRNA markers for forensic body fluid identification
Poster: microRNA Quantitation by Real-time PCR
Poster: TaqMan-based miRNA Profiles Classify Embryonic Stem and Differentiated Cells
Product Bulletin: Quantitate microRNAs with the specificity and sensitivity of real-time PCR
QRC: TaqMan® Small RNA Assays
Design Process for Small RNA Assays
TaqMan® MicroRNA Assays FAQs
Brochure: TaqMan microRNA Assays
Product Categories
Related Applications
Related Products
Species | TaqMan® MicroRNA Assays are available for a range of species including human, mouse, rat, Drosophila, C. elegans, and Arabidopsis. We will continue to increase the number of miRNA assays for these species in alignment with the Sanger miRBase Registry. |
Assay Components | TaqMan® MicroRNA Assays are delivered in two tubes: one containing the RT primer, the other containing the specific pre-formulated TaqMan® Assay (TaqMan® probe and PCR primer set). |
Available Assays | Download: Complete list of TaqMan® MicroRNA Assays |
Shipping and Storage | Shipped at ambient temperature. |
Catalog # | Availability | # of RT/PCR Reactions |
---|---|---|
4427975 | Inventoried | S: 50/150 |
4440885 | Made to Order | XS: 25/75 |
4440886 | Made to Order | S: 50/150 |
4440887 | Made to Order | M: 750/750 |
4440888 | Made to Order | L: 2900/2900 |
Application Note: Endogenous Controls for Real-Time Quantitation of miRNA Using TaqMan® MicroRNA Assays
Application Note: High Correlation of miRNA Quantitation Data from Matched FFPE and Snap-Frozen Tissues using TaqMan® MicroRNA Assays
Customer Focus Story: TaqMan® MicroRNA Assays help researchers validate miRNA markers for forensic body fluid identification
Poster: microRNA Quantitation by Real-time PCR
Poster: TaqMan-based miRNA Profiles Classify Embryonic Stem and Differentiated Cells
Product Bulletin: Quantitate microRNAs with the specificity and sensitivity of real-time PCR
QRC: TaqMan® Small RNA Assays
Design Process for Small RNA Assays
TaqMan® MicroRNA Assays FAQs
Brochure: TaqMan microRNA Assays