Search Thermo Fisher Scientific
Organism Type | Mouse | The single-cell cloned LAP3 line was derived from NIH 3T3 cells co-transfected with pHbALAP267 and pSV2-neo after G418 selection, that are capable of expressing exogenous genes upon IPTG induction |
Tissues | Embryo | |
Phenotype | Adherent | |
Primary | no | |
Application | Transient Transfection |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||