The right gas chromatography modules for your application
Thermo Scientific instant connect (iConnect) injectors and detectors redefine flexibility and usability in GC and GC-MS analysis. Save on cost and bench space by keeping back-up modules in place of entire GC systems and ensure continuous operation by dramatically reducing maintenance downtime—just replace modules with clean back-ups for off-line maintenance and be back to running samples again in minutes. Swap out and exchange modules to easily reconfigure your GC system and be always ready for incoming samples.
Injectors
iConnect injectors offer greater versatility for your gas chromatography analyses.
iConnect Split/Splitless (SSL) Injector
Suitable for most GC and GC-MS applications. Equipped with an optimized thermal profile to permit accurate injection of a broad range of analytes.
iConnect Split/Splitless (SSL) Injector
iConnect Split/Splitless with Backflush (SSL-BKF) Injector
Ideal for eliminating heavy or undesired compounds. Backflush capabilities negate the need for additional auxiliary gas.
iConnect Split/Splitless with Backflush (SSL-BKF) Injector
iConnect Helium Saver Split/Splitless (SSL) Injector
Proprietary injector design that greatly reduces helium consumption during operation while using it as carrier gas. Does not require instrument method changes.
iConnect Helium Saver Split/Splitless (SSL) Injector
iConnect Programmable Temperature Vaporizing (PTV) Injector
Combines “discrimination-free” cold injector performance with robustness of vaporizing injectors. Ideal for thermally labile compounds and extended volatility ranges.
iConnect Programmable Temperature Vaporizing (PTV) Injector
iConnect Programmable Temperature Vaporizing with Backflush (PTV-BKF) Injector
Ideal for eliminating heavy or undesired compounds. Offers backflush capabilities and sub-ambient operation with a cryo cooling option.
iConnect Programmable Temperature Vaporizing with Backflush (PTV-BKF) Injector
iConnect Cold-On-Column (COC) Injector
True cold-on-column injector for extending GC analysis to very high boiling compounds. Ideal for thermal labile and degradable compounds.
  iConnect Cold-On-Column (COC) Injector
iConnect Gas Sampling Valve (GSV) Injector
Rapidly introduces precise amounts of gas samples directly into capillary or packed columns, making it ideal for gas analysis.
iConnect Gas Sampling Valve (GSV) Injector
Online purchase is not available in all regions.
Detectors
iConnect detectors are designed for enhanced productivity in gas chromatography workflows.
iConnect Flame Ionization Detector (FID)
Achieve sensitive detection of organic compounds over a wide linear dynamic range. Ideal for hydrocarbons and related organic compound analyses.
iConnect Gas Sampling Valve (GSV) Injector
iConnect Thermal Conductivity Detector (TCD)
Provides thermal stability and fast response times for capillary and packed column applications. Ideal for gas analysis at ppm levels and determination of unknowns.
iConnect Thermal Conductivity Detector (TCD)
iConnect Electron Capture Detector (ECD)
Achieve outstanding selectivity and sensitivity for trace-level detection and analysis of compounds with electronegative functional groups.
iConnect Electron Capture Detector (ECD)
iConnect Nitrogen Phosphorus Detector (NPD)
Multiple dedicated sources permit selectivity and sensitivity for the determination of organic compounds containing nitrogen or phosphorus.
iConnect Nitrogen Phosphorus Detector (NPD)
iConnect Flame Photometric Detector (FPD)
Exceptional sensitivity and response stability for determination of sulfur, phosphorous, or tin-containing compounds.
iConnect Flame Photometric Detector (FPD)
iConnect Pulse Discharge Detector (PDD)
Requires high purity helium as discharge gas and is ideal for gas analysis at ppb/ppm levels.
iConnect Pulse Discharge Detector (PDD)
Online purchase is not available in all regions.