therapudic-drug-monitoring
Optimize patient dosages with confidence
Therapeutic drug monitoring (TDM) tests provide quantitative results showing the correlation between the drug concentration in blood and a beneficial therapeutic effect. Too much drug can cause toxicity, and too little, a lack of efficacy. Individual patient factors such as age, ethnicity, metabolism, and history can affect drug performance. TDMs aid physicians in determining the optimal drug dosage for patients by monitoring serum or plasma drug concentrations to ensure effectiveness and patient compliance.
Thermo Fisher Scientific provides a broad range of TDM assays including full menus of antibiotics, antiepileptics and antiarrythmics as well as assays to monitor immunosuppressant drug levels in organ transplant patients.
plasma-or-serum
Jaon, K.K, Therapeutic Drug Monitoring Technologies, Companies and Markets, Jain PharmaBiotech, Basel, Switzerland Tech Report, p.14. February 2012.
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Assays. Manufactured since 1986, they provide an excellent range of Thermo Scientific™ DRI™ TDM Assays as well as CEDIA TDM Assays and Calibrators.
Streamline testing with Thermo Scientific™ CEDIA™ Therapeutic Drug Monitoring (TDM) Calibrators, a single calibrator kit that can be used with multiple CEDIA TDM assays. These economical kits are valued for their accuracy, precision and lot-to-lot dependability. They are available for cardiac or antiarrhythmic, antiepileptic and antibiotic drug monitoring in human serum and plasma. Each kit includes both high and low calibrators.
Receive reliable performance on a wide range of analyzers with Thermo Scientific™ DRI™ Therapeutic Drug Monitoring (TDM) Assays. Thermo Scientific TDM assays have been manufactured since 1986 and provide an excellent range of Thermo Scientific™ CEDIA TDM Assays, Thermo Scientific™ CEDIA TDM Calibrators and DRI TDM Assays with a wide dynamic measuring range of analytes.
Get relief from time-consuming and costly QC standard lot changes for up to 5 years with ready-to-use, liquid-assayed Thermo Scientific™ MAS™ Liquimmune™ Immunoassay Controls. With a new 5-year shelf life, these multi-analyte controls simplify QC while monitoring immunoassay test procedures on most automated instruments. These controls are also available in a convenient Vista™ vial format. Extended claim may not apply for all countries.
Consolidate general chemistry and serum protein QC processing into a single product offering three distinct levels with Thermo Scientific™ MAS™ Omni·CORE Controls. Value assignment is provided for key integrated instrument systems.
Consolidate routine immunoassay, tumor marker, and specialty immunoassay QC processing into a single product with Thermo Scientific™ MAS™ Omni·IMMUNE Immunoassay Controls.
Consolidate your immunoassay and tumor marker controls into a single QC product with Thermo Scientific™ MAS™ Omni·IMMUNE™ PRO Immunoassay Controls. This QC product contains the same analyte panel as the Omni·IMMUNE control, but also has the following additional analytes: Anti-Thyroglobulin (Anti-Tg), Anti-Thyroid Peroxidase (Anti-TPO), and Sex Hormone Binding Globulin (SHBG).
Check therapeutic drug monitoring test procedures with ready-to-use, liquid assayed multi-analyte Thermo Scientific™ MAS™ PAR™ TDM Controls, therapeutic drug controls with assayed values designed for therapeutic drugs and common immunoassayed test procedures on most automated instruments.
Accurately measure therapeutic drug levels in serum and plasma with the Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Assays. These liquid, ready-to-use assays are a convenient alternative to more expensive and time-consuming technologies. QMS (Quantitative Microsphere System) immunoassays have excellent precision and accuracy, optimized performance on a wide range of general chemistry analyzers and lot-to-lot consistency.
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.
Accurately measure therapeutic drug levels in serum and plasma with Thermo Scientific™ QMS™ Therapeutic Drug Monitoring (TDM) Calibrators, which employ advanced microparticle technology that is easy-to-use.
PrecisionInAnti-rejectionMonitoring
Precision in anti-rejection monitoring
Methodist Dallas Medical Center performs hundreds of liver and kidney transplants per year. With such volume, the medical center must be hyper-vigilant about lab result quality and efficiency.
Read more ›
Thermo Fisher Scientific products are distributed worldwide. Not all intended uses and applications mentioned here are available in every country.