Robust ICP-OES Systems
Our robust ICP-OES systems offer easy-to-use software and multi-element detection technology far superior to that of single-element AAS and multi-element microwave plasma techniques. They are ideal for laboratories with low sample throughput requirements and users new to the technique or who require a simple solution for multi-elemental analysis.
Learn more
Advanced, High-Throughput ICP-OES Systems
Analyze high-matrix trace elemental samples with sensitive multi-element detection and meet your data requirements. These instruments include both the iFR analysis mode, which measures the entire wavelength range in one measurement, and a eUV analysis mode, which measures the UV region of the spectrum with greater sensitivity.
Learn more
Semi-quantitative analysis for the iCAP PRO Series ICP-OES
Do you need to screen samples for their elemental composition as part of method development or to inform further analysis? The new semi-quantitative analysis tool within the Qtegra ISDS Software leverages artificial intelligence to enable you to do this in the simplest way. Find out how by reading this product spotlight.
Read now
iCAP PRO Series ICP-OES System product tour
Are you curious to see what our iCAP PRO Series ICP-OES Systems look like close up? We've designed a 3D product tour to help you explore these systems in a self-guided manner. Find videos, literature, and detailed descriptions of key features and capabilities.
View product tour › 
 iCAP PRO ICP-OESiCAP PRO X ICP-OESiCAP PRO XP ICP-OESiCAP PRO XPS ICP-OES
Plasma viewVertical Radial or Vertical Duo
Start up time1 hour (from off)15 minutes (from standby)5 minutes (from standby)
Wavelength rangeiFR mode 167.021 nm - 852.145 nmiFR mode 167.021 nm - 852.145 nm
eUV mode 167.021 nm - 240.063 nm
Resolution @ 200nm7 pm
Measurement modesiFRiFR and eUV
Camera technologyCID821
Full spectrum captureYes
RF source radial1150 W750, 1150 or 1350 W750 to 1600 W
RF source duo1150 W750, 1150 or 1350 W750 to 1600 W
Gas flow controlsOptimized MFCVariable MFC
Plasma gasFixed at 12.5 L/minFixed, optimized at
8.5, 12.5 or 14.5 L/min
0 to 20 L/min
Auxiliary gasFixed at 0.5 L/minSelectable
0.5, 1 or 1.5 L/min
0 to 2 L/min
Nebulizer gas0.3 to 0.8 L/min0 to 1.5 L/min0.0 to 1.5 L/min
Additional gas0 - 0.25 L/min
Video Player is loading.
Current Time 0:00
Duration 2:17
Loaded: 5.58%
Stream Type LIVE
Remaining Time 2:17
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Learn how the iSC-65 Autosampler offers exceptional performance in streamlining workflows and sample throughput.
    The Thermo Scientific iSC-65 Autosampler is a workhorse partner of the iCAP PRO Series ICP-OES. It has been designed with reliability from the ground up to ensure all samples are analyzed the first time.
    • Efficiency and productivity: The iSC-65 Autosampler allows sample-specific probe depths, avoiding unnecessary reruns due to blocked sample introduction systems. With the advanced Step Ahead feature, the autosampler probe is sent to rinse while data is still being acquired, using sample in the probe line to complete the analysis. 
    • Durable and robust design: Acid-resistant construction ensures confident sampling as well as longevity and robust operation in laboratory environments.
    • Visual feedback: Through LED lighting, provides clear, easy-to-see visual feedback at a distance on autosampler and analysis run status for technicians in crowded laboratories.
    • Unattended analyses: With the advanced quality control capabilities in Qtegra ISDS Software, the iSC-65 Autosampler instills confidence in high throughout, unattended ICP-OES and ICP-MS analyses.
    • Low contamination sample introduction: Utilize the iSC-65 Autosampler with the Thermo Scientific iCAP Qnova Series ICP-MS to eliminate manual sample introduction and the potential of contamination when analyzing ultra-pure high sensitivity samples.
    Ordering information
    Partnering in sustainability - ACT label
    Reduce your lab’s environmental impact with the iCAP PRO Series ICP-OES, recipient of the ACT label from My Green Lab. The ACT label, the standard for determining the environmental impact of laboratory products, helps labs make informed, sustainable purchasing decisions.
    Learn more about the ACT label ›
    * Required field
    *
    *
    *
    *
    *
    *
    *
    *
    *
     Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
    This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
    Style Sheet for Global Design System