Lipofectamine LTX Reagent kit box and two tubes, one with a red cap and one with a green cap
Lipofectamine LTX Reagent is a plasmid transfection reagent that offers a balance of potency and gentleness for your cells, resulting in high transfection efficiencies and viabilities. This reagent provides an advanced solution for gene expression in common cell lines, particularly Chinese Hamster Ovary (CHO) cells, and challenging cell types such as primary neural progenitor cells, primary fibroblasts, and primary epithelial cells.
Lipofectamine LTX reagent offers:
  • Superior delivery—transfect common cell lines and challenging primary and sensitive cells, with high efficiency
  • Superior performance—combine high transfection efficiency with maximal (>90%) cell viability
  • Optimized protocol—rapid, simple protocol provided for most cell lines
Request a sample  Protocol library
Ordering information for Lipofectamine LTX with Plus Reagent
Catalog # Name Size Price (USD) Qty
A12621 Lipofectamine™ LTX Reagent with PLUS™ Reagent, 0.1 mL Each
110.65
Online Exclusive
116.00
Save 5.35 (5%)
15338030 Lipofectamine™ LTX Reagent with PLUS™ Reagent, 0.3 mL Each
302.65
Online Exclusive
315.00
Save 12.35 (4%)
15338100 Lipofectamine™ LTX Reagent with PLUS™ Reagent, 1 mL Each
854.65
Online Exclusive
940.00
Save 85.35 (9%)
15338500 Lipofectamine™ LTX Reagent Each
12,670.00
Lipofectamine LTX has broad compatibility with cell lines
In addition to being excellent for transfecting primary and difficult-to-transfect cells, Lipofectamine LTX reagent provides a versatile approach to transfecting established cell lines, including those that are sensitive to other transfection reagents. Data for Lipofectamine vs FuGENE® has shown that the Lipofectamine LTX reagent leads to exceptional transfection efficiency with lower toxicity (Figure 1).
Bar graph showing transfection efficiency of Lipofectamine LTX vs Fugene reagents across ten cell lines demonstrating overall better performance of Lipofectamine LTX.
Figure 1. Lipofectamine LTX Reagent combines high transfection efficiency with low cytotoxicity superior to FuGENE 6 and FuGENE HD transfection reagents. Activity/toxicity analysis measures the ratio of peak gene expression to cytotoxicity, with higher ratios indicating optimization of transfection efficiency with low cytotoxicity levels. Ten cell lines were transfected with luciferase-expressing plasmids following recommended protocols for the respective reagents. Cells were analyzed for toxicity using the Invitrogen CyQUANT Cytotoxicity Assay Kit (Cat. No. V23111) and assayed for luciferase expression. The data were normalized to the activity/toxicity ratios for the FuGENE HD reagent.
Transfect primary cells efficiently with Lipofectamine LTX reagent and PLUS reagent
Primary cells have traditionally been difficult to transfect. Lipofectamine LTX reagent, when combined with PLUS Reagent, solves the problem of transfecting primary cells, including primary neural progenitor cells (Figure 2).
Three micrographs demonstrating how Lipofectamine LTX with PLUS Reagent efficiently transfects primary neural cells; the 1,000 ng image shows the most GFP activity
Figure 2. Lipofectamine LTX reagent with PLUS Reagent efficiently transfects primary neural progenitor cells. Lipofectamine LTX reagent (1.5 μL) was used to transfect murine embryonic primary neural progenitor cells with the indicated quantities of a GFP-expressing plasmid in the presence of PLUS Reagent (2.5 μL). GFP expression was analyzed 24 hours post-transfection. Data courtesy of the Beverly Davidson laboratory, University of Iowa.
Lipofectamine LTX reagent uses a simple, rapid, and serum-compatible protocol
Lipofectamine LTX reagent has a protocol that is easy to use with improved performance. When using this protocol, there is no need to remove transfection complexes or change medium following transfection. You can spend less hands-on time, and get better results, faster.
Line graph comparing the performance of the Lipofectamine LTX with PLUS Reagent’s old and new protocols for six cell lines
Figure 3. Comparison of performance between the current protocol with an old protocol for use of Lipofectamine LTX PLUS Reagent.
Plasmid transfection protocol
How to perform plasmid DNA transfection with Lipofectamine LTX with PLUS Reagent
Resources
Our Lipofectamine reagent-specific protocols have been optimized for efficiency, viability, and reproducibility across a broad range of cell types. Search this library to find protocols and citations curated to fit your experimental needs. Filter by cell type, payload, and product of choice to easily unlock your answers.
Explore protocols and citations
Transfection FAQs
Search our extensive collection of Transfection FAQs to find the answers related to Lipofectamine LTX with PLUS reagent.
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments