Ribonuclease- (RNase-) free buffers and reagents are rigorously tested for nuclease contamination (RNase and DNase) as part of the routine quality control process. These reagents have all been shown to lack detectable non-specific endonucleases, exonucleases, and RNase activity. To protect your experimental results, common lab buffers, such as Tris-EDTS (TE) buffer and phosphate buffered saline (PBS), are available in nuclease-free, RNase-free formats.
Maintaining an RNase-free lab Nuclease-free water
RNase-free TE buffer
Invitrogen RNase-free TE buffer, pH 8.0 product photo
Tris-EDTA (TE) is a commonly used pH buffer for labs storing RNA and DNA samples. TE buffer consists of 10 mM Tris (adjusted to the specified pH with HCl) and 1 mM EDTA. These TE buffers are filtered through a 0.2 μm filter into a sterile autoclavable bottle, are nuclease-free, and ready for use without additional preparation.
Features of RNase-free TE buffer include:
  • Available in pH 7.0 or 8.0
  • Certified RNase-free
  • Regular quality control lot testing for contaminating non-specific endonucleases, exonucleases, and RNases
  • Manufactured in facilities that are designed to prevent nuclease contamination
Catalog # Name Size Price (USD) Qty
AM9849 TE, pH 8.0, RNase-free, 500 mL Each
65.65
Online Exclusive
69.25
Save 3.60 (5%)
AM9858 TE, pH 8.0, RNase-free, 1 L Each
93.65
Online Exclusive
101.00
Save 7.35 (7%)
AM9860 TE, pH 7.0, RNase-free, 10 x 1 mL Each
66.65
Online Exclusive
70.00
Save 3.35 (5%)
AM9861 TE, pH 7.0, RNase-free, 50 mL Each
64.65
Online Exclusive
68.75
Save 4.10 (6%)
RNase-free PBS for cell culture applications
Invitrogen RNase-free PBS, pH 7.4 product photo
Phosphate-buffered saline (PBS) is a phosphate buffer saline solution used in a cell culture applications. RNase-free PBS helps keep contaminants out of samples and research. PBS that is considered molecular biology grade should not contain contaminants such as proteases, RNases, and DNases.
Features of RNase-free PBS include:
  • 10X dilution— ready to use upon dilution to the desired concentration
  • Certified RNase-free
  • Regular quality control lot testing for contaminating non-specific endonucleases, exonucleases, and RNases
Catalog # Name Size Price (USD) Qty
AM9624 PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free, 500 mL Each
66.65
Online Exclusive
70.75
Save 4.10 (6%)
AM9625 PBS - Phosphate-Buffered Saline (10X) pH 7.4, RNase-free, 1 L Each
97.65
Online Exclusive
105.00
Save 7.35 (7%)
Additional RNase-free reaction buffers and chemicals
Thermo Fisher offers a selection of RNase-free buffers and reagents that are commonly used in reactions containing RNA and DNA. Use these products in your experiments with confidence, knowing that they have passed rigorous quality checks for contaminating non-specific endonucleases, exonucleases, and RNase activity.
Glycogen and GlycoBlue coprecipitant for nucleic acid recovery
Invitrogen Glycogen (5 mg/ml) product photo
Glycogen is a carbohydrate molecule widely used in labs to co-precipitate with RNA or DNA for nucleic acid recovery. Similarly, GlycoBlue coprecipitant has a blue dye molecule linked to glycogen for visibility of the nucleic acid pellet.
Both Glycogen and GlycoBlue coprecipitant have the following features:
  • Ideal with RT-PCR
  • Increases pellet mass and visibility
  • Quantitative recovery of low concentrations (ng/mL) of nucleic acid
  • Prevents pellet loss in nuclease protection assays
  • RNase- and DNase-free
Catalog # Name Size Price (USD) Qty
AM9510 Glycogen (5 mg/ml) Each
252.65
Online Exclusive
266.00
Save 13.35 (5%)
AM9515 GlycoBlue™ Coprecipitant (15 mg/mL), 300 μL Each
129.65
Online Exclusive
137.00
Save 7.35 (5%)
AM9516 GlycoBlue™ Coprecipitant (15 mg/mL), 5 x 300 μL Each
323.65
Online Exclusive
344.00
Save 20.35 (6%)
Proteinase K to inactivate protein activity
Ambion Recombinant Proteinase K Solution (20 mg/ml) product photo
Proteinase K is commonly used in molecular biology labs to treat for contaminating nucleases (RNase, DNase). Proteinase K is a serine protease that digests a broad range of proteins.
Recombinant Proteinase K solutions can be used for:
  • Preparation of RNA and high molecular weight DNA for gel electrophoresis
  • Terminate enzymatic reactions, such as phosphatase or nuclease treatments
  • Treatment of samples with high protein concentrations to facilitate downstream phenol extraction
Catalog # Name Size Price (USD) Qty
AM2546 Recombinant Proteinase K Solution (20 mg/mL), 1.25 mL Each
102.65
Online Exclusive
107.00
Save 4.35 (4%)
AM2548 Recombinant Proteinase K Solution (20 mg/mL), 5 x 1.25 mL Each
311.65
Online Exclusive
334.00
Save 22.35 (7%)
RNase-free EDTA for RNA sample storage
Invitrogen RNase-free EDTA (0.1 mM), pH 8.0 product photo
Ethylenediaminetetraacetic acid (EDTA) is a chemical that is universally used in labs for RNA sample storage. Features of RNase-free EDTA include:
  • Prepared in nuclease-free (RNase-free, DNase-free), ultrapure water
Catalog # Name Size Price (USD) Qty
AM9912 EDTA (0.1 mM), pH 8.0, RNase-free Each
73.75
RNase-free salmon sperm DNA for hybridization experiments
Invitrogen Salmon Sperm DNA, sheared (10 mg/mL) product photo
Sheared salmon sperm DNA is a common blocking reagent used in molecular biology labs. It is useful in blocking during northern prehybridization and hybridization. Features of RNase-free salmon sperm DNA include:
  • Treated with Proteinase K
  • RNase- and DNase-free
Catalog # Name Size Price (USD) Qty
AM9680 Salmon Sperm DNA, sheared (10 mg/mL) Each
383.65
Online Exclusive
411.00
Save 27.35 (7%)
RNase-free Tris for electrophoresis assays
Invitrogen RNase-free Tris, pH 8.0 product photo
Tris solution is used in molecular biology labs in electrophoresis assays. Tris is the main ingredient for TAE (Tris acetate EDTA) and TBE (Tris borate EDTA) buffers regularly used in DNA gel electrophoresis. RNase-free Tris has the following features:
  • Available in pH 8.0
  • RNase-free
  • Regular lot testing for contaminating non-specific endonucleases, exonucleases, and RNases
Catalog # Name Size Price (USD) Qty
AM9855G Tris (1 M), pH 8.0, RNase-free, 100 mL Each
57.65
Online Exclusive
61.00
Save 3.35 (5%)
AM9856 Tris (1 M), pH 8.0, RNase-free, 500 mL Each
115.65
Online Exclusive
124.00
Save 8.35 (7%)
THE RNA Storage Solution for RNA pellet resuspension
Invitrogen THE RNA Storage Solution product photo
THE RNA Storage Solution is an RNA stabilizing solution used to resuspend and store RNA for future downstream assays, such as:
  • RT-PCR
  • In vitro transcription
  • Northern analysis
  • Nuclease protection assays
View other RNA stabilizers
Catalog # Name Size Price (USD) Qty
AM7000 THE RNA Storage Solution, 10 x 1 mL Each
67.65
Online Exclusive
72.00
Save 4.35 (6%)
AM7001 THE RNA Storage Solution, 50 mL Each
67.65
Online Exclusive
72.50
Save 4.85 (7%)
TechNotes articles
Support
CMD Wide-format style fixes
For Research Use Only. Not for use in diagnostic procedures.
Stylesheet for Classic Wide Template adjustments