Cas9 lentivirus for screening and high-efficiency editing
Invitrogen LentiArray Cas9 Lentivirus provides an efficient method to drive stable Cas9 nuclease expression in a wide variety of cell types, including hard-to-transfect cells. The LentiArray Cas9 Lentivirus construct includes a human codon-optimized Cas9 with two nuclear localization signals to facilitate efficient delivery into the nucleus. This CRISPR-Cas9 lentivirus gives you the ability to create a stable pool of Cas9-expressing cells or to isolate stable Cas9-expressing clones.
LentiArray Cas9 Lentivirus can be used in screening applications with LentiPool and LentiArray CRISPR libraries or targeted gene experiments with LentiArray CRISPR gRNAs or TrueGuide Synthetic gRNAs.
Ready-to-transduce component of CRISPR viral delivery system
Map showing the structure of the Invitrogen LentiArray Cas9 Lentivirus construct
Invitrogen LentiArray Cas9 Lentivirus map
In this CRISPR viral delivery system, an EFS promoter on the lentiviral construct drives Cas9 protein expression, while a blasticidin resistance gene allows for straightforward selection of transduced cells. In addition to the nuclear localization signals, the encoded Cas9 nuclease also includes the V5 epitope tag for easy detection of Cas9 proteins.
Order Cas9 lentivirus products
LentiArray Cas9 Lentivirus is provided as a high titer, ready-to-use lentivirus at 1x107 TU/mL and is available in two sizes. For creating your Cas9-expressing pools and isolating Cas9-stable clones, select the 100 μL size. For co-infection in either single gene targeting or screening applications, select the larger 1 mL size.
In addition, order Invitrogen LentiArray CRISPR Positive and Negative Control Lentiviruses for optimizing transduction and antibiotic selection conditions in your experiments.
Catalog # Name Size Price (USD) Qty
A32064 LentiArray™ Cas9 Lentivirus Each
723.65
Online Exclusive
762.00
Save 38.35 (5%)
A32069 LentiArray™ Cas9 Lentivirus Each
2,359.65
Online Exclusive
2,610.00
Save 250.35 (10%)
Resources and support for CRISPR-Cas9 lentivirus
For Research Use Only. Not for use in diagnostic procedures.