Heraguard ECO Clean Bench
The Thermo Scientific Heraguard ECO clean bench delivers outstanding product protection, energy efficiency and user comfort. Dual DC motors and advanced airflow design provide advanced protection, quiet operation and low impact to the lab environment. Our clean benches meet ISO class 5 minimum conditions achieved in the work area and EU GMP Grade A work environment. They are available in four sizes: 900 mm, 1200 mm, 1500 mm, or 1800 mm (1800 mm size with optional extra height).
Efficiency, safety and functionality combined
Smart Ports
Easily pass tubing and wiring through sidewall to accommodate many application needs.
SmartPort convenience of clean benches
Side windows
Safety glass panels include pass-through tubing ports for convenience, while enabling better visibility and a more open workspace.
Heraguard ECO side window
Intuitive control panel
Performance and operational control status provided through easy to use and read, eye-level panel.
Easy touch-button Heraguard ECO control panel
Features
1500 B2 BSC detailed features
1 DC Motors
Deliver better energy efficiency and low heat emission into the laboratory.
2 Optional UV Light 
Fitted into the top of the workspace, the UV light disinfects the work area.  
3 Customizable worktops 
Choose from stainless steel or melamine work surfaces that best suit your application and work style.
4 Windscreen protection (optional)
Built to be easily flipped down when the clean bench is in use, the windscreen protects users against the dry purified air.
5 Easy prefilter exchange 
The prefilter on the intake side can be changed by hand, without the need for tools. Prefilter exchange is made economical and easy.
Horizontal laminar flow
Clean benches provide a clean (aseptic) space to work with a product or specimen without contaminating it with particles such as micro-organisms. They protect the product, they do not protect the user or the environment.
Horizontal laminar flow
Air is pulled in from the top and passes through a pre-filter, before it is then pushed through the HEPA filter. A horizontal laminar flow is produced within the work area. The laminar flow (unidirectional flow) helps protect the product from the worker and also contamination from the environment. The work area becomes a (positively pressurized) particle free aseptic clean zone which is being continually bathed in clean air. 
Airflow graphic for horizontal laminar flow
Applications
  • Hospitals (Pharmacy) — drug dispensing, IV mixture prep, media prep, drug compounding
  • Cleanrooms
  • Hospitals (microbiology dept.) — Tissue Culture (nonbiological)
  • IVF departments
  • Pharmaceutical research
  • Cleanrooms — GMP suites
  • Pharmaceutical microbiology
  • Sterile packaging and filling
  • Micro-electronics assembly and semi-conductor production
  • Plant cell culture/agriculture
  • Food and drink industry
  • Cosmetics industry
  • Aerospace industry
Popular applications for clean benches
Certifications
  • Agency: EAC, CSA
  • CE:  2014/35/EU, 2014/30/EU, 2011/65/EU incl. (EU) 2015/863 Amending Annex II to Directive 2011/65/EU
  • Standards:  EN 61010-1:2010 + A1: 2019, EN 61010-1, EN 61326-1
Resources
Ordering information
Size/Width
0.9m (3-ft)
1.2m (4-ft)
1.5m (5-ft)
1.8m (6-ft)
1.8m (6-ft)
950 mm interior height
230V
51029701
51029702
51029703
51029704
51029705
120V
51029696
51029697
51029698
51029699
51029700
Many options are available. Please check with your local Sales representative.