Precision Water Bath PDF
Precision redefined
Thermo Scientific Precision water baths are setting a new standard with enhanced features that simplify workflows and maximize productivity. Their rugged construction, compact footprint, advanced temperature controls, ease of use, and safety features clearly make our water baths a smart choice for your smart lab.
Browse our complete portfolio or let one of our experts help you make a selection.
Contact us
Thermo Scientific Precision water baths are:
  • Compact—Save valuable bench space with the small footprint of our general purpose baths.
  • Effortless—Automatically preheat and turn off your bath for efficient work scheduling with new auto-on and auto-off timers.
  • Intuitive—Simplify parameter setting and monitoring with new icon-based controller interface to help ensure safe operation.
  • Safe—Protect your work with features such as audible alarm,  low-level detection and high-temperature cut-offs.
  • Everywhere—Access worldwide service and support when you need it for added peace of mind.
Download brochure
Featured precision water baths
Rugged, high-performance baths designed to maintain water temperature from ambient to 100°C and are ideal for a wide range of lab applications.
An ideal choice when temperature uniformity and control are particularly critical, as when working with enzymes or in serological applications.
Equipped with next-generation shaker motor designed to support sensitive life science and QA/QC applications.
Designed specifically for fecal coliform determination, with advanced temperature controller factory preset to 35.0, 41.5 and 44.4°C. 
Applications
Thermo Scientific Precision Water Baths
 General PurposeCirculatingColiformShakingDubnoff
Bacteriological  Examinations
Coagulation Tests  
Coliform  Determinations  
Copper Strip Corrosion Tests  
Crude Oil Studies   
Cytochemistry  
Dialysis    
Demulsibility Studies   
Enzyme Studies   
Electrophoresis Gel Destaining    
Environmental Studies
Food Processing QC 
Genetic Studies 
Hormone Studies 
Immunological Research 
Incubation for Microbiological Assays 
Incubation for Microcentrifugation Tubes   
Melting Agar    
Metallurgical Analysis 
Molecular Biology 
Protein Analysis   
Radioactive Isotope Uptake Studies 
Radiochemistry  
Serological Research 
Thawing 
Thawing Cryopreservation Vials    
Tissue Culture Research 
Virology Research 
Warming Reagents   
Water Quality Research