Erlenmeyer flasks
Nalgene Erlenmeyer Flasks are a safer alternative to glass. They won’t break, chip or shatter amid the bustle and bumps of everyday lab work. Enjoy their light weight and ease of handling. Trust Nalgene Erlenmeyer Flasks to be made from the highest quality laboratory-grade polypropylene copolymer (PPCO), polycarbonate (PC), polymethylpentene (PMP) and fluoropolymer (FEP) materials for dependably low extractables across a wide variety of lab applications.
Choose polypropylene for its low cost and good chemical compatibility with a wide range of lab chemicals. Get very good chemical resistance and clarity, too, with PMP. FEP provides the ultimate in chemical resistance, temperature range and inertness for the most demanding lab applications. Or, choose polycarbonate for its window-like clarity making it an ideal culture flask. Screw cap flasks are leakproof.*
Which flasks are best for your application?
Nalgene PPCO
Erlenmeyer Flask
Nalgene PC
Erlenmeyer Flask
Nalgene PC
Baffled
Erlenmeyer Flask
Nalgene PC
Erlenmeyer Flask
with Screw Cap
Nalgene PMP
Erlenmeyer Flask
with Screw Cap
Nalgene FEP
Erlenmeyer Flask
with Screw Cap
Traditional style,
milky white,
chemical resistant
Traditional style, window clearTraditional style,
window clear
with baffles
Threaded screw
cap design,
window clear
Threaded screw
cap design,
good clarity,
chemical resistant
Threaded screw
cap design,
superior chemical resistance
Mixing reagentsMedia prep, 
culture techniques
Suspension,
culture techniques
Media storage,
culture techniques
Reagent prep,
storage, titrations
Aggressive chemical
prep, trace
metals work
No capNo capNo capScrew on cap, leakproof*Screw on cap, leakproof*Screw on cap, leakproof*
No graduationsGraduations molded, mLGraduations
molded, mL
Graduations molded, mLGraduations molded, mLGraduations
molded, mL
125mL-2000mL125mL-500mL250mL-2000mL125mL-500mL125mL-1000mL125mL-250mL
PPCO flaskPC flaskPC flaskPC flask
w/PP closure
PMP flask
w/PP closure
FEP flask
w/ETFE closure
Milky white translucentWindow clearWindow clearWindow clearGood clarityHazy white
translucent
Temp. range
-40oC to 121oC
Temp. range
-135oC to 135oC
Temp. range
-135oC to 135oC
Temp. range
-135oC to 135oC
Temp. range
20oC to 153oC
Temp. range
-270oC to 205oC
AutoclavableAutoclavable (shortens life)Autoclavable (shortens life)Autoclavable (shortens life)AutoclavableAutoclavable
Complies with
Part 21
CFR 177.1520
for food use
Complies with
Part 21
CFR 177.1580
for food use
Complies with
Part 21
CFR 177.1580
for food use
Complies with
Part 21
CFR 177.1580
for food use
No food complianceComplies with
Part 21
CFR 177.1550
for food use
BPA-FreeContains BPAContains BPAContains BPABPA-FreeBPA-Free
Phthalate-freePhthalate-freePhthalate-freePhthalate-freePhthalate-freePhthalate-free
General chemical compatibility
 Nalgene PPCO 
Erlenmeyer Flask
Nalgene PC 
Erlenmeyer Flask
Nalgene PC 
Baffled 
Erlenmeyer Flask
Nalgene PC
Erlenmeyer Flask
with Screw Cap
Nalgene PMP 
Erlenmeyer Flask 
with Screw Cap
Nalgene FEP 
Erlenmeyer Flask 
with Screw Cap
Acids, dilute or weakEEEEEE
Acids, strong, concentratedGN N N EE
Alcohols, aliphaticEGGGEE
AldehydesGFFFGE
Bases/alkaliENNN EE
EstersGNN N EE
Hydrocarbons, aliphaticGGGGGE
Hydrocarbons, aromatic NNN N N E
Hydrocarbons, halogenated NNNN N E
Ketones, aromaticNN N N FE
Oxidizing agents, strongFFFFGE
For a more detailed chemical resistance guide by chemical and application temperature, please refer to Chemical resistance guide or request a printed poster for your lab or contact Nalgene Technical Support.
 E No damage after 30 days constant exposure at 20 oC.
 G Little or no damage after 30 days constant exposure at 20oC.
 F Some effect after 7 days constant exposure at 20oC.
 N Not recommended. Immediate damage may occur.
*The term “leakproof” applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes; b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
^ Except oxidizing acids. For oxidizing acids, see “Oxidizing agents, strong.”
Start every day with dependable labware | Thermo Scientific Nalgene labware
Your work depends on quality and consistency, and we believe every lab should have the best tools. Watch Video
Video Player is loading.
Current Time 0:00
Duration 1:46
Loaded: 0.00%
Stream Type LIVE
Remaining Time 1:46
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Nalgene side-arm vacuum flasks
    Nalgene side-arm vacuum flasks are rugged and ready for daily lab use. Replace all your glass vacuum flasks with Nalgene vacuum flasks which can take a beating without a chip, crack or shatter. Molded in one piece for full vacuum service. 
    Support
    Need technical support? Contact our expert team for technical and application support of Laboratory Products at technicalsupport@thermofisher.com.
    California Proposition 65 Warning: Products manufactured with polycarbonate (PC), polyethylene terephthalate (PET), polyethylene terephthalate glycol (PETG) or polystyrene (PS) contain chemicals known to the State of California to cause cancer, birth defects or other reproductive harm.