Search Thermo Fisher Scientific
Applied Biosystems™ TaqMan™ MicroRNA Assays are innovative tools for miRNA research—from isolation through discovery, profiling, quantitation, validation and functional analysis. There are now even more choices for detection of mature, biologically active miRNA, designed to support a growing range of miRNA applications. The new Applied Biosystems™ TaqMan Advanced miRNA Assays use ligation-based universal reverse transcription for a streamlined and highly sensitive workflow. The standard TaqMan MicroRNA Assays, which employ target-specific stem-loop reverse transcription primers for 3’ extended templates, continue to cover a range of species using standard TaqMan Assay–based real-time PCR.
Enable highly sensitive and specific quantification of mature miRNAs using qPCR. Find your affordable solution using the search tool below.
Search for pre-designed array plates ›
Search for pre-designed array cards ›
What chromosome position are you interested in?
Genome Version: Build GRCh38 for
New TaqMan Advanced miRNA Assays use universal reverse transcription and provide highly sensitive quantification.
Standard TaqMan MicroRNA Assays use a target-specific stem-loop reverse transcription primer.
TaqMan Pri-miRNA Assays detect and measure primary miRNAs from human, mouse, or rat species.
The content for our standard TaqMan MicroRNA Assays and the new TaqMan Advanced miRNA Assays is aligned with release 22 of the miRBase database. For a complete list of standard TaqMan MicroRNA Assays, TaqMan Advanced miRNA Assays, and other technical resources, please visit our TaqMan MicroRNA Assays resource page.
TaqMan Non-coding RNA Assays for reliable detection and quantitation of non-coding human, mouse, or rat transcripts longer than 200 nucleotides.
Custom assays to quantitate small RNA are ideal to quantitate your small RNA when a predesigned assay is unavailable—all you need is the sequence of interest.
Predesigned TaqMan Advanced miRNA 96-well Plates are now available in both fast and standard format. In addition, custom 96-well plates containing TaqMan Advanced miRNA assays or the classic chemistry TaqMan miRNA assays are available with the TaqMan Custom Plating Service.
Predesigned TaqMan Advanced miRNA array cards are now available, as well as custom TaqMan array cards containing TaqMan Advanced miRNA assays or the classic chemistry TaqMan miRNA assays that are ordered using our configurator tool. TaqMan array cards are the ideal solution for profiling large numbers of miRNAs and samples with limited material (each reaction requires only 2 µL of sample). TaqMan array cards are compatible with the 7900HT, ViiA 7, QuantStudio 7, and QuantStudio 12K Flex real-time PCR systems.
Order your TaqMan MicroRNA Assays pre-plated into 384-well plates with the TaqMan Custom Plating Service. Such plates are perfect for analyzing a large number of samples for a few target miRNAs or non-coding RNAs or for evaluating a few samples for a large number of targets.
These plates accommodate 3,072 reactions at once, delivering the high-throughput capabilities required for large profiling studies. Designed for use with the Applied Biosystems OpenArray Real-Time PCR System and the QuantStudio 12K Flex Real-Time PCR System.
TaqMan MicroRNA Assays FAQs
Product Bulletin: Quantitate microRNAs with the specificity and sensitivity of real-time PCR
Download complete list of TaqMan MicroRNA Assays
Protocol for generating Custom Pools of RT primers
Brochure: TaqMan microRNA Assays
For Research Use Only. Not for use in diagnostic procedures.