Front angle on the VeritiPro Thermal Cycler, including the user interface home screen
The Applied Biosystems Veriti Thermal Cycler set a new standard for innovation when this thermocycler launched in 2007. It was the first thermal cycler to employ an LCD touchscreen interface, multiple temperature zone block, and web-enabled remote-control software. Building on these innovations, the VeritiPro Thermal Cycler upgrade features the latest block technology and connectivity options. These advancements provide higher ramp rates, quieter runs, and more intuitive VeriFlex temperature optimization setup while continuing to offer the same high-end performance.
Request quote Request demo Virtual tour
Watch video
96- and 384-well options for gradient thermocycler VeritiPro Thermal Cycler
The VeritiPro Thermal Cycler comes with two options:
  1. 96-well fixed block with 6 VeriFlex zones for user-independent temperature control
  2. 384-well fixed and isothermal block
Linear gradient PCR optimization
Gradient temperature control is a feature of thermal cyclers to help primer annealing in PCR. However, thermal cyclers equipped with “better-than-gradient” technology are alternatives that help improve temperature control for primer annealing. These metal blocks often come with three or more segmented blocks, each with separate heating and cooling elements. VeritiPro Thermal Cycler is equipped with “better-than-gradient” technology, namely Veriflex Blocks.
Video Player is loading.
Current Time 0:00
Duration 0:58
Loaded: 0%
Stream Type LIVE
Remaining Time 0:58
1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
    • en (Main), selected
    Video: Introducing the linear gradient thermocycler, VeritiPro Thermal Cycler
    We've made Veriti Thermal Cycler even better—Meet VeritiPro Thermal Cycler. Watch this video to view features and capabilities of our gradient thermocycler, VeritiPro Thermal Cycler.
    Stay in control with Thermal Cycler Fleet Control Software
    Diagram of multiple thermal cyclers connected to one tablet to represent Fleet Control Software
    Manage multiple users, methods, and instruments with a single intuitive interface securely behind your firewall.
    Learn more about Thermal Cycler Fleet Control Software
    High-performance PCR and qPCR plastics for optimal results
    Five multi-colored PCR tube plates
    Applied Biosystems PCR plastics have been designed and validated to work with our thermal cyclers for more than 25 years. Options for every format and throughput need are available to ensure consistent, precise results for end-point PCR applications. Quickly find the right fit and compatible plastics for your desired model and block formats by using our plastics selection guide.
    Explore online PCR and qPCR plastics selection tool
    Ordering information
    Catalog # Name Size Price (USD) Qty
    A47396 VeritiPro™ Thermal Cycler + Plastics Package, 96 well, 8-tube strips, 1 package Each
    7,146.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    11,910.00
    Save 4,764.00 (40%)
    A47397 VeritiPro™ Thermal Cycler + Plastics Package, 96 well, EnduraPlates, 1 package Each
    7,146.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    11,910.00
    Save 4,764.00 (40%)
    A47398 VeritiPro™ Thermal Cycler + Plastics Package, 96 well, TriFlex plates, 1 package Each
    7,146.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    11,910.00
    Save 4,764.00 (40%)
    A47399 VeritiPro™ Thermal Cycler, 384-well, 1 Package Each
    7,062.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    11,770.00
    Save 4,708.00 (40%)
    Catalog # Name Size Price (USD) Qty
    A48766 VeritiPro™ Thermal Cycler + Extended Warranty + Plastics Package, 96 well, ABRC, 8-tube strips, 1 package Each
    7,370.40
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,284.00
    Save 4,913.60 (40%)
    A48767 VeritiPro™ Thermal Cycler + Extended Warranty + Plastics Package, 96 well, REX, 8-tube strips, 1 package Each
    7,407.60
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,346.00
    Save 4,938.40 (40%)
    A48768 VeritiPro™ Thermal Cycler + Extended Warranty + Plastics Package, 96 well, ABRC, EnduraPlates, 1 package Each
    7,370.40
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,284.00
    Save 4,913.60 (40%)
    A48769 VeritiPro™ Thermal Cycler + Extended Warranty + Plastics Package, 96 well, REX, EnduraPlates, 1 package Each
    7,407.60
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,346.00
    Save 4,938.40 (40%)
    A48770 VeritiPro™ Thermal Cycler + Extended Warranty + Plastics Package, 96 well, ABRC, TriFlex Plates, 1 package Each
    7,370.40
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,284.00
    Save 4,913.60 (40%)
    A48771 VeritiPro™ Thermal Cycler + Extended Warranty + Plastics Package, 96 well, REX, TriFlex Plates, 1 package Each
    7,407.60
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,346.00
    Save 4,938.40 (40%)
    A50327 VeritiPro™ Thermal Cycler, 384-well, 1 Package Each
    7,105.20
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    11,842.00
    Save 4,736.80 (40%)
    A50328 VeritiPro™ Thermal Cycler, 384-well, 1 Package Each
    7,326.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    12,210.00
    Save 4,884.00 (40%)
    Catalog # Name Size Price (USD) Qty
    A48141 VeritiPro™ Thermal Cycler, 96 well, 1 instrument Each
    7,709.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    11,860.00
    Save 4,151.00 (35%)
    A48140 VeritiPro™ Thermal Cycler, 384-well, 1 Instrument Each
    7,592.00
    Special offer
    Online exclusive
    Ends: 01-Jun-2025
    11,680.00
    Save 4,088.00 (35%)
    Catalog # Name Size Price (USD) Qty
    A40070 Thermal Cycler Fleet Control Software Each
    2,180.00
    Resources
    Thermal Cycler Software and Firmware Files
    Download the latest updates for your instrument
    Interested in private label thermal cyclers?
    Submit an inquiry
    Support
    Service
    We provide a full range of service options for all Applied Biosystems thermal cyclers.
    View service plans
    Multiplex Dynamic Temperature Verification Service
    Instrument qualification service: Have one of our engineers perform temperature qualifications at your site.
    View temperature service options
     Download MDTV flyer
    For Research Use Only. Not for use in diagnostic procedures.