Protein Enrichment Strategies
Numerous techniques are available for the isolation of cellular proteins based on their post-translational modification (PTM) or active binding site. Protein enrichment is essential for isolating low-abundant proteins or isolating protein classes. Enrichment of specific proteins or protein complexes can most easily be accomplished using target-specific immobilized antibody or PTM specific affinity binding. The novel Thermo Scientific™ ActivX™ probes and enrichment kits enable the specific targeting of several enzyme classes, including kinases, GTPases, and serine hydrolases.
  • Comprehensive—reagents and kits for multiple protein enrichment strategies
  • Optimized—all reagents and kits are designed to maximize protein yield and reduce nonspecific binding
  • Convenient—easy-to-use spin formats enable more rapid enrichment of proteins
  • Flexible—kits can be used with either cultured mammalian cells or tissues
Choose the right protein enrichment kit for your application
 Phospho-
protein Enrichment Kit
Glycoprotein Enrichment Kit, ConAGlycoprotein Enrichment Kit, WGAUbiquitin Enrichment KitKinase Enrichment Kit with ATP ProbeActivX™ TAMRA-FP Serine Hydrolase Probe
TargetPhosphorylated proteinsα-linked mannose and terminal glucose residuesN-acetyl glucosamine (GlcNAc)Polyubiquitin-ated proteinsATP binding sitesActive serine hydrolase enzymes
Binding/
labeling mechanism
Metal chelate affinity to phosphate groupsLectin affinity to sugarsLectin affinity to sugarsAntibody affinity to targetBiotinylated ATP analog  to active-site lysineTAMRA-labeled fluoro
phosphonate binds to active site serine
Processing time2 hr<1 hr<1 hr<45 min6.5 hr3 hr
Enrichment formatIMAC-agaroseImmobilized conA-lectin-agaroseImmobilized WGA-lectin-agaroseImmobilized polyubiquitin affinity-agaroseImmobilized strepavidin-agaroseImmobilized anti-TAMRA mAb-agarose
 90003
(10 columns)
89804 (10 rxn)89805 (10 rxn)89899 (15 rxn)88310 (16 rxn)88318 (6.8 µg)
For downstream mass spectrometry analysis, peptide enrichment and/or fractionation is often necessary to detect peptides from low-abundance proteins and/or for reducing sample complexity for proteomic analysis.
Featured product data
Faster and higher phosphoprotein yields with the Pierce Phosphoprotein Enrichment Kit.
KitPhosphoprotein yield (ug)*Enrichment time (hours)
Pierce Phosphoprotein Enrichment Kit3001.5
Supplier Q kit884.5
Supplier I kit52**3.5
Supplier C kit1603
Supplier E kitToo dilute to determine5
*From 2 mg total protein.
**Based on maximum 1 mg load per manufacturer’s protocol.
Related products for protein enrichment
货号 产品名称 规格 单价 (CNY) 数量
89881 Pierce™ Cell Surface Protein Isolation Kit Each
9,199.00
16116 Active Rho Pull-Down and Detection Kit Each
11,551.00
88311 ActivX™ Desthiobiotin-ATP Probe Each
2,913.00
飞享价
截止至: 2025/12/31
3,596.00
共减 683.00 (19%)
88312 Pierce™ Kinase Enrichment Kit with ADP Probe Each
6,047.00
飞享价
截止至: 2025/12/31
7,465.00
共减 1,418.00 (19%)
88313 ActivX™ Desthiobiotin-ADP Probe Each
3,631.00
88315 ActivX™ Desthiobiotin-GTP Probe Each
2,754.00
飞享价
截止至: 2025/12/31
3,399.00
共减 645.00 (19%)
88316 ActivX™ Azido-FP Serine Hydrolase Probe Each
4,066.00
飞享价
截止至: 2025/12/31
5,019.00
共减 953.00 (19%)
88318 ActivX™ TAMRA-FP Serine Hydrolase Probe Each
4,146.00
飞享价
截止至: 2025/12/31
5,118.00
共减 972.00 (19%)
MA1041 TAMRA Monoclonal Antibody (5G5) 100 µg
5,701.00
20372 Pierce™ D-Galactose Agarose Each
3,350.00
仅供科研使用,不可用于诊断目的。