STEMPRO® Adipogenesis Differentiation Kit
STEMPRO® Adipogenesis Differentiation Kit
Complete differentiation of hMSCs into fat cells
STEMPRO® Chondrogenesis Differentiation Kit
Complete differentiation of hMSCs into cartilage cells
STEMPRO® Osteogenesis Differentiation Kit
Complete differentiation of hMSCs into bone cells
  • Easy to use
  • Reliable hMSC differentiation
  • Produced under cGMP
hMSC Differentiation Kits
  • Easy-to-use differentiation media kits demonstrate and satisfy multipotency criteria of hMSCs
  • Complements STEMPRO® MSC SFM, MesenPRO RS™ Medium, and MSC-qualified FBS cell expansion systems
  • Reliable induction of hMSCs into adipocyte progenitors/adipocytes, chondrocyte progenitors/cartilage and osteoblasts/osteocytes
  • Produced under cGMP; each lot is performance qualified using PCR
  • No or little adaptation required from cell expansion cultures
  • Reconstituted differentiation kits (basal medium plus supplement) are stable for up to one month.
STEMPRO® Adipogenesis Differentiation Kit
The STEMPRO® Adipogenesis Differentiation Kit supports adipogenesis from bone marrow–derived hMSCs (Figures 1A and 1B), human adipose-derived stem cells (ADSCs) (Figure 1C), and hMSCs derived from human embryonic stem cells.
Adipogenesis-induced differentiation using the STEMPRO® Adipogenesis Differentiation Kit
Figure 1.  Adipogenesis-induced differentiation using the STEMPRO® Adipogenesis Differentiation Kit.
A. Bone marrow–derived hMSCs induced under adipogenic conditions for 14 days, fixed and stained for oil red O, a marker for lipid-rich vesicles. 
B. Bone marrow derived hMSCs induced under adipogenic conditions for 7 days, fixed and lipid vesicles visualized with LipidTOX Green neutral lipid stain (green). Hoechst 33342 applied as a nuclear counterstain (blue).
C. Human ADSCs induced under adipogenic conditions for 12 days produce lipid containing adipocytes as demonstrated by oil red O staining (200X magnification). Data provided by K. Hicok, Cytori Therapeutics.
STEMPRO® Osteogenesis Differentiation Kit
The STEMPRO® Osteogenesis Differentiation Kit supports osteogenesis from bone marrow–derived hMSCs (Figures 2A and 2B), human adipose-derived stem cells (ADSCs) (Figure 2C), and hMSCs derived from human embryonic stem cells.
Osteogenesis-induced differentiation using the STEMPRO® Osteogenesis Differentiation Kit.
Figure 2. Osteogenesis-induced differentiation using the STEMPRO® Osteogenesis Differentiation Kit.
A. Bone marrow–derived hMSCs induced under osteogenic conditions for 14 days, fixed and stained for alkaline phosphatase (a marker for proliferating osteoblasts).
B. Bone marrow derived hMSCs induced under osteogenic conditions for 28 days, fixed and stained for Alizarin Red S (a dye that specifically binds to calcium matrix formations).
C. Human ADSCs induced under osteogenic conditions for 28 days produce calcium-containing mineralized matrix as demonstrated by alizarin red dye incorporation (200X magnification). Data provided by K. Hicok, Cytori Therapeutics.
Ordering Information
Catalog # Name Size Price (USD) Qty
A1007001 StemPro™ Adipogenesis Differentiation Kit Each
238.39
Special offer
Online exclusive
Ends: 21-Jun-2025
264.00
Save 25.61 (10%)
A1007101 StemPro™ Chondrogenesis Differentiation Kit Each
249.81
Special offer
Online exclusive
Ends: 21-Jun-2025
275.00
Save 25.19 (9%)
A1007201 StemPro™ Osteogenesis Differentiation Kit Each
238.74
Special offer
Online exclusive
Ends: 21-Jun-2025
264.00
Save 25.26 (10%)