showcase.par.50779.image.0.0.1
  • Low passage
  • Each lot from single donor
  • MesenPRO RS Medium reduces ADSC doubling time
Human adipose-derived stem cells
  • Human ADSCs isolated from human lipoaspirate tissue and cryopreserved from primary cultures
  • Low passage—ADSCs passaged once after isolation
  • Expanded in MesenPRO RS Medium—a revolutionary new reduced-serum (2%) medium that reduces ADSC doubling times
  • Kit contains human ADSCs and MesenPRO RS Medium—components ensure optimal ADSC growth and expansion
ADSCs have demonstrated very similar phenotypic and functional characteristics to bone marrow-derived mesenchymal stem cells. They can be expanded to 4–5 passages before they lose their ability to grow or differentiate into all potential phenotypes.
ADSC Characterization
Figure 1.  The cell surface protein profile of ADSCs was analyzed by flow cytometry at P2 or P3 and the following criteria was used to characterize the ADSCs 
Positive markers:
CD29, CD44, CD73, CD90, CD105, CD166 (>95%)
Negative markers:
CD14, CD31, CD45, Lin1 (< 2%).
Bar graph showing percentage of ADSCs expressing various cell surface proteins.
Differentiation potential of Human ADSCs
Each vial of ADSCs contains cells that can differentiate into multiple mature cell phenotypes in vitro including adipocytes, osteoblasts, and chondrocytes (Figure 1). In vitro differentiation into non-mesenchymal cell types, such as neuronal and glial progenitors, hepatocytes and vascular endothelial progenitors, cardiomyocytic, pancreatic and epithelial phenotypes has also been described. In addition, ADSCs secrete pro-angiogenic, immunomodulatory, and anti-apoptotic factors. ADSCs can be used for studies of adult stem cell differentiation, tissue engineering, and potential future clinical applications or as cells amenable to delivery of recombinant DNA constructs.
Figure 1: Three panel image showing ADSC-derived chondrocytes stained with safranin orange, ADSC-derived osteoblasts stained with Alizarin red, and ADSC-derived adipocytes stained with Oil Red O.
Figure 1.  Differentiation potential of human ADSCs
A. ADSCs induced to differentiate towards chondrocytes for 29 days and then stained with Safranin Orange dye ( pellet cross-sectional staining) for proteoglycan content, image captured using 4x objective lens.
B. ADSCs induced to differentiate towards osteoblasts for 29 days and then stained with Alizarin Red dye (which stains mineralized extracellular matrix), image captured using 4x objective lens.
C. ADSCs induced to differentiate towards adipocytes for 14 days and then stained with Oil-Red-O (which stains lipid vacuoles) and counterstained with hematoxylin, image captured using 10x objective lens.
Components of the STEMPRO Human ADSC Kit
 The Kit contains:
  1. Human adipose-derived stem cells (~1 x 106 cells)
  2. MesenPRO RS Medium
These components ensure optimal ADSC growth and expansion. Each lot of ADSCs originates from a single donor of human lipoaspirate tissue.
Ordering Information
Catalog # Name Size Price (USD) Qty
R7788110 StemPro™ Human Adipose-Derived Stem Cell Kit Each
1,128.00
12746012 MesenPRO RS™ Medium Each
265.09
Special offer
Online exclusive
Ends: 21-Jun-2025
307.00
Save 41.91 (14%)
15240062 Antibiotic-Antimycotic (100X), 100 mL Each
54.65
Online Exclusive
57.50
Save 2.85 (5%)
12662029 Fetal Bovine Serum, mesenchymal stem cell-qualified, USDA-approved regions, 500 mL Each
958.65
Online Exclusive
1,010.00
Save 51.35 (5%)
14190144 DPBS, no calcium, no magnesium, 500 mL bottle Each
30.65
Online Exclusive
32.63
Save 1.98 (6%)
25030081 L-Glutamine (200 mM), 100 mL Each
37.65
Online Exclusive
40.88
Save 3.23 (8%)
25200072 Trypsin-EDTA (0.25%), phenol red, 500 mL Each
64.25
12604013 TrypLE™ Express Enzyme (1X), no phenol red, 100 mL Each
38.65
Online Exclusive
42.22
Save 3.57 (8%)
15250061 Trypan Blue Solution, 0.4% Each
27.36
12585014 Insulin, human recombinant, zinc solution Each
72.75
23017015 Laminin Mouse Protein, Natural Each
264.65
Online Exclusive
286.00
Save 21.35 (7%)
410200 CD73 Monoclonal Antibody (7G2) 100 µg
488.00
AHU0051 AHU0051 -
MHCD10500 CD105 Monoclonal Antibody (SN6) 500 µL
381.00
398401 STRO-1 Monoclonal Antibody (STRO-1) 100 µL
474.00
PHG9204 Human TGF-beta 1 Recombinant Protein 5 µg
356.65
Online Exclusive
380.00
Save 23.35 (6%)
About STEMPRO Human ADSC Kit
Mesenchymal Stem Cell Research Tools
For Research Use Only. Not for use in diagnostic procedures.