Gattermann and Gattermann-Koch Formylation
In 1897, L Gattermann and J.A. Koch successfully introduced an aldehyde group onto toluene by using formyl chloride (HCOCl) as an acylating agent under Friedel-Crafts acylation conditions. Subsequently, the addition of a formyl group into electron-rich aromatic rings by application of CO/HCl/Lewis acid catalysts (AlX3, FeX3, where X = Cl, Br, I) to prepare aromatic aldehydes became known as the Gattermann-Koch formylation.
The scope of the Gattermann-Koch reaction is limited due to the lack of suitable substrates, as it is mostly restricted to alkylbenzenes; so Gattermann introduced a modification which allowed the formylation of phenols, phenolic ethers, and heteroaromatic compounds such as pyrroles and indoles.
The main drawback of the Gattermann formylation was that it called for the use of anhydrous hydrogen cyanide (HCN). To avoid handling HCN, R. Adams generated it in situ from zinc cyanide and hydrochloric acid, and this became known as the Adams modification. This method has become more widely used in organic synthesis.
Mechanism of the Gattermann Formylation using the Adams Modification
The benzofuran-derived natural product, caleprunin A, was synthesized by R. Stevenson, et al. using the Gattermann formylation as the key step.
Compounds with applications as diverse as fluorescent probes of peptide structures in dynamic environments and potent anti-tumor agents have been synthesized using the Gattermann formylation.
Review available Thermo Scientific products that are used in Gattermann-Koch reactions:
Catalog # Name Size Price (USD) Qty
L15836.14 Zinc cyanide, 98% Each
34.65
Online Exclusive
38.10
Save 3.45 (9%)
L15836.22 Zinc cyanide, 98% Each
35.65
Special offer
Online exclusive
Ends: 30-Jun-2025
41.20
Save 5.55 (13%)
L15836.0B Zinc cyanide, 98% Each
127.65
Special offer
Online exclusive
Ends: 30-Jun-2025
150.00
Save 22.35 (15%)
Catalog # Name Size Price (USD) Qty
44470.06 -
44470.14 -
88488.06 -
A11892.30 Aluminum chloride, anhydrous, granular, 99% Each
30.65
Special offer
Online exclusive
Ends: 30-Jun-2025
36.30
Save 5.65 (16%)
A11892.0B Aluminum chloride, anhydrous, granular, 99% Each
80.65
Special offer
Online exclusive
Ends: 26-Sep-2025
94.40
Save 13.75 (15%)
L18489.22 Aluminum chloride, 98+%, extra pure, anhydrous powder Each
46.65
Special offer
Online exclusive
Ends: 30-Jun-2025
54.70
Save 8.05 (15%)
L18489.36 Aluminum chloride, 98+%, extra pure, anhydrous powder Each
88.65
Online Exclusive
98.50
Save 9.85 (10%)
14552.06 -
14552.14 -
14552.22 -
44313.14 -
44313.22 -
43879.06 Each -
43879.14 -
Vote for your favorite named reaction
Learn more about the most popular named reactions in chemistry based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now