The Mitsunobu reaction—a nucleophilic substitution reaction
In 1967, O. Mitsunobu demonstrated the acylation of secondary alcohols with carboxylic acids in the presence of diethyl azodicarboxylate (DEAD) and triphenylphosphine. Later it was discovered that optically active secondary alcohols underwent total inversion of configuration under these reaction conditions, and this procedure was found to be general for the synthesis of optically active amines, azides, ethers, thioethers, and alkanes. Since then, the substitution of primary and secondary alcohols with nucleophiles in the presence of a dialkylazodicarboxylate and a trialkyl- or triarylphosphine has become known as the Mitsunobu reaction.
An important development of this reaction was made by T. Mukaiyama, who prepared inverted tert-alkyl carboxylates from chiral tertiary alcohols via alkoxydiphenylphosphines formed in situ from 2,6-dimethyl-1,4-benzoquinone.
The synthesis of the potent antitumor antibiotic (+)-duocarmycin A utilized the Mitsunobu reaction during the final stage, a special case where the reaction is used to create new carbon-carbon bonds.
Mechanism of the Mitsunobu reaction
Review available Thermo Scientific products for the Mitsunobu reaction:
Catalog # Name Size Price (USD)
L10386.14 Diisopropyl azodicarboxylate, 94% Each
54.65
Online Exclusive
60.50
Save 5.85 (10%)
L10386.22 Diisopropyl azodicarboxylate, 94% Each
96.65
Special offer
Online exclusive
Ends: 26-Sep-2025
114.00
Save 17.35 (15%)
45131.14
45131.36
A11437.14 Dimethyl acetylenedicarboxylate, 98% Each
73.65
Special offer
Online exclusive
Ends: 30-Jun-2025
86.80
Save 13.15 (15%)
A11437.22 Dimethyl acetylenedicarboxylate, 98% Each
226.65
Special offer
Online exclusive
Ends: 30-Jun-2025
266.00
Save 39.35 (15%)
A11437.36 Dimethyl acetylenedicarboxylate, 98% Each
1,040.65
Special offer
Online exclusive
Ends: 30-Jun-2025
1,224.00
Save 183.35 (15%)
L02502.18 Triphenylphosphine, powder, 99% Each
37.65
Online Exclusive
41.30
Save 3.65 (9%)
L02502.30 Triphenylphosphine, powder, 99% Each
73.65
Special offer
Online exclusive
Ends: 30-Jun-2025
86.40
Save 12.75 (15%)
L02502.0B Triphenylphosphine, powder, 99% Each
237.65
Special offer
Online exclusive
Ends: 30-Jun-2025
279.00
Save 41.35 (15%)
14112.14
A14089.30 Triphenylphosphine, flake, 99% Each
51.65
Special offer
Online exclusive
Ends: 26-Sep-2025
60.80
Save 9.15 (15%)
A14089.0B Triphenylphosphine, flake, 99% Each
147.65
Special offer
Online exclusive
Ends: 30-Jun-2025
174.00
Save 26.35 (15%)
A14089.0I Triphenylphosphine, flake, 99% Each
528.65
Special offer
Online exclusive
Ends: 30-Jun-2025
622.00
Save 93.35 (15%)
Vote for your favorite named reaction
Learn more about the most popular named reactions based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now