Search Thermo Fisher Scientific
Organism Type | Human | The GripTite™ 293 MSR Cell Line is a genetically engineered Human Embryonic Kidney (HEK 293) cell line that expresses the human macrophage scavenger receptor and strongly adheres to standard tissue culture plates for dependable results. Developed from a 293-H subclone, GripTite™ 293 MSR cells show the same fast cell growth, high transfection efficiency, and high-level expression characteristics of the parental 293-H cells. Unlike most 293 cells, GripTite® 293 MSR cells don't wash away during the repeated manipulations of routine tissue culture or plate washing protocols. Their superior adherence enables reliable reproduction of ligand-binding, enzymatic, or immunofluorescence assay results. |
Tissues | Kidney | |
Phenotype | Adherent | |
Primary | no | |
Application | Transfection: Gene Expression |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||