Nazarov Cyclization
In 1941, the Russian chemist Ivan Nikolaevich Nazarov extensively studied the cyclization reactions of allyl vinyl ketones to the corresponding 2-cyclopentanones. Although cyclizations following this general procedure had been observed before Nazarov's work, it was his study of the rearrangements of allyl vinyl ketones that marked the first close examination of this process. Nazarov correctly reasoned that the allylic olefin isomerized in situ to form a divinyl ketone before ring closure to the cyclopentenone product. Subsequently, the protic or Lewis acid-catalyzed ring closure of divinyl ketones through formation of the corresponding pentadienylic cations became known as the Nazarov cyclization.
This reaction became useful in organic synthesis due to the frequent appearance of the cyclopentanone ring in natural products and in the formation of synthetic intermediates in total synthesis campaigns. For example, in the synthesis of (±)-trichodiene by K.E. Harding and colleagues, the Nazarov cyclization was employed as a key ring-forming step to control the stereochemistry.
Mechanism of the Nazarov cyclization reaction
Review available Thermo Scientific products for the Nazarov cyclization reaction:
Catalog # Name Size Price (USD) Qty
L04295.30 Acetic anhydride, 99+% Each
34.90
L04295.0B Acetic anhydride, 99+% Each
72.30
L04295.0E Acetic anhydride, 99+% Each
111.00
A16037.36 Sodium hydroxide, pellets, 98% Each
30.65
Special offer
Online exclusive
Ends: 30-Jun-2025
36.00
Save 5.35 (15%)
A18395.36 Sodium hydroxide, flake, 98% Each
30.65
Special offer
Online exclusive
Ends: 26-Sep-2025
35.30
Save 4.65 (13%)
A18395.0E Sodium hydroxide, flake, 98% Each
87.65
Special offer
Online exclusive
Ends: 30-Jun-2025
103.00
Save 15.35 (15%)
A18395.0C Sodium hydroxide, flake, 98% Each
285.65
Online Exclusive
317.00
Save 31.35 (10%)
A15275.AE Boron trifluoride diethyl etherate, 98+% Each
35.65
Special offer
Online exclusive
Ends: 26-Sep-2025
42.10
Save 6.45 (15%)
A15275.AP Boron trifluoride diethyl etherate, 98+% Each
65.65
Online Exclusive
73.10
Save 7.45 (10%)
A15275.AU Boron trifluoride diethyl etherate, 98+% Each
102.65
Special offer
Online exclusive
Ends: 30-Jun-2025
120.00
Save 17.35 (14%)
Vote for your favorite named reaction
Learn more about the most popular named reactions in chemistry based on feedback from our customers. Do you have a favorite reaction? Let us know!
Vote now