Selenium • Non-Metals
Primary XPS region: Se3d
Overlapping regions: Li1s, Mg2p
Binding energies of common chemical states:
Chemical stateBinding energy Se3d5/2
Se metal55.1 eV
Experimental information
N/A
Interpretation of XPS spectra
  • Se3d peak has overlapping spin-orbit components (Δ=0.86eV, intensity ratio=0.735).
    • Loss feature is located at 58.5 eV, visible in Se3d spectrum for metal.
  • Se LMM peaks (between 150eV–400eV) overlap with many XPS peaks from other elements and selenium’s own XPS peaks.
About this element
Symbol: Se
Date of discovery: 1817
Name origin: Greek selênê
Appearance: gray
Discoverer: Jöns Berzelius
Obtained from: refining of lead, copper, nickel
Melting point: 4904 K
Boiling point: 958 K
Density[kg/m3]: 4819
Molar volume: 16.42 × 10-6 m3/mol
Protons/Electrons: 34
Neutrons: 45
Shell structure: 2,8,18,6
Electron configuration: [Ar]3d104s24p4
Oxidation state: ±2,4,6
Crystal structure: monoclinic, hexagonal
Selenium was discovered by Jöns Jakob Berzelius 1817 and derives its name from the Greek word "selene", meaning moon. Although selenium is naturally found in a few rare minerals, it is primarily produced as a byproduct of copper refining. Selenium exists in two crystalline forms: crystalline monoclinic selenium is a deep red, while crystalline hexagonal selenium, the most stable form, is metallic gray. Selenium’s photovoltaic and photoconductive properties contribute to its use in photocopying, toning of photographic prints, photocells, light meters, and solar cells. A semi-conductive material below its melting point, selenium is also used in electronic applications.
Join-the-conversation-banner-in-blogs-social-graphic-1143250-1000x172
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.