Real-Time PCR Software for Biopharmaceutical Analytical Assays
 AccuSEQ integrated software for residual host cell DNA testing and mycoplasma detection testing
The AccuSEQ Real-Time PCR Software provides fast and easy data interpretation of analytical output from contaminants and impurities testing. This AccuSEQ software is a key component of an all-inclusive solution that provides you with the actionable results needed during biopharmaceutical manufacturing processes. The AccuSEQ Real-Time PCR Software is part of a complete validated solution from sample prep, real-time PCR, data analysis, and report output for contaminant and impurity testing.
Contact us  Order now
The AccuSEQ software was designed to facilitate the data interpretation of essential tests required for GMP manufacturing, including in-process testing and lot-release testing.
  • Integrated, easy-to-use interface allows for consistency across multiple data applications and reduces the time needed to train your team
  • Single software platform for all current and future resDNASEQ, MycoSEQ, and ViralSEQ real-time PCR assays, as well as custom experiments
  • Security, audit, and e-signature capabilities to help enable 21 CFR Part 11 compliance
  • Available for 7500 Fast and QuantStudio 5, Block PCR instruments for easy integration of SEQ solutions into your current QC laboratory
  • Flexible throughput of 10–500 samples per week
  • Complete traceability with assay specific output, eliminating manual calculations
  • Comprehensive data management and reporting features
Workflow
Thermo Fisher Scientific offers an integrated solution for the entire analytical workflow, consisting of sample prep, SEQ assays, qPCR Instrument and the AccuSEQ software. During sample preparation, enable reliable, consistent recovery of your target DNA and RNA with Thermo Fisher’s proven PrepSEQ chemistry. There are various assays to detect impurities, identify contamination, and quantify titers using the resDNASEQ, MycoSEQ, and ViralSEQ family of assays. Finally, you have the AccuSEQ Software to run the assays on the 7500 Fast/QuantStudio 5 instruments and to analyze the data from the instrument to produce actionable results.
Integrated products for analytic workflow
On this page
Getting the right start
Create custom templates tailored to your needs
The AccuSEQ Software v3.2 dashboard is intuitive and user-friendly. Use the home page to start a new experiment, get immediate updates for ongoing experiments, and access quick links to the most recent experiments. Find your most recent experiments under “Home”. Check the status and time remaining of your real-time PCR run under “Monitor The Run”. Browse through factory default templates (validated and optimized templates for assays including MycoSEQ and resDNASEQ), user saved templates, or create custom templates tailored to your needs under “Create New Experiment”.
Key features
The AccuSEQ Software v3.2 plate set up interface provides customers a visual approach to setting up their experiment details on a 96-well plate. Users can input details (sample, controls, dilutions, etc.) and designate colors for each well, allowing for quick visualization of the 96-well layout. Table-view plate setup is also available. The “Define & Setup Standard” feature allows users to expedite their standards set up on the plate. Simply enter information about the target and standard curve parameters, and this feature provides well arrangement on the plate for the Standards.
Plate setup for Mycoplasma Plus Kit using AccuSEQ Software v3.2
The AccuSEQ Software allows for great flexibility with experiments, with options to modify call settings to meet individual specifications. This feature allows customers to modify the thresholds for every call built as part of the decision tree in a presence/absence call. The AccuSEQ Software can also review calls to be overridden. This can be done when you do not agree with the system calls and want to override it with your own assessment.
Manual call edit feature on AccuSEQ Software v3.2
Manual call edit feature on AccuSEQ Software v3.2 using 7500 Fast System
Predefined templates allow for a streamlined experiment set-up, where factory default templates for Thermo Fisher Scientific’s SEQ products get you up and running faster.
AccuSEQ Software v3.2 factory default templates
AccuSEQ Software v3.2 factory default templates
AccuSEQ Software v2.2 factory default templates
AccuSEQ Software v2.2 factory default templates
The AccuSEQ Software v3.2 plate set up interface provides customers a visual approach to setting up their experiment details on a 96-well plate. Users can input details (sample, controls, dilutions, etc.) and designate colors for each well, allowing for quick visualization of the 96-well layout. Table-view plate setup is also available. The “Define & Setup Standard” feature allows users to expedite their standards set up on the plate. Simply enter information about the target and standard curve parameters, and this feature provides well arrangement on the plate for the Standards.
Plate setup for Mycoplasma Plus Kit using AccuSEQ Software v3.2
The AccuSEQ Software allows for great flexibility with experiments, with options to modify call settings to meet individual specifications. This feature allows customers to modify the thresholds for every call built as part of the decision tree in a presence/absence call. The AccuSEQ Software can also review calls to be overridden. This can be done when you do not agree with the system calls and want to override it with your own assessment.
Manual call edit feature on AccuSEQ Software v3.2
Manual call edit feature on AccuSEQ Software v3.2 using 7500 Fast System
Predefined templates allow for a streamlined experiment set-up, where factory default templates for Thermo Fisher Scientific’s SEQ products get you up and running faster.
AccuSEQ Software v3.2 factory default templates
AccuSEQ Software v3.2 factory default templates
AccuSEQ Software v2.2 factory default templates
AccuSEQ Software v2.2 factory default templates
Mycoplasma detection using the AccuSEQ Software
For rapid and accurate mycoplasma detection, the AccuSEQ software offers a plug-in analysis module for automated presence or absence calls when using the MycoSEQ Mycoplasma Detection system.
Key features and benefits:
  • Optimized and validated for the MycoSEQ Mycoplasma Detection assay
  • Automated detection of PCR inhibition in sample wells
  • Automated detection of discriminatory positive control (DPC)
  • Robust data analysis parameters for the MycoSEQ Mycoplasma Detection assay
  • Flexibility for experiments; call settings may also be modified to meet the specifications of individual laboratories
Residual host cell DNA analysis using AccuSEQ software
The AccuSEQ software offers factory default templates to facilitate experiment set up for the accurate and sensitive quantitation of host cell DNA impurities in both the purification process and in drug substance samples.
Key features and benefits:
  • Streamlined resDNASEQ experiment setup (using factory default templates) and data reporting
  • Automatic calculations of the resDNASEQ standard curve with curve fitting parameters
  • Accurate quantitation of samples with % CV
  • Computation of key metrics including % Recovery, Total DNA in pg/mL, and pg DNA/mg Protein
  • Flexibility for custom experiments to meet the specifications of individual laboratories
  • Customizable output layout that can be exported to a variety of formats
  • Comprehensive reports which help eliminate the need for Excel sheets for calculations and preserve the audit trail
After the experiment run, the AccuSEQ Software v3.2 automatically sets up the standard curve calculations and provides a visual display of the results with relevant curve fitting parameters, such as slope, intercept and efficiency, and R2.
The AccuSEQ Software v3.2 shows the standard curve plot with samples overlayed and the results analysis tab on the right.
The resDNASEQ Results Table in AccuSEQ Software v3.2 shows key metrics including Quantity Mean, Quantity % CV, % Recovery, Total DNA per mL, and Total DNA per Protein Concentration.
The report section allows results to be exported in multiple formats (.xls, .xlsx, .txt, and .csv) and provides the ability to customize how this content appears in a printed report.
The AccuSEQ Software v3.2 showing the Print report section where users can customize report contents.
Snapshot of the resDNASEQ “Result Summary” from the PDF report generated by the v3.2 software. The Comprehensive Result Summary includes all important metrics required in the resDNASEQ experiment such as % CV, % Recovery, and Total DNA per mL (dilution adjusted).
After the experiment run, the AccuSEQ Software v3.2 automatically sets up the standard curve calculations and provides a visual display of the results with relevant curve fitting parameters, such as slope, intercept and efficiency, and R2.
The AccuSEQ Software v3.2 shows the standard curve plot with samples overlayed and the results analysis tab on the right.
The resDNASEQ Results Table in AccuSEQ Software v3.2 shows key metrics including Quantity Mean, Quantity % CV, % Recovery, Total DNA per mL, and Total DNA per Protein Concentration.
The report section allows results to be exported in multiple formats (.xls, .xlsx, .txt, and .csv) and provides the ability to customize how this content appears in a printed report.
The AccuSEQ Software v3.2 showing the Print report section where users can customize report contents.
Snapshot of the resDNASEQ “Result Summary” from the PDF report generated by the v3.2 software. The Comprehensive Result Summary includes all important metrics required in the resDNASEQ experiment such as % CV, % Recovery, and Total DNA per mL (dilution adjusted).
Rapid sterility testing using the AccuSEQ software
For rapid and accurate sterility testing, the AccuSEQ software offers presence or absence calls when using the SteriSEQ Rapid Sterility Testing Kit, allowing for easy analysis.
Key features and benefits:
  • Automated analysis with presence/absence calls
  • Bulk edit feature to allow for custom rule edits
  • Discriminatory positive control to help eliminate false positives
  • Predefined templates to allow for streamlined experiment setup
  • Comprehensive reports which help eliminate the need for Excel sheets for calculations and preserve the audit trail
The AccuSEQ Software v3.2 shows the “automated presence or absence calls” of sterility testing.
ViralSEQ information
Detection of MMV and vesivirus with AccuSEQ software
For rapid and accurate MMV and vesivirus detection, the AccuSEQ software offers:
  • Robust, optimized, and validated system—data analysis designed for use with ViralSEQ Detection Systems
  • Automated detection—rapid assessment of PCR inhibition in sample wells
  • Analysis options—identify true positive from false positives with discriminatory positive control
  • Flexible call settings—meets specifications for unique lab requirements
Quantitation of Sf-rhabdovirus with AccuSEQ software
The AccuSEQ software offers factory streamlined experimental set up for the accurate and sensitive quantitation of Sf-rhabodvirus contamination in both the purification process and in drug substance samples.
Key features and benefits:
  • Streamlined experiment set-up—use factory default templates for easy data reporting
  • Automated calculations—help eliminate the need for Excel sheets for calculations and preserve the audit trail
  • Comprehensive reports—compute key metrics including percent recovery, total RNA in pg/mL, and pg RNA/mg Protein
  • Flexibility—easily adjust for custom experiments to meet the specifications and reporting needs
Custom experiments using AccuSEQ software
There is also the custom experiment mode (“Create New”) where users can use the AccuSEQ software to create custom experiment setups. The custom mode guides the user through the experiment properties, qPCR run method details, and plate setup where there are easy-to-use tools for defining and setting up standards, controls, and unknowns. This custom workflow also supports ViralSEQ Mouse Minute Virus (MMV), vesivirus, and resDNASEQ SF9-Baculovirus duplex assays.
Customized output for SEQ Analytical Assays
The AccuSEQ Software produces customized output for SEQ assays, helping users to export complete, traceable, and simple outputs, greatly streamlining the analysis process. Rather than outputting Ct and requiring further downstream calculations, the customized output feature allows you to have a specialized output to match your needs. For example, the LentiQuant physical titer outputs the viral particles/mL that help determine the titer of lentivirus in viral supernatants and purified viruses.
Assay
Specialized output
ViralSEQ MMV detection kit
  • Call
  • Call assessment 
ViralSEQ Vesivirus detection kit 
  • Call
  • Call assessment 
MycoSEQ Plus mycoplasma detection kit 
  • Call
  • Call assessment 
SteriSEQ detection kit 
  • Call
  • Call assessment 
All resDNASEQ Quantitative Kits
  • All resDNASEQ Quantitative Kits
           Ct, Ct mean
  • Qty, Qty mean, Qty % CV
  • Dilution adjusted
  • % Recovery
resDNASEQ Quantitative E1A DNA Fragment Length Kits
  • All resDNASEQ Quantitative Kits            
            Ct, Ct mean
  • Qty, Qty mean, Qty % CV
  • Dilution adjusted
  • % Recovery
resDNASEQ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kits
ViralSEQ Quantitative Sf-Rhabdovirus Kit
  • % Recovery
  • Mass (g)
ViralSEQ Lentivirus Physical Titer Kit
  • Viral particles per mL (VP/mL)
ViralSEQ Lentivirus Proviral DNA Titer Kit
  • Provirus copies per cell
Customized output for Quantitative Sf9 and Baculovirus DNA Kit using the AccuSEQ Software v3.2
Customized output for Quantitative Sf9 and Baculovirus DNA Kit using the AccuSEQ Software v2.2
Customized output for Lentivirus Physical Titer Kit using the AccuSEQ Software v3.2
Customized output for Lentivirus Physical Titer Kit using the AccuSEQ Software v2.2
Assay
Specialized output
ViralSEQ MMV detection kit
  • Call
  • Call assessment 
ViralSEQ Vesivirus detection kit 
  • Call
  • Call assessment 
MycoSEQ Plus mycoplasma detection kit 
  • Call
  • Call assessment 
SteriSEQ detection kit 
  • Call
  • Call assessment 
All resDNASEQ Quantitative Kits
  • All resDNASEQ Quantitative Kits
           Ct, Ct mean
  • Qty, Qty mean, Qty % CV
  • Dilution adjusted
  • % Recovery
resDNASEQ Quantitative E1A DNA Fragment Length Kits
  • All resDNASEQ Quantitative Kits            
            Ct, Ct mean
  • Qty, Qty mean, Qty % CV
  • Dilution adjusted
  • % Recovery
resDNASEQ Quantitative Plasmid DNA - Kanamycin Resistance Gene Kits
ViralSEQ Quantitative Sf-Rhabdovirus Kit
  • % Recovery
  • Mass (g)
ViralSEQ Lentivirus Physical Titer Kit
  • Viral particles per mL (VP/mL)
ViralSEQ Lentivirus Proviral DNA Titer Kit
  • Provirus copies per cell
Customized output for Quantitative Sf9 and Baculovirus DNA Kit using the AccuSEQ Software v3.2
Customized output for Quantitative Sf9 and Baculovirus DNA Kit using the AccuSEQ Software v2.2
Customized output for Lentivirus Physical Titer Kit using the AccuSEQ Software v3.2
Customized output for Lentivirus Physical Titer Kit using the AccuSEQ Software v2.2
Security, audit, and E-signature (SAE) feature and compliance services for AccuSEQ software
Security, audit, and E-signature (SAE) is a key feature of the AccuSEQ software as it supports our customers in meeting their local regulations like 21 CFR Part 11 compliance. The software security, audit, and electronic signature (SAE) manager includes options that let you easily organize and track your security settings, such as:
  • Security—username and password restrictions, and security policies such as password expiration and user account suspension
  • Audit—automatically audits changes to experiments in the AccuSEQ Software and modifications to settings in the SAE module, and sorts and prints audit records at the experiment and systems level
  • Electronic signature—selection of an electronic signature mode (enable or disable), on-demand electronic signature, electronic signature settings, number of electronic signatures required, and electronic signature actions (review, approve, or sign off)
Other key benefits include:
  • Database storage for improved data security
  • Customizable user role and permission
  • Two levels of auditing including comprehensive reporting of audit events
Since AccuSEQ software is validated with respective real-time PCR systems (7500Fast and QuantStudio 5, 0.1mL block), Thermo Fisher Scientific offers compliance services like IQOQ (installation and operation qualification) and CSV (computer system validation) services that save customers time and money with compliance audits. For more information on compliance services, please contact your local sales or field support representative.
Computer System Validation (CSV)
This comprehensive document starts with a master validation and ends with a summary report. The Thermo Fisher Scientific Computer System Validation (CSV) consulting service develops and produces results in documentation form as tangible evidence, assuring that all software features within scope of data security, auditing, and e-signature are suitable for intended use.
The CSV offering provides complete or partial consulting services with 12 industry standard validation documents and execution for collection of objective evidence:
Validation Plan
Operational Qualification (OQ) with objective evidence
Validation Risk Assessment (RA)
Performance Qualification (PQ) with objective evidence
User Requirements Specification (URS)
Traceability matrix
System Configuration Specification (SCS)
21 CFR Part 11 / Annex 11 / GAMP™ 5 Assessment
Validation test plan
Quality assurance review
Installation Qualification (IQ) with objective evidence
Validation summary report
AccuSEQ File Management Tool
The AccuSEQ File Management Tool is a specially designed feature that allows non-IT/database customers to manage their data in the AccuSEQ database with a simple user interface. The File Management Tool is built to help with backups, archiving, file access, as well as security.
Data in AccuSEQ Real-Time PCR Software v3.2 can be backed up in three ways:
AccuSEQ File Management Tool—Use the tool to backup, purge, or restore selected experiments in a date range or to backup all data stored in the AccuSEQ Real‑Time PCR Software v3.2. Use the tool to:
  • Backup experiments—Backup selected experiments in a date range or backup all data stored in the AccuSEQ Real‑Time PCR Software v3.2 database
  • Purge experiments—Permanently delete selected experiments from the database
  • Restore experiments—Restore selected experiments in a date range or restore all data stored in the AccuSEQ Real‑Time PCR Software v3.2 database.
Manual backup—Use export functions in the software to backup after each experiment. You can:
  • Manually export the experiment data to an EDS file
  • Manually export the experiment and audit reports to PDF files
  • Manually export new templates to an EDT file
  • Manually export SAE audit records (if changes were made)
Manual export of EDS, EDT, and PDF files can take several minutes per experiment. Only EDS and EDT files can be reimported. PDF files cannot be reimported.
Backup and Restore Utility—Use to backup experiments stored in AccuSEQ Real‑Time PCR Software v3.2
Selecting the right AccuSEQ Software for you
The Applied Biosystems QuantStudio 5, 0.1 mL block system is the newer real-time PCR system validated with AccuSEQ Software v3.2.
Learn more about our QS5, 0.1 mL system ›
The Applied Biosystems 7500 Fast system is our real-time PCR system validated with AccuSEQ Software v2.2.
Learn more about our 7500 Fast system ›
 AccuSEQ v3.2 SoftwareAccuSEQ v2.2 Software
Instrument supportedQuantStudio 5, 0.1 mL block7500 FAST
CompatibilityWin10Win10
InterfaceBrowser-basedApplication-based
Templates predefined
MycoSEQ
MycoSEQ Plus
ResDNASEQ
ViralSEQ
MycoSEQ
MycoSEQ Plus
ResDNASEQ
ViralSEQ
Template format
Customizable template option
Open or locked
Customizable template
Open
Data storageDatabaseFile-based
Data back-up and restoreAvailableNot available
Data securityImproved access controlStandard
Ordering information
Analytics Knowledge Hub
Explore the Analytics Knowledge Hub from Thermo Fisher Scientific, providing a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our products in your bioprocess workflow.
Explore the Hub