PrepSEQ reagents for nucleic acid extraction, dna purification, and genomic DNA isolation
Solutions for contaminant and impurity testing
Optimization for the best recovery of your genomic DNA and RNA is crucial for in-process and lot release testing and requires the best sample preparation possible. The success of a QC analytical test is critically dependent on the sample preparation used to remove PCR interferants. We offer a range of sample prep solutions using our reliable PrepSEQ chemistry in both manual and automated formats to enable superior recovery efficiency for applications where consistent absolute quantitation and high sensitivity detection are required.
Contact us  Order now
PrepSEQ chemistry is a universal sample preparation workflow for contaminant and impurity testing used in multiple applications, including residual host cell DNA quantitation, mycoplasma detection, Minute Mouse Virus (MMV), and viral detection. The benefits of the PrepSEQ kit include:
  • Easy-to-use—simple protocol featuring magnetic bead based technology for efficient sample extraction, clean up and elution
  • Sensitive—isolation of genomic DNA from total lysate of test samples
  • Increased throughput with flexible automation formats
  • Rapid—routine screening of high-value cell lines in combination with the resDNASEQ Quantitative Residual Host Cell DNA Kit
  • Accurate—determination of absence or presence of contaminants with the MycoSEQ Mycoplasma Detection Kit
Universal sample preparation for impurity and contaminant testing
Get reliable, consistent quantitative recovery of your target DNA and RNA with a comprehensive solution that is built on years of successful performance. PrepSEQ technology enables consistent recovery from matrices varying in complexity, whether it is pH, salt quantity, or protein balance. The nucleic acid extraction workflow includes our proprietary magnetic particle and binding solution that not only enables high binding but complete elution of the purified sample. The PrepSEQ solution offers flexible protocols for manual sample preparation or automation protocols, allowing you to process 1–96 extractions at one time, with high recovery to help enable you to meet regulatory guidelines. 
PrepSEQ nucleic acid extraction workflow
Figure 1. The PrepSEQ nucleic acid extraction workflow includes our proprietary magnetic particle and binding solution that not only enables high binding but complete elution of the purified sample.
Automated sample preparation solutions
Boost lab productivity by reducing hands-on sample preparation time, increasing throughput, reducing costs, and improving the quality of sample extractions. Intuitive to operate and pre-loaded with protocols, our automated sample preparation instruments help meet your throughput needs. Our automated solutions offer multi-assay, time-saving, cost-effective DNA extraction for residual host cell DNA, mycoplasma, MMV, and Vesivirus.
Sample preparation solutions for residual DNA and RNA quantitation
Applied Biosystems PrepSEQ Residual DNA Sample Preparation Kit
PrepSEQ Residual DNA Sample Preparation Kits are optimized for consistent recovery of DNA and RNA from various sample matrices. The kit has flexible protocols for both manual and automated workflows to enable consistent, reliable recovery for even the most challenging samples. The PrepSEQ Residual DNA Sample Prep Kit can be used with the resDNASEQ quantitative residual host cell DNA kits that are available for CHO, Human, HEK293, E. coli, Vero, MDCK, NS0, and Pichia pastoris cell lines.
Thermo Scientific KingFisher Apex and Kingfisher Flex Sample Purification Instruments
We offer a high throughput solution for the automation of the PrepSEQ workflow to purify samples for residual host cell DNA analysis. The complete sample DNA and RNA extraction and 96-well preparation for qPCR analysis is achieved in less than three hours using PrepSEQ/resDNASEQ kits. The KingFisher Apex and KingFisher Flex Instruments automate the PrepSEQ workflow to deliver reliable high DNA/RNA recovery with consistent performance, regardless of sample complexity.
Read more ›
Applied Biosystems PrepSEQ Nucleic Acid Sample Preparation Kit
The PrepSEQ Nucleic Acid Sample Preparation Kit is optimized for highly efficient, quantitative, and consistent recovery of DNA and RNA from diverse samples types, including cell culture, raw material (e.g., serum), and in-process downstream purification samples, such as column purification or bulk drug substance samples.
Sample preparation solutions for contaminant detection
Applied Biosystems PrepSEQ 1-2-3 Nucleic Acid Extraction Kit
Conduct manual sample preparation for the extraction of DNA and RNA from mycoplasma cells, MMV isolates, and/or Vesivirus isolates. Minimizing cellular DNA and/or RNA in the final sample extraction is critical to mycoplasma DNA detection where high amounts of cellular DNA and RNA cause PCR inhibition and high background. PrepSEQ 1-2-3 kits allow for sample cleanup of mycoplasma genomic DNA from research cell lines, stem cell lines, cellular, and tissue therapy samples with high efficiency. The workflow offers a rapid and easy to use protocol for results in under 2 hours.
AutoMate Express Nucleic Acid Extraction System
We offer an easy-to-use benchtop instrument that enables hands-free automation of the nucleic acid extraction process for the MycoSEQ Mycoplasma Detection, ViralSEQ MMV Detection, and ViralSEQ Vesivirus Detection testing workflows. The instrument uses the established PrepSEQ nucleic acid extraction chemistry packaged into a cartridge format for quantitative recovery of DNA and RNA. The Automate Express offers out-of-the-box automation for easy integration into existing workflows and lab configurations. There is minimal hands-on time with this closed system that is “load and go” with prefilled reagent cartridges and preprogrammed protocols. The Automate Express magnetic particle processor instrument offers low to medium throughput of up to 13 extractions in one run. Read more ›
PrepSEQ Express Nucleic Acid Extraction Kit
This ready-to-go kit contains pre-filled reagents and plastic tubes and tips to perform hands-free extraction of purified nucleic acid from 52 samples. The kit is designed for use on the AutoMate Express Nucleic Acid Extraction System. The cartridges utilize the established PrepSEQ nucleic acid extraction chemistry for quantitative recovery of DNA and RNA from diverse samples types, including cell culture, raw material, and in-process samples. Learn more ›
Ordering information—PrepSEQ products for residual DNA and RNA quantitation
Ordering information—PrepSEQ products for contaminant testing
Analytics Knowledge Hub
Explore the Analytics Knowledge Hub from Thermo Fisher Scientific, providing a robust collection of webinars, articles, eBooks, infographics, and other digital resources to help you optimize the use of our products in your bioprocess workflow.
Explore the Hub
Bioprocessing resources
Access a targeted collection of scientific application notes, case studies, posters, white papers and more for bioprocessing:
For Research Use Only. Not for use in diagnostic procedures.