Find the best fit for your business
Many factors can go into the selection of a food safety metal detector - from the unique challenges of your application from product content that may mimic metals to packaging materials, environmental noise, washdown procedures, line location and more.  The comparison table below can help to determine which system best supports your needs, based on popular requirements. 
For assistance in determining the best system for your needs or to talk with a specialist, click the Contact Sales button above.
Compare metal detectors
SENTINEL 5000
SENTINEL 3000
SENTINEL 1000
APEX 300
APEX 100
APEX Rx
Conveyor Install Supported
Combo with Checkweigher Install Supported
External infeed
Integral to checkweigher frame
External infeed
Integral to checkweigher
External infeed
Drop Through/Gravity Install Supported
300D and 300P systems
Tablet/capsule only
Pipeline Install Supported
Package Types Supported
Cartons/Boxes
Pouches/Bags
Bulk Flow Products
Plastic Bottles & Jars (No Metal Lids)
Tablets and Capsules
Construction
Stainless steel,
epoxy
aperture
Stainless steel,
epoxy
aperture
Stainless steel,
epoxy
aperture
Stainless steel,
epoxy
aperture
Stainless steel,
epoxy
aperture
Stainless steel,
epoxy
aperture
User Interface
Graphical, color touchscreen  Graphical, color touchscreen Graphical, color touchscreen
ABS plastic control panel,
display and keypad
ABS plastic control panel,
display and keypad
ABS plastic control panel,
display and keypad
USB Port
Standard
Standard
Standard
Built-In Line Filter
Enhanced Statistics
Frequency Range
Up to five frequencies in real time, adjustable with software in the field(50–1000kHz)  Up to five frequencies in real time, adjustable with software in the field(50–1000kHz) 
A fully-flexible frequency optimized per application, adjustable with software in the field (50 - 1000kHz)
Dual frequencies set in factory (50–460 kHz) Dual frequencies set in factory (50–460 kHz)
Dual frequencies set in factory (700–900 kHz)
Typical Sensitivity
Best, up to 70% smaller volume metal than previous technology
Best, up to 30% smaller volume metal than previous technology
Better, up to 25% smaller volume metal than previous technology
Good
Good
Best
Operating Temperature
–10° to +40°C
(+14° to +104°F)
–10° to +40°C
(+14° to +104°F)
–10° to +40°C
(+14° to +104°F)
–10° to +40°C
(+14° to +104°F)
–10° to +40°C
(+14° to +104°F)
–10° to +40°C
(+14° to +104°F)
Product Temperature Maximum
–10° to 55°C
(14° to 131°F)
–10° to 55°C
(14° to 131°F)
–10° to 55°C
(14° to 131°F)
–10° to 55°C
(14° to 131°F)
–10° to 55°C
(14° to 131°F)
–10° to 55°C
(14° to 131°F)
High temperature option –10° to 120°C (14° to 248°F)
Kynar plastic with metal flange/gasket
Kynar plastic with metal flange/gasket
Belt Speed Range
0.5–1000m/ min
0.5–1000m/
min
0.5–1000m/
min
0.5–1000m/
min
0.5–1000m/
min
0.5–100m/min
Apertures
Wide range
Limited to those used with combos
Wide range
Narrow range
Wide range
Small only
Specific Aperture Sizes
Width: 75 to 2000 mm, Height: 75 to 600 mm. Increments of 25 mm up to 200 mm and 50 mm above 200mm
Width: 100 to 450 mm, Height: 75 to 250 mm. Increments of 25 mm up to 200 mm and 50 mm above 200mm
Width: 75 to 2000 mm, Height: 75 to 600 mm. Increments of 25 mm up to 200 mm and 50 mm above 200mm
Width: 75 to 550 mm, Height: 50 to 550 mm. Increments of 25 mm up to 200 mm and 50 mm above 200mm
Width: 50 to 1750 mm, Height: 50 to 600 mm. Increments of 25 mm up to 200 mm and 50 mm above 200mm
Width: 100 mm, Height: 25, 35 & 50 mm
Ratings
IP69K, HD option includes thermal shock protection
IP69K
IP69K
IP69K, Type 3 enclosure
IP69K, Type 3 enclosure
Type 3 enclosure, metal detector and control panel only
Auditcheck
Relative Price
$$$$
$$$
$$$
$$
$
$$$$
1x1 image pixel for data collection