Archaeological sciences have particular value when it can provide information about the age and origin of archaeological ancient materials and deposits. With use of isotopic fingerprints, one can trace ancient metals and identify different metal deposits, so that metals can be related to specific ore deposits. Also by analyzing human remains on C, N, O, S, P and Sr isotopic signatures, archaeologists can study the dietary habits of an individual or group, determine where a person grew up or where they lived in the last years of their life. In addition, absolute age determinations by, e.g. Re-Os and U-Th isotope systems provide valuable information for archaeologists.
*
*
*
*
*
*
*
*
*
 Please confirm you would like to receive marketing and promotional email messages about Thermo Fisher Scientific products and services.
This site is protected by reCAPTCHA and the Google Privacy Policy and Terms of Service apply.
CMD Form Code v3.0
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
CMD Wide-format style fixes
CMD SchemaApp code