Element Crystal cub
Francium • Alkali Metal
Symbol: Fr
Date of discovery: 1939
Name origin: France
Appearance: unknown
Discoverer: Marguerite Perey
Obtained from: decay of actinium
Melting point: 300 K
Boiling point: 950 K
Density[kg/m3]: unknown
Molar volume: unknown
Protons/Electrons: 87
Neutrons: 136
Shell structure: 2,8,18,32,18,8,1
Electron configuration: [Rn]7s1
Oxidation state: 1
Crystal structure: cubic
While its existence was predicted by Mendeleev in the late nineteenth century, francium was discovered in 1939 by Marguerite Perey of the Curie Institute in Paris. She named the new element after her country, France. It is the last element to be discovered in nature and has the lowest electronegativity and electron affinity of all the elements. Francium is the second rarest element found in the earth’s crust. It is the longest-lived isotope of actinium, with a half life of about 22 minutes.
Products
Nexsa G2 XPS
  • Micro-focus X-ray sources
  • Unique multi-technique options
  • Dual-mode ion source for monoatomic & cluster ion depth profiling
Style Sheet for Instrument Cards Original
K-Alpha XPS
  • High resolution XPS
  • Fast, efficient, automated workflow
  • Ion source for depth profiling
ESCALAB QXi XPS
  • High spectral resolution
  • Multi-technique surface analysis
  • Extensive sample preparation and expansion options
Contact us
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Style Sheet for Global Design System
Style Sheet for Komodo Tabs
Style Sheet to change H3 to p with em-h3-header class
Style Sheet to change H2 style to p with em-h2-header class
Style Sheet for Products Table Specifications
Fix for left nav
Style Sheet for Support and Service footer
Style Sheet for Fonts
Style Sheet for Cards
Electron microscopy services for
the materials science
To ensure optimal system performance, we provide you access to a world-class network of field service experts, technical support, and certified spare parts.