Search Thermo Fisher Scientific
Organism Type | Human | Morphology: round cells, single or in clumps, lymphoblast-like Species: human male 19 years old; Tissue: peripheral blood; Tumor: leukemia, T cell; Derived from: a patient with acute lymphoblastic leukemia (ALL) in relapse; Validated by HLA-DP beta: confirmed as human by PCR Depositor: Dr G. Canti, Pharmacology Dept, Milan, Italy Cell line available for distribution Properties: G->A mutation at codon 248 of the p53; p53 is not expressed; CD1+ CD2+ CD3+ CD4+ CD5+ CD6+ CD7+ CD19- EBV-; high level of terminal deoxynucleotidyl transferase |
Tissues | Blood | |
Phenotype | Suspension | |
Primary | no | |
Application | Transfection: Gene Expression |
Maintaining healthy cells is the key to experimental success and reproducible research results. To give you confidence in the health of your cells every step of the way, we’ve highlighted the technologies and products within cell biology that are critical to maintaining optimal cell health. No matter how you are using your cells, you can count on these products to help keep them healthy.
|
|
|
| |||