Digest-All 4 (Proteinase K)
Product Image
Invitrogen™

Digest-All 4 (Proteinase K)

Have Questions?
Catalog NumberQuantity
003011
also known as 00-3011
50 mL
Catalog number 003011
also known as 00-3011
Price (USD)
343.00
Each
-
Add to cart
Quantity:
50 mL
Price (USD)
343.00
Each
Add to cart
Ask our AI about this Product
This product may be used to pretreat formalin-fixed, paraffin-embedded tissues (see Invitrogen Anatomical Pathology catalog page 140), and will improve the signal of certain antigens. It contains 50 mL of ready-to-use Proteinase K. Digest-All™ 4 is stable for 12 months.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Recommended StorageStore in refrigerator (2–8°C).
Quantity50 mL
Unit SizeEach
Have questions about this product? Ask our AI assisted search.
Generating response
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Los clientes que vieron este artículo también vieron



Documents & Downloads

Certificates

Lot #Certificate TypeDateCatalog Number(s)
3064059Certificate of AnalysisNov 26, 2024003011
3036059Certificate of AnalysisOct 03, 2024003011
2993861Certificate of AnalysisAug 13, 2024003011
2810800Certificate of AnalysisJan 09, 2024003011
2804410Certificate of AnalysisNov 10, 2023003011
5 results displayed, search above for a specific certificate

Safety Data Sheets

Share catalog number, name or link

1x1 image pixel for data collection