Thermo Scientific Chemicals

1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals

製品番号(カタログ番号): 022913.M1
4 x 1 L, Each
Thermo Scientific Chemicals

1,4-Dioxane, HPLC Grade, 99% min, Thermo Scientific Chemicals

製品番号(カタログ番号): 022913.M1
4 x 1 L, Each
数量
製品番号(カタログ番号): 022913.M1
または、製品番号022913-M1

化学物質識別子

CAS
123-91-1
IUPAC Name
1,4-dioxane
Molecular Formula
C4H8O2
InChI Key
RYHBNJHYFVUHQT-UHFFFAOYSA-N
SMILES
C1COCCO1
Residual water
0.03%
Residue on Evaporation
5ppm

概要

1,4-Dioxane HPLC Grade is used as a mobile phase in high performance liquid chromatography and liquid chromatography coupled with mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

ドキュメントおよびダウンロード

証明書

    よくあるご質問(FAQ)

    引用および参考文献

    Search citations by name, author, journal title or abstract text