Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Increase productivity and efficiency using the Thermo Scientific™ AutoEXEC 32-Run Expandable Gas and Liquid Flow Computer. It is the industry's first flow computer that measures natural gas and petroleum liquids simultaneously. This rugged, easy-to-use system calculates flow at a rate of up to 10 times per second on four to 32 runs to reduce capital expenditures.
Product Type | 32-Run Gas and Liquid Flow Computer |
Enclosure Rating | NEMA 4X |
Data Memory | 2 MB RAM, battery-backed plus 4 MB non-volatile serial flash |
Input Power | 90/250 VAC, 9/30 VDC |
Live Density Input | UGC, 4 to 20 mA |
Output Power | 12 VDC |
Program Memory | 6 MB Flash |
Additional Factors/Equations | FWV (manual); FWV (partial); FWV (full); FWS |
Audit Trails | 200 alarm events, 100 different types of alarms |
Certifications/Compliance | Class I Div 2 (Groups and CD), ATEX zone 2 for hazardous area locations, CE, GOST (pending) |
Communication Ports | CPU Board: 2 RS232, 2 Ethernet, 1 Slave USB |
Data Storage | 65 days of daily, 35 days of hourly |
Depth (English) Card Cage | 10.62 in. |
Depth (English) Cover | 12.68 in. |
Depth (Metric) Card Cage | 270 mm |
Depth (Metric) Cover | 319.28 mm |
Diagnostic | AGA 10 SoS |
Differential Meters | AGA 3/ASTM 2530–1992; AGA 3–1985; GOST; V-Cone; Annubar; Slotted-DP |
Display | 4 x 16 character LCD |
Energy | AGA 5 |
Ethylene Density Calculation | API 2565 (Ch 11.3.2.1); Ethylene NBS 1045 |
FCC Compliance | Complies with the limits for a Class A computing device with Part 15 of the FCC rules |
Height (English) | 10.5 in. |
Height (English) Card Cage | 10.5 in. |
Height (English) Cover | 10.5 in. |
Height (Metric) | 266.7 mm |
Height (Metric) Card Cage | 266.7 mm |
Keypad | 4 x 4 (16 key) input, metal keypad button, calculator configuration |
Linear Meters | AGA 7; AGA 9 |
No. of Alarms | 200 alarm events, 23 different types of alarms |
Operating Humidity Range | 0 to 95% RH, Non-condensing |
Operating Temperature Range | -40°C to +85°C (-40°F to +185°F) |
Processor | 32 bit FreescaleTM Coldfire™, operates at 200MHz, processes at 300 million instructions per second (MIPS) |
Propylene Density Calculation | API Ch 11.3.3.2 |
Supercompressibility | AGA 8 Gross-1992; AGA 8 Detail-1992; AGA 8 Short-1985; NX-19; NX-19 Analysis; GERG |
Turbine Meter Linearization | 10-Point Frq/K-Factor Table |
Volume Correction Factor | Consistent with API 2540/ASTM D1250-80/IP 200; 5/6 A/B; 23/24 A/B; 53/54 A/B; 6/24/54 C; CH 11.1 2004; Note: natural gas liquids (NGL) and liquefied petroleum gases (LPG): OLD 23/24, OLD 53/54; Table E is new standard to replace OLD 23/24 |
Width (English) | 17.25 in. |
Width (English) Card Cage | 17 in. |
Width (Metric) | 438.15 mm |
Width (Metric) Card Cage | 431.8 mm |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Product Type | Price (USD) | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
10124435 | Each | 32-Run Gas and Liquid Flow Computer | Request A Quote | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|